Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 194687105-194687330 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM43A
Entrez Gene ID
131583
Gene Name
family with sequence similarity 43 member A
Gene Aliases
-
UniGene
Hs.435080
Gene Chromosome Location
Chr.3: 194685893-194689037 on Build GRCh38
Transcripts
NM_153690.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4116205 c.308G>A p.G103D Chr.3: 194687134-194687134 on Build GRCh38
4996321 c.370G>A p.E124K Chr.3: 194687196-194687196 on Build GRCh38
6420605 c.395G>C p.G132A Chr.3: 194687221-194687221 on Build GRCh38
5974705 c.309C>T p.G103G Chr.3: 194687135-194687135 on Build GRCh38
5864153 c.443G>A p.R148Q Chr.3: 194687269-194687269 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761289488 Chr.3: 194687243-194687243 on Build GRCh38
rs752031180 Chr.3: 194687267-194687267 on Build GRCh38
rs140783529 Chr.3: 194687249-194687249 on Build GRCh38
rs778114503 Chr.3: 194687286-194687286 on Build GRCh38
rs758077922 Chr.3: 194687144-194687144 on Build GRCh38
rs1002687759 Chr.3: 194687208-194687208 on Build GRCh38
rs199570017 Chr.3: 194687192-194687192 on Build GRCh38
rs768342224 Chr.3: 194687218-194687218 on Build GRCh38
rs776158491 Chr.3: 194687221-194687221 on Build GRCh38
rs756228435 Chr.3: 194687168-194687168 on Build GRCh38
rs756353665 Chr.3: 194687274-194687274 on Build GRCh38
rs1041625524 Chr.3: 194687194-194687194 on Build GRCh38
rs528918225 Chr.3: 194687244-194687244 on Build GRCh38
rs112620464 Chr.3: 194687213-194687213 on Build GRCh38
rs907046142 Chr.3: 194687197-194687197 on Build GRCh38
rs766026930 Chr.3: 194687258-194687258 on Build GRCh38
rs370376419 Chr.3: 194687163-194687163 on Build GRCh38
rs760189155 Chr.3: 194687214-194687214 on Build GRCh38
rs750266265 Chr.3: 194687141-194687141 on Build GRCh38
rs201243224 Chr.3: 194687166-194687166 on Build GRCh38
rs765127611 Chr.3: 194687140-194687140 on Build GRCh38
rs771610085 Chr.3: 194687182-194687182 on Build GRCh38
rs142628946 Chr.3: 194687147-194687147 on Build GRCh38
rs1052663311 Chr.3: 194687177-194687177 on Build GRCh38
rs762808685 Chr.3: 194687256-194687256 on Build GRCh38
rs775214286 Chr.3: 194687203-194687203 on Build GRCh38
rs143158704 Chr.3: 194687270-194687270 on Build GRCh38
rs749370333 Chr.3: 194687181-194687181 on Build GRCh38
rs561543874 Chr.3: 194687202-194687202 on Build GRCh38
rs1050946673 Chr.3: 194687216-194687216 on Build GRCh38
rs757242906 Chr.3: 194687136-194687136 on Build GRCh38
rs945992577 Chr.3: 194687185-194687185 on Build GRCh38
rs778053297 Chr.3: 194687174-194687174 on Build GRCh38

View Full Product Details