Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 37707355-37707628 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FRMPD1
Entrez Gene ID
22844
Gene Name
FERM and PDZ domain containing 1
Gene Aliases
FRMD2
UniGene
Hs.163990
Gene Chromosome Location
Chr.9: 37602979-37746904 on Build GRCh38
Transcripts
NM_014907.2 XM_011517803.2 XM_011517804.2 XM_011517805.2 XM_017014480.1 XM_017014482.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3657401 c.135G>A p.R45R Chr.9: 37707449-37707449 on Build GRCh38
3657402 c.156C>T p.I52I Chr.9: 37707470-37707470 on Build GRCh38
2775747 c.116C>A p.A39E Chr.9: 37707430-37707430 on Build GRCh38
4599322 c.163C>T p.R55* Chr.9: 37707477-37707477 on Build GRCh38
5027072 c.222T>A p.S74S Chr.9: 37707536-37707536 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761695738 Chr.9: 37707399-37707399 on Build GRCh38
rs746495979 Chr.9: 37707467-37707467 on Build GRCh38
rs762817179 Chr.9: 37707492-37707492 on Build GRCh38
rs764280280 Chr.9: 37707578-37707578 on Build GRCh38
rs750395582 Chr.9: 37707424-37707424 on Build GRCh38
rs539493025 Chr.9: 37707429-37707429 on Build GRCh38
rs773099173 Chr.9: 37707486-37707486 on Build GRCh38
rs368412884 Chr.9: 37707512-37707512 on Build GRCh38
rs752733863 Chr.9: 37707570-37707570 on Build GRCh38
rs767590379 Chr.9: 37707567-37707567 on Build GRCh38
rs141611918 Chr.9: 37707505-37707505 on Build GRCh38
rs139243880 Chr.9: 37707443-37707443 on Build GRCh38
rs138301491 Chr.9: 37707452-37707452 on Build GRCh38
rs754221636 Chr.9: 37707589-37707589 on Build GRCh38
rs776607210 Chr.9: 37707398-37707398 on Build GRCh38
rs754153341 Chr.9: 37707516-37707516 on Build GRCh38
rs752234880 Chr.9: 37707490-37707493 on Build GRCh38
rs759771040 Chr.9: 37707388-37707388 on Build GRCh38
rs139830645 Chr.9: 37707478-37707478 on Build GRCh38
rs78006962 Chr.9: 37707575-37707575 on Build GRCh38
rs765683914 Chr.9: 37707488-37707488 on Build GRCh38
rs7031966 Chr.9: 37707463-37707463 on Build GRCh38
rs751634587 Chr.9: 37707504-37707504 on Build GRCh38
rs766314187 Chr.9: 37707493-37707493 on Build GRCh38
rs542144899 Chr.9: 37707558-37707558 on Build GRCh38
rs531882907 Chr.9: 37707531-37707531 on Build GRCh38
rs112589819 Chr.9: 37707449-37707449 on Build GRCh38
rs371289474 Chr.9: 37707601-37707601 on Build GRCh38
rs149646564 Chr.9: 37707456-37707456 on Build GRCh38
rs2296556 Chr.9: 37707444-37707444 on Build GRCh38
rs375020833 Chr.9: 37707423-37707423 on Build GRCh38
rs761712793 Chr.9: 37707497-37707497 on Build GRCh38
rs747829777 Chr.9: 37707537-37707537 on Build GRCh38
rs969189967 Chr.9: 37707498-37707498 on Build GRCh38
rs774999716 Chr.9: 37707383-37707383 on Build GRCh38
rs751605302 Chr.9: 37707442-37707442 on Build GRCh38
rs2296557 Chr.9: 37707395-37707395 on Build GRCh38
rs752933775 Chr.9: 37707510-37707510 on Build GRCh38
rs766396709 Chr.9: 37707441-37707441 on Build GRCh38
rs73445151 Chr.9: 37707521-37707521 on Build GRCh38
rs760911278 Chr.9: 37707577-37707577 on Build GRCh38
rs765321276 Chr.9: 37707420-37707420 on Build GRCh38
rs776407042 Chr.9: 37707471-37707471 on Build GRCh38
rs572307245 Chr.9: 37707477-37707477 on Build GRCh38
rs138252723 Chr.9: 37707520-37707520 on Build GRCh38
rs202041675 Chr.9: 37707507-37707507 on Build GRCh38
rs150458691 Chr.9: 37707451-37707451 on Build GRCh38
rs768259137 Chr.9: 37707470-37707470 on Build GRCh38
rs768223771 Chr.9: 37707525-37707525 on Build GRCh38
rs759337102 Chr.9: 37707565-37707565 on Build GRCh38
rs770842730 Chr.9: 37707555-37707555 on Build GRCh38
rs866932127 Chr.9: 37707546-37707546 on Build GRCh38
rs889384165 Chr.9: 37707574-37707574 on Build GRCh38
rs762781500 Chr.9: 37707552-37707552 on Build GRCh38
rs749802152 Chr.9: 37707461-37707461 on Build GRCh38
rs769520391 Chr.9: 37707539-37707539 on Build GRCh38

View Full Product Details