Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 227246568-227246841 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL4A3
Entrez Gene ID
1285
Gene Name
collagen type IV alpha 3 chain
Gene Aliases
-
UniGene
Hs.570065
Gene Chromosome Location
Chr.2: 227164565-227314792 on Build GRCh38
Transcripts
NM_000091.4 XM_005246277.3 XM_005246280.3 XM_006712245.3 XM_011510555.1 XM_017003295.1 NR_102371.1

Gene Symbol
LOC654841
Entrez Gene ID
654841
Gene Name
uncharacterized LOC654841
Gene Aliases
-
UniGene
Hs.659982
Gene Chromosome Location
Chr.2: 227221052-227325164 on Build GRCh38
Transcripts
NM_000091.4 XM_005246277.3 XM_005246280.3 XM_006712245.3 XM_011510555.1 XM_017003295.1 NR_102371.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4855413 c.440C>T p.P147L Chr.2: 227246737-227246737 on Build GRCh38
4855412 c.440C>T p.P147L Chr.2: 227246737-227246737 on Build GRCh38
1669818 c.434G>T p.G145V Chr.2: 227246731-227246731 on Build GRCh38
5601291 c.435G>A p.G145G Chr.2: 227246732-227246732 on Build GRCh38
6327707 c.424G>T p.G142C Chr.2: 227246721-227246721 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200909738 Chr.2: 227246651-227246651 on Build GRCh38
rs573893388 Chr.2: 227246717-227246717 on Build GRCh38
rs772893456 Chr.2: 227246791-227246791 on Build GRCh38
rs748850089 Chr.2: 227246750-227246750 on Build GRCh38
rs759949369 Chr.2: 227246656-227246658 on Build GRCh38
rs757421963 Chr.2: 227246726-227246726 on Build GRCh38
rs559126084 Chr.2: 227246720-227246720 on Build GRCh38
rs777243346 Chr.2: 227246643-227246643 on Build GRCh38
rs773953946 Chr.2: 227246768-227246768 on Build GRCh38
rs766279179 Chr.2: 227246710-227246710 on Build GRCh38
rs774119052 Chr.2: 227246677-227246677 on Build GRCh38
rs761720322 Chr.2: 227246602-227246602 on Build GRCh38
rs75683214 Chr.2: 227246696-227246696 on Build GRCh38
rs781415019 Chr.2: 227246737-227246737 on Build GRCh38
rs183877521 Chr.2: 227246609-227246609 on Build GRCh38
rs372041612 Chr.2: 227246691-227246691 on Build GRCh38
rs376452150 Chr.2: 227246769-227246769 on Build GRCh38
rs10178458 Chr.2: 227246719-227246719 on Build GRCh38
rs762513527 Chr.2: 227246670-227246670 on Build GRCh38
rs555034953 Chr.2: 227246716-227246716 on Build GRCh38
rs373559251 Chr.2: 227246738-227246738 on Build GRCh38
rs768262083 Chr.2: 227246758-227246758 on Build GRCh38
rs750189238 Chr.2: 227246695-227246695 on Build GRCh38
rs774781634 Chr.2: 227246635-227246635 on Build GRCh38
rs767127367 Chr.2: 227246786-227246786 on Build GRCh38
rs763908601 Chr.2: 227246675-227246675 on Build GRCh38
rs201086447 Chr.2: 227246658-227246658 on Build GRCh38
rs761752842 Chr.2: 227246681-227246681 on Build GRCh38
rs373201683 Chr.2: 227246810-227246810 on Build GRCh38
rs765065879 Chr.2: 227246648-227246648 on Build GRCh38
rs758520877 Chr.2: 227246814-227246814 on Build GRCh38
rs764180248 Chr.2: 227246816-227246816 on Build GRCh38
rs369948212 Chr.2: 227246798-227246798 on Build GRCh38
rs776062774 Chr.2: 227246749-227246749 on Build GRCh38
rs751081959 Chr.2: 227246788-227246788 on Build GRCh38

View Full Product Details