Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 40936098-40936365 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DLL4
Entrez Gene ID
54567
Gene Name
delta like canonical Notch ligand 4
Gene Aliases
AOS6, hdelta2
UniGene
Hs.511076
Gene Chromosome Location
Chr.15: 40929333-40939060 on Build GRCh38
Transcripts
NM_019074.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6403441 c.1310G>A p.C437Y Chr.15: 40936297-40936297 on Build GRCh38
5383242 c.1342C>T p.P448S Chr.15: 40936329-40936329 on Build GRCh38
4464462 c.1335C>T p.A445A Chr.15: 40936322-40936322 on Build GRCh38
1372729 c.1321G>A p.V441I Chr.15: 40936308-40936308 on Build GRCh38
6429291 c.1284C>A p.C428* Chr.15: 40936271-40936271 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374162819 Chr.15: 40936308-40936308 on Build GRCh38
rs373338394 Chr.15: 40936257-40936257 on Build GRCh38
rs779764235 Chr.15: 40936223-40936224 on Build GRCh38
rs750221835 Chr.15: 40936208-40936208 on Build GRCh38
rs535125961 Chr.15: 40936258-40936258 on Build GRCh38
rs374587964 Chr.15: 40936286-40936286 on Build GRCh38
rs889609750 Chr.15: 40936141-40936141 on Build GRCh38
rs760234335 Chr.15: 40936285-40936285 on Build GRCh38
rs575946294 Chr.15: 40936233-40936233 on Build GRCh38
rs776252375 Chr.15: 40936290-40936290 on Build GRCh38
rs751873412 Chr.15: 40936323-40936323 on Build GRCh38
rs780355579 Chr.15: 40936213-40936213 on Build GRCh38
rs753874647 Chr.15: 40936189-40936189 on Build GRCh38
rs758218516 Chr.15: 40936211-40936211 on Build GRCh38
rs929814147 Chr.15: 40936126-40936126 on Build GRCh38
rs761840661 Chr.15: 40936190-40936190 on Build GRCh38
rs370339149 Chr.15: 40936273-40936273 on Build GRCh38
rs771130564 Chr.15: 40936267-40936267 on Build GRCh38
rs751697404 Chr.15: 40936316-40936316 on Build GRCh38
rs752793079 Chr.15: 40936334-40936334 on Build GRCh38
rs762820678 Chr.15: 40936314-40936314 on Build GRCh38
rs781379364 Chr.15: 40936324-40936324 on Build GRCh38
rs765165782 Chr.15: 40936193-40936193 on Build GRCh38
rs751673521 Chr.15: 40936218-40936218 on Build GRCh38
rs368013738 Chr.15: 40936340-40936340 on Build GRCh38
rs371847465 Chr.15: 40936341-40936341 on Build GRCh38
rs778252260 Chr.15: 40936337-40936337 on Build GRCh38
rs750346321 Chr.15: 40936313-40936313 on Build GRCh38
rs556538770 Chr.15: 40936146-40936146 on Build GRCh38
rs376378103 Chr.15: 40936306-40936306 on Build GRCh38
rs758675187 Chr.15: 40936253-40936253 on Build GRCh38
rs962595544 Chr.15: 40936182-40936182 on Build GRCh38
rs376300590 Chr.15: 40936262-40936262 on Build GRCh38
rs370387440 Chr.15: 40936246-40936246 on Build GRCh38
rs1006837437 Chr.15: 40936142-40936142 on Build GRCh38
rs750653634 Chr.15: 40936231-40936231 on Build GRCh38
rs1016842579 Chr.15: 40936145-40936145 on Build GRCh38
rs755422423 Chr.15: 40936335-40936335 on Build GRCh38
rs755141768 Chr.15: 40936220-40936220 on Build GRCh38
rs370793294 Chr.15: 40936307-40936307 on Build GRCh38

View Full Product Details