Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 99956988-99957233 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNMBP
Entrez Gene ID
23268
Gene Name
dynamin binding protein
Gene Aliases
ARHGEF36, TUBA
UniGene
Hs.500771
Gene Chromosome Location
Chr.10: 99875571-100009953 on Build GRCh38
Transcripts
NM_015221.3 XM_006717735.2 XM_011539559.2 NR_024130.2

Gene Symbol
DNMBP-AS1
Entrez Gene ID
100188954
Gene Name
DNMBP antisense RNA 1
Gene Aliases
NCRNA00093, bA287G8.2
UniGene
Hs.656104
Gene Chromosome Location
Chr.10: 99927209-99958998 on Build GRCh38
Transcripts
NM_015221.3 XM_006717735.2 XM_011539559.2 NR_024130.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7199623 c.460A>T p.M154L Chr.10: 99957014-99957014 on Build GRCh38
7149850 c.421C>G p.Q141E Chr.10: 99957053-99957053 on Build GRCh38
6647673 c.411C>T p.S137S Chr.10: 99957063-99957063 on Build GRCh38
5432965 c.481G>C p.D161H Chr.10: 99956993-99956993 on Build GRCh38
4010993 c.400C>A p.H134N Chr.10: 99957074-99957074 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748739600 Chr.10: 99957063-99957063 on Build GRCh38
rs375722443 Chr.10: 99957104-99957104 on Build GRCh38
rs371156348 Chr.10: 99957046-99957046 on Build GRCh38
rs776548346 Chr.10: 99957166-99957166 on Build GRCh38
rs201560883 Chr.10: 99957140-99957140 on Build GRCh38
rs755058310 Chr.10: 99957023-99957023 on Build GRCh38
rs370012463 Chr.10: 99957136-99957136 on Build GRCh38
rs771715835 Chr.10: 99957155-99957155 on Build GRCh38
rs761567960 Chr.10: 99957078-99957078 on Build GRCh38
rs760408417 Chr.10: 99957163-99957163 on Build GRCh38
rs777875614 Chr.10: 99957123-99957123 on Build GRCh38
rs758692399 Chr.10: 99957111-99957111 on Build GRCh38
rs142256001 Chr.10: 99957048-99957048 on Build GRCh38
rs376375839 Chr.10: 99957183-99957183 on Build GRCh38
rs753790207 Chr.10: 99957107-99957107 on Build GRCh38
rs759556841 Chr.10: 99957182-99957182 on Build GRCh38
rs868363517 Chr.10: 99957144-99957144 on Build GRCh38
rs113700467 Chr.10: 99957096-99957096 on Build GRCh38
rs573860858 Chr.10: 99957137-99957137 on Build GRCh38
rs778947567 Chr.10: 99957035-99957035 on Build GRCh38
rs566667971 Chr.10: 99957189-99957189 on Build GRCh38
rs980144796 Chr.10: 99957160-99957160 on Build GRCh38
rs530209398 Chr.10: 99957062-99957062 on Build GRCh38
rs747635325 Chr.10: 99957141-99957141 on Build GRCh38
rs748462462 Chr.10: 99957044-99957044 on Build GRCh38
rs146379907 Chr.10: 99957108-99957108 on Build GRCh38
rs138795130 Chr.10: 99957022-99957022 on Build GRCh38
rs772860432 Chr.10: 99957161-99957161 on Build GRCh38
rs139729649 Chr.10: 99957110-99957110 on Build GRCh38
rs985098351 Chr.10: 99957176-99957176 on Build GRCh38
rs922498924 Chr.10: 99957154-99957154 on Build GRCh38
rs374469305 Chr.10: 99957070-99957070 on Build GRCh38
rs541350785 Chr.10: 99957197-99957197 on Build GRCh38
rs750298221 Chr.10: 99957011-99957011 on Build GRCh38
rs766241031 Chr.10: 99957106-99957106 on Build GRCh38
rs952878688 Chr.10: 99957118-99957118 on Build GRCh38
rs781740819 Chr.10: 99957126-99957126 on Build GRCh38
rs143249554 Chr.10: 99957019-99957019 on Build GRCh38
rs772943085 Chr.10: 99957083-99957083 on Build GRCh38
rs747337569 Chr.10: 99957124-99957124 on Build GRCh38
rs756094335 Chr.10: 99957015-99957015 on Build GRCh38
rs766222713 Chr.10: 99957165-99957165 on Build GRCh38
rs201587745 Chr.10: 99957045-99957045 on Build GRCh38
rs767195437 Chr.10: 99957082-99957082 on Build GRCh38
rs757699328 Chr.10: 99957125-99957125 on Build GRCh38
rs773872765 Chr.10: 99957073-99957073 on Build GRCh38
rs753955196 Chr.10: 99957197-99957198 on Build GRCh38

View Full Product Details