Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 49634719-49634958 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAAF2
Entrez Gene ID
55172
Gene Name
dynein axonemal assembly factor 2
Gene Aliases
C14orf104, CILD10, KTU, PF13
UniGene
Hs.231761
Gene Chromosome Location
Chr.14: 49625174-49635230 on Build GRCh38
Transcripts
NM_001083908.1 NM_018139.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7306262 c.421G>A p.V141I Chr.14: 49634729-49634729 on Build GRCh38
1147296 c.391T>A p.Y131N Chr.14: 49634759-49634759 on Build GRCh38
1369918 c.399delG p.R134fs*45 Chr.14: 49634751-49634751 on Build GRCh38
5848130 c.211C>T p.H71Y Chr.14: 49634939-49634939 on Build GRCh38
4657137 c.396G>A p.A132A Chr.14: 49634754-49634754 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs534863337 Chr.14: 49634935-49634935 on Build GRCh38
rs969552686 Chr.14: 49634788-49634788 on Build GRCh38
rs756604353 Chr.14: 49634765-49634765 on Build GRCh38
rs1043037410 Chr.14: 49634859-49634859 on Build GRCh38
rs914228343 Chr.14: 49634823-49634823 on Build GRCh38
rs1014750801 Chr.14: 49634772-49634772 on Build GRCh38
rs1000776853 Chr.14: 49634877-49634877 on Build GRCh38
rs563306994 Chr.14: 49634821-49634821 on Build GRCh38
rs1002627627 Chr.14: 49634775-49634775 on Build GRCh38
rs899878401 Chr.14: 49634835-49634835 on Build GRCh38
rs932908231 Chr.14: 49634922-49634922 on Build GRCh38
rs980944471 Chr.14: 49634926-49634926 on Build GRCh38
rs745566638 Chr.14: 49634774-49634774 on Build GRCh38
rs774533054 Chr.14: 49634822-49634822 on Build GRCh38
rs931555570 Chr.14: 49634807-49634807 on Build GRCh38
rs997340567 Chr.14: 49634845-49634845 on Build GRCh38
rs888453539 Chr.14: 49634930-49634930 on Build GRCh38
rs919604642 Chr.14: 49634792-49634792 on Build GRCh38
rs1037086952 Chr.14: 49634937-49634937 on Build GRCh38
rs1004740371 Chr.14: 49634931-49634931 on Build GRCh38
rs768581485 Chr.14: 49634814-49634814 on Build GRCh38
rs759580456 Chr.14: 49634746-49634746 on Build GRCh38
rs921529892 Chr.14: 49634898-49634898 on Build GRCh38
rs746522193 Chr.14: 49634790-49634790 on Build GRCh38
rs980425229 Chr.14: 49634791-49634791 on Build GRCh38
rs566365192 Chr.14: 49634900-49634900 on Build GRCh38
rs904534333 Chr.14: 49634872-49634872 on Build GRCh38
rs944251462 Chr.14: 49634829-49634829 on Build GRCh38
rs900049390 Chr.14: 49634738-49634738 on Build GRCh38
rs771875922 Chr.14: 49634776-49634776 on Build GRCh38
rs754170049 Chr.14: 49634770-49634770 on Build GRCh38
rs1048364248 Chr.14: 49634756-49634756 on Build GRCh38
rs1035574559 Chr.14: 49634784-49634784 on Build GRCh38
rs919591102 Chr.14: 49634938-49634938 on Build GRCh38
rs533083141 Chr.14: 49634787-49634787 on Build GRCh38
rs886050529 Chr.14: 49634763-49634763 on Build GRCh38
rs541232616 Chr.14: 49634739-49634739 on Build GRCh38
rs771603303 Chr.14: 49634773-49634773 on Build GRCh38
rs200484042 Chr.14: 49634766-49634766 on Build GRCh38
rs529603226 Chr.14: 49634828-49634828 on Build GRCh38
rs1048899988 Chr.14: 49634924-49634924 on Build GRCh38
rs767758073 Chr.14: 49634880-49634880 on Build GRCh38
rs754367020 Chr.14: 49634904-49634904 on Build GRCh38
rs767631402 Chr.14: 49634757-49634757 on Build GRCh38
rs549781788 Chr.14: 49634818-49634818 on Build GRCh38
rs757622361 Chr.14: 49634915-49634915 on Build GRCh38
rs1041192624 Chr.14: 49634897-49634897 on Build GRCh38
rs549457480 Chr.14: 49634887-49634887 on Build GRCh38
rs956518652 Chr.14: 49634883-49634883 on Build GRCh38
rs759705265 Chr.14: 49634833-49634833 on Build GRCh38
rs1024818468 Chr.14: 49634906-49634906 on Build GRCh38
rs1004283663 Chr.14: 49634768-49634768 on Build GRCh38
rs958625302 Chr.14: 49634785-49634785 on Build GRCh38
rs977164190 Chr.14: 49634863-49634863 on Build GRCh38
rs752703053 Chr.14: 49634886-49634886 on Build GRCh38
rs761666035 Chr.14: 49634819-49634819 on Build GRCh38
rs752795172 Chr.14: 49634762-49634762 on Build GRCh38

View Full Product Details