Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 54398538-54398799 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITGA5
Entrez Gene ID
3678
Gene Name
integrin subunit alpha 5
Gene Aliases
CD49e, FNRA, VLA-5, VLA5A
UniGene
Hs.505654
Gene Chromosome Location
Chr.12: 54395261-54419266 on Build GRCh38
Transcripts
NM_002205.4 NR_120486.1 NR_120487.1

Gene Symbol
LOC102724050
Entrez Gene ID
102724050
Gene Name
uncharacterized LOC102724050
Gene Aliases
-
UniGene
Hs.670941
Gene Chromosome Location
Chr.12: 54353691-54467030 on Build GRCh38
Transcripts
NM_002205.4 NR_120486.1 NR_120487.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
281992 c.2876C>T p.A959V Chr.12: 54398664-54398664 on Build GRCh38
5704500 c.2905C>T p.R969* Chr.12: 54398635-54398635 on Build GRCh38
1562018 c.2938C>T p.R980C Chr.12: 54398602-54398602 on Build GRCh38
3359824 c.2920C>T p.Q974* Chr.12: 54398620-54398620 on Build GRCh38
1362785 c.2842C>T p.R948W Chr.12: 54398698-54398698 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754879532 Chr.12: 54398565-54398565 on Build GRCh38
rs1043237001 Chr.12: 54398696-54398696 on Build GRCh38
rs538642834 Chr.12: 54398634-54398634 on Build GRCh38
rs893534435 Chr.12: 54398718-54398718 on Build GRCh38
rs4016638 Chr.12: 54398610-54398610 on Build GRCh38
rs964852457 Chr.12: 54398774-54398774 on Build GRCh38
rs1006654322 Chr.12: 54398754-54398754 on Build GRCh38
rs145576475 Chr.12: 54398601-54398601 on Build GRCh38
rs765192718 Chr.12: 54398664-54398664 on Build GRCh38
rs762012604 Chr.12: 54398611-54398611 on Build GRCh38
rs757921132 Chr.12: 54398675-54398675 on Build GRCh38
rs549185294 Chr.12: 54398630-54398630 on Build GRCh38
rs767565107 Chr.12: 54398615-54398615 on Build GRCh38
rs752566250 Chr.12: 54398666-54398666 on Build GRCh38
rs370125974 Chr.12: 54398697-54398697 on Build GRCh38
rs576679952 Chr.12: 54398765-54398765 on Build GRCh38
rs184620316 Chr.12: 54398755-54398755 on Build GRCh38
rs748606920 Chr.12: 54398724-54398724 on Build GRCh38
rs774947402 Chr.12: 54398609-54398609 on Build GRCh38
rs747681987 Chr.12: 54398574-54398574 on Build GRCh38
rs773311162 Chr.12: 54398747-54398747 on Build GRCh38
rs773318883 Chr.12: 54398616-54398616 on Build GRCh38
rs150955083 Chr.12: 54398687-54398687 on Build GRCh38
rs775915497 Chr.12: 54398604-54398604 on Build GRCh38
rs754720093 Chr.12: 54398652-54398652 on Build GRCh38
rs779566600 Chr.12: 54398707-54398707 on Build GRCh38
rs577771775 Chr.12: 54398605-54398605 on Build GRCh38
rs909831932 Chr.12: 54398592-54398592 on Build GRCh38
rs1031887294 Chr.12: 54398775-54398775 on Build GRCh38
rs778445958 Chr.12: 54398572-54398572 on Build GRCh38
rs377040337 Chr.12: 54398698-54398698 on Build GRCh38
rs757866345 Chr.12: 54398579-54398579 on Build GRCh38
rs142582287 Chr.12: 54398679-54398679 on Build GRCh38
rs766127225 Chr.12: 54398623-54398623 on Build GRCh38
rs772101538 Chr.12: 54398726-54398726 on Build GRCh38
rs117897179 Chr.12: 54398603-54398603 on Build GRCh38
rs148870761 Chr.12: 54398621-54398621 on Build GRCh38
rs377062655 Chr.12: 54398678-54398678 on Build GRCh38
rs751241594 Chr.12: 54398681-54398681 on Build GRCh38
rs777299350 Chr.12: 54398581-54398581 on Build GRCh38
rs769099725 Chr.12: 54398701-54398701 on Build GRCh38
rs758515446 Chr.12: 54398650-54398650 on Build GRCh38

View Full Product Details