Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 122716622-122716849 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HCAR3
Entrez Gene ID
8843
Gene Name
hydroxycarboxylic acid receptor 3
Gene Aliases
GPR109B, HCA3, HM74, PUMAG, Puma-g
UniGene
Hs.458425
Gene Chromosome Location
Chr.12: 122714756-122716892 on Build GRCh38
Transcripts
NM_006018.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4979699 c.31C>T p.L11L Chr.12: 122716707-122716707 on Build GRCh38
6677840 c.64C>T p.R22* Chr.12: 122716674-122716674 on Build GRCh38
288213 c.9G>A p.R3R Chr.12: 122716729-122716729 on Build GRCh38
2225271 c.105G>A p.G35G Chr.12: 122716633-122716633 on Build GRCh38
6295630 c.10C>A p.H4N Chr.12: 122716728-122716728 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768138411 Chr.12: 122716730-122716730 on Build GRCh38
rs774932390 Chr.12: 122716696-122716696 on Build GRCh38
rs760643532 Chr.12: 122716756-122716756 on Build GRCh38
rs373933634 Chr.12: 122716776-122716776 on Build GRCh38
rs376720686 Chr.12: 122716764-122716764 on Build GRCh38
rs779321079 Chr.12: 122716680-122716680 on Build GRCh38
rs768051285 Chr.12: 122716688-122716690 on Build GRCh38
rs3825155 Chr.12: 122716686-122716686 on Build GRCh38
rs368127235 Chr.12: 122716700-122716700 on Build GRCh38
rs747568632 Chr.12: 122716695-122716695 on Build GRCh38
rs755098499 Chr.12: 122716765-122716765 on Build GRCh38
rs372512150 Chr.12: 122716668-122716668 on Build GRCh38
rs750386853 Chr.12: 122716672-122716672 on Build GRCh38
rs190370423 Chr.12: 122716731-122716731 on Build GRCh38
rs148038408 Chr.12: 122716825-122716825 on Build GRCh38
rs756135211 Chr.12: 122716674-122716674 on Build GRCh38
rs148858491 Chr.12: 122716658-122716658 on Build GRCh38
rs761686251 Chr.12: 122716657-122716657 on Build GRCh38
rs762656705 Chr.12: 122716720-122716720 on Build GRCh38
rs766441103 Chr.12: 122716759-122716759 on Build GRCh38
rs3825159 Chr.12: 122716796-122716796 on Build GRCh38
rs3825156 Chr.12: 122716744-122716744 on Build GRCh38
rs1031388275 Chr.12: 122716797-122716797 on Build GRCh38
rs781730752 Chr.12: 122716784-122716784 on Build GRCh38
rs759984438 Chr.12: 122716674-122716674 on Build GRCh38
rs560288602 Chr.12: 122716679-122716679 on Build GRCh38
rs748402120 Chr.12: 122716681-122716681 on Build GRCh38
rs3825162 Chr.12: 122716827-122716827 on Build GRCh38
rs752986518 Chr.12: 122716770-122716770 on Build GRCh38
rs3825158 Chr.12: 122716793-122716793 on Build GRCh38
rs186655475 Chr.12: 122716642-122716642 on Build GRCh38
rs375551658 Chr.12: 122716643-122716643 on Build GRCh38
rs3825161 Chr.12: 122716826-122716826 on Build GRCh38
rs754057666 Chr.12: 122716675-122716675 on Build GRCh38
rs553165107 Chr.12: 122716757-122716757 on Build GRCh38
rs747515470 Chr.12: 122716777-122716777 on Build GRCh38
rs751372302 Chr.12: 122716645-122716645 on Build GRCh38
rs3825157 Chr.12: 122716791-122716791 on Build GRCh38
rs750335615 Chr.12: 122716755-122716755 on Build GRCh38
rs757899404 Chr.12: 122716781-122716781 on Build GRCh38
rs564699572 Chr.12: 122716741-122716741 on Build GRCh38
rs374235993 Chr.12: 122716693-122716693 on Build GRCh38
rs12229648 Chr.12: 122716769-122716769 on Build GRCh38
rs3825160 Chr.12: 122716807-122716807 on Build GRCh38

View Full Product Details