Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 51010115-51010362 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNG1
Entrez Gene ID
3755
Gene Name
potassium voltage-gated channel modifier subfamily G member 1
Gene Aliases
K13, KCNG, KV6.1, kH2
UniGene
Hs.118695
Gene Chromosome Location
Chr.20: 51003656-51023176 on Build GRCh38
Transcripts
NM_002237.3 XM_006723785.2 XM_011528800.1 XM_011528801.1 XM_011528802.1 XM_011528803.2 XM_011528804.1 XM_011528805.2 XM_011528806.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5707486 c.211G>A p.G71S Chr.20: 51010128-51010128 on Build GRCh38
4694290 c.157C>T p.R53C Chr.20: 51010182-51010182 on Build GRCh38
3547700 c.72C>T p.F24F Chr.20: 51010267-51010267 on Build GRCh38
5744050 c.112G>A p.A38T Chr.20: 51010227-51010227 on Build GRCh38
3693579 c.101C>A p.A34D Chr.20: 51010238-51010238 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778419121 Chr.20: 51010192-51010192 on Build GRCh38
rs759726603 Chr.20: 51010146-51010146 on Build GRCh38
rs765174727 Chr.20: 51010152-51010152 on Build GRCh38
rs988882138 Chr.20: 51010320-51010320 on Build GRCh38
rs533277438 Chr.20: 51010333-51010333 on Build GRCh38
rs913335879 Chr.20: 51010324-51010324 on Build GRCh38
rs751331756 Chr.20: 51010306-51010306 on Build GRCh38
rs773106992 Chr.20: 51010289-51010289 on Build GRCh38
rs985715958 Chr.20: 51010227-51010227 on Build GRCh38
rs374169800 Chr.20: 51010174-51010174 on Build GRCh38
rs760751986 Chr.20: 51010273-51010273 on Build GRCh38
rs772208137 Chr.20: 51010262-51010262 on Build GRCh38
rs548363128 Chr.20: 51010185-51010185 on Build GRCh38
rs780703373 Chr.20: 51010180-51010180 on Build GRCh38
rs750155529 Chr.20: 51010159-51010159 on Build GRCh38
rs759168251 Chr.20: 51010321-51010321 on Build GRCh38
rs762916287 Chr.20: 51010294-51010294 on Build GRCh38
rs779187215 Chr.20: 51010234-51010234 on Build GRCh38
rs766134943 Chr.20: 51010172-51010172 on Build GRCh38
rs919816936 Chr.20: 51010196-51010196 on Build GRCh38
rs747674837 Chr.20: 51010181-51010181 on Build GRCh38
rs746064109 Chr.20: 51010252-51010252 on Build GRCh38
rs754614377 Chr.20: 51010175-51010175 on Build GRCh38
rs774621397 Chr.20: 51010135-51010135 on Build GRCh38
rs767080993 Chr.20: 51010332-51010332 on Build GRCh38
rs947391143 Chr.20: 51010340-51010340 on Build GRCh38
rs372397663 Chr.20: 51010195-51010195 on Build GRCh38
rs766155892 Chr.20: 51010297-51010297 on Build GRCh38
rs768778949 Chr.20: 51010276-51010276 on Build GRCh38
rs775466736 Chr.20: 51010148-51010148 on Build GRCh38
rs200434033 Chr.20: 51010270-51010270 on Build GRCh38
rs371150331 Chr.20: 51010147-51010147 on Build GRCh38
rs758172905 Chr.20: 51010170-51010170 on Build GRCh38
rs536118359 Chr.20: 51010215-51010215 on Build GRCh38
rs761694072 Chr.20: 51010149-51010149 on Build GRCh38
rs771399675 Chr.20: 51010232-51010232 on Build GRCh38

View Full Product Details