Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 219532094-219532323 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASIC4
Entrez Gene ID
55515
Gene Name
acid sensing ion channel subunit family member 4
Gene Aliases
ACCN4, BNAC4
UniGene
Hs.87469
Gene Chromosome Location
Chr.2: 219514170-219538772 on Build GRCh38
Transcripts
NM_182847.2 XM_005246671.2 XM_017004439.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1326397 c.1236+2T>A p.? Chr.2: 219532130-219532130 on Build GRCh38
720608 c.1228G>A p.E410K Chr.2: 219532120-219532120 on Build GRCh38
4091726 c.1234C>T p.R412W Chr.2: 219532126-219532126 on Build GRCh38
7143440 c.1209G>A p.Q403Q Chr.2: 219532101-219532101 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs750769790 Chr.2: 219532126-219532126 on Build GRCh38
rs1038129946 Chr.2: 219532146-219532146 on Build GRCh38
rs371570850 Chr.2: 219532171-219532171 on Build GRCh38
rs759497969 Chr.2: 219532183-219532183 on Build GRCh38
rs776385892 Chr.2: 219532181-219532181 on Build GRCh38
rs200868720 Chr.2: 219532135-219532135 on Build GRCh38
rs756510718 Chr.2: 219532127-219532127 on Build GRCh38
rs1022031899 Chr.2: 219532150-219532150 on Build GRCh38
rs1020282770 Chr.2: 219532195-219532195 on Build GRCh38
rs544029545 Chr.2: 219532186-219532186 on Build GRCh38
rs777615707 Chr.2: 219532169-219532169 on Build GRCh38
rs757985394 Chr.2: 219532294-219532294 on Build GRCh38
rs774414742 Chr.2: 219532185-219532185 on Build GRCh38
rs766790420 Chr.2: 219532196-219532196 on Build GRCh38
rs59761991 Chr.2: 219532148-219532148 on Build GRCh38
rs777526690 Chr.2: 219532302-219532302 on Build GRCh38
rs555094972 Chr.2: 219532140-219532140 on Build GRCh38
rs546484806 Chr.2: 219532262-219532262 on Build GRCh38
rs765130281 Chr.2: 219532184-219532184 on Build GRCh38
rs779630344 Chr.2: 219532282-219532282 on Build GRCh38
rs755480746 Chr.2: 219532274-219532274 on Build GRCh38
rs373384533 Chr.2: 219532189-219532189 on Build GRCh38
rs767798894 Chr.2: 219532122-219532122 on Build GRCh38
rs780774383 Chr.2: 219532209-219532209 on Build GRCh38
rs767672359 Chr.2: 219532188-219532188 on Build GRCh38
rs754205418 Chr.2: 219532270-219532270 on Build GRCh38
rs202019494 Chr.2: 219532166-219532166 on Build GRCh38
rs1001480270 Chr.2: 219532182-219532182 on Build GRCh38
rs374509292 Chr.2: 219532286-219532286 on Build GRCh38
rs923465021 Chr.2: 219532257-219532257 on Build GRCh38
rs747603402 Chr.2: 219532288-219532288 on Build GRCh38
rs746734801 Chr.2: 219532170-219532170 on Build GRCh38
rs747700795 Chr.2: 219532160-219532160 on Build GRCh38
rs149591037 Chr.2: 219532256-219532256 on Build GRCh38
rs769234826 Chr.2: 219532303-219532303 on Build GRCh38
rs754395464 Chr.2: 219532142-219532142 on Build GRCh38
rs757444378 Chr.2: 219532114-219532114 on Build GRCh38

View Full Product Details