Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 29609286-29609531 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD13B
Entrez Gene ID
124930
Gene Name
ankyrin repeat domain 13B
Gene Aliases
-
UniGene
Hs.662164
Gene Chromosome Location
Chr.17: 29589769-29614761 on Build GRCh38
Transcripts
NM_152345.4 XM_005257920.4 XM_011524319.2 XM_011524320.2 XM_011524321.2 XM_017024174.1 XM_017024175.1 XM_017024176.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
309132 c.781C>T p.R261C Chr.17: 29609380-29609380 on Build GRCh38
1381854 c.777C>A p.G259G Chr.17: 29609376-29609376 on Build GRCh38
4065197 c.794C>T p.T265M Chr.17: 29609393-29609393 on Build GRCh38
705748 c.814G>A p.E272K Chr.17: 29609413-29609413 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs771475263 Chr.17: 29609304-29609304 on Build GRCh38
rs146795223 Chr.17: 29609385-29609385 on Build GRCh38
rs747543951 Chr.17: 29609434-29609434 on Build GRCh38
rs944076670 Chr.17: 29609504-29609504 on Build GRCh38
rs369846246 Chr.17: 29609355-29609355 on Build GRCh38
rs549198487 Chr.17: 29609394-29609394 on Build GRCh38
rs770925792 Chr.17: 29609505-29609505 on Build GRCh38
rs569937779 Chr.17: 29609370-29609370 on Build GRCh38
rs117661862 Chr.17: 29609328-29609328 on Build GRCh38
rs773911634 Chr.17: 29609456-29609456 on Build GRCh38
rs201684708 Chr.17: 29609466-29609466 on Build GRCh38
rs777891112 Chr.17: 29609430-29609430 on Build GRCh38
rs747813610 Chr.17: 29609460-29609460 on Build GRCh38
rs758452197 Chr.17: 29609409-29609409 on Build GRCh38
rs774036741 Chr.17: 29609315-29609315 on Build GRCh38
rs887099828 Chr.17: 29609476-29609476 on Build GRCh38
rs781697868 Chr.17: 29609445-29609445 on Build GRCh38
rs374823174 Chr.17: 29609313-29609313 on Build GRCh38
rs933224638 Chr.17: 29609384-29609384 on Build GRCh38
rs535994428 Chr.17: 29609436-29609436 on Build GRCh38
rs779530201 Chr.17: 29609307-29609307 on Build GRCh38
rs567329432 Chr.17: 29609432-29609432 on Build GRCh38
rs374732691 Chr.17: 29609451-29609451 on Build GRCh38
rs747799243 Chr.17: 29609343-29609343 on Build GRCh38
rs199786725 Chr.17: 29609393-29609393 on Build GRCh38
rs529080158 Chr.17: 29609320-29609320 on Build GRCh38
rs771504698 Chr.17: 29609465-29609465 on Build GRCh38
rs374046551 Chr.17: 29609448-29609448 on Build GRCh38
rs142904329 Chr.17: 29609314-29609314 on Build GRCh38
rs114355910 Chr.17: 29609467-29609467 on Build GRCh38
rs371863320 Chr.17: 29609323-29609323 on Build GRCh38
rs372062091 Chr.17: 29609339-29609339 on Build GRCh38

View Full Product Details