Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 31399153-31399404 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DSG1-AS1
Entrez Gene ID
101927718
Gene Name
DSG1 antisense RNA 1
Gene Aliases
-
UniGene
Hs.625473
Gene Chromosome Location
Chr.18: 31343368-31426988 on Build GRCh38
Transcripts
NR_110788.1 NM_001134453.2 NM_177986.4

Gene Symbol
DSG4
Entrez Gene ID
147409
Gene Name
desmoglein 4
Gene Aliases
CDGF13, CDHF13, HYPT6, LAH
UniGene
Hs.407618
Gene Chromosome Location
Chr.18: 31376777-31415791 on Build GRCh38
Transcripts
NR_110788.1 NM_001134453.2 NM_177986.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6083120 c.1038G>T p.Q346H Chr.18: 31399304-31399304 on Build GRCh38
708113 c.1024G>A p.A342T Chr.18: 31399290-31399290 on Build GRCh38
6083122 c.1027C>T p.P343S Chr.18: 31399293-31399293 on Build GRCh38
5741235 c.1016A>G p.Y339C Chr.18: 31399282-31399282 on Build GRCh38
5603434 c.1049G>A p.G350E Chr.18: 31399315-31399315 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754093240 Chr.18: 31399278-31399278 on Build GRCh38
rs1015625692 Chr.18: 31399343-31399343 on Build GRCh38
rs76349777 Chr.18: 31399368-31399368 on Build GRCh38
rs767929125 Chr.18: 31399254-31399254 on Build GRCh38
rs949200320 Chr.18: 31399216-31399216 on Build GRCh38
rs768184020 Chr.18: 31399313-31399313 on Build GRCh38
rs148911860 Chr.18: 31399294-31399294 on Build GRCh38
rs779728387 Chr.18: 31399296-31399296 on Build GRCh38
rs368181590 Chr.18: 31399222-31399222 on Build GRCh38
rs576113919 Chr.18: 31399224-31399224 on Build GRCh38
rs112376128 Chr.18: 31399291-31399291 on Build GRCh38
rs762395269 Chr.18: 31399336-31399336 on Build GRCh38
rs766803039 Chr.18: 31399351-31399351 on Build GRCh38
rs772511697 Chr.18: 31399340-31399340 on Build GRCh38
rs140183798 Chr.18: 31399208-31399208 on Build GRCh38
rs545724872 Chr.18: 31399182-31399182 on Build GRCh38
rs937914148 Chr.18: 31399200-31399200 on Build GRCh38
rs757089004 Chr.18: 31399287-31399287 on Build GRCh38
rs192864615 Chr.18: 31399355-31399355 on Build GRCh38
rs998283060 Chr.18: 31399345-31399345 on Build GRCh38
rs750695206 Chr.18: 31399255-31399255 on Build GRCh38
rs748887910 Chr.18: 31399302-31399302 on Build GRCh38
rs752781605 Chr.18: 31399378-31399378 on Build GRCh38
rs979394777 Chr.18: 31399187-31399187 on Build GRCh38
rs773738568 Chr.18: 31399342-31399342 on Build GRCh38
rs976499394 Chr.18: 31399221-31399221 on Build GRCh38
rs200590887 Chr.18: 31399346-31399346 on Build GRCh38
rs1055430710 Chr.18: 31399201-31399201 on Build GRCh38
rs1028386849 Chr.18: 31399360-31399360 on Build GRCh38
rs774942514 Chr.18: 31399247-31399247 on Build GRCh38
rs766747911 Chr.18: 31399261-31399261 on Build GRCh38
rs759768071 Chr.18: 31399365-31399365 on Build GRCh38
rs372942507 Chr.18: 31399347-31399347 on Build GRCh38
rs756498035 Chr.18: 31399256-31399256 on Build GRCh38
rs925317178 Chr.18: 31399191-31399191 on Build GRCh38
rs762185432 Chr.18: 31399249-31399249 on Build GRCh38
rs369018054 Chr.18: 31399283-31399283 on Build GRCh38
rs773666252 Chr.18: 31399314-31399314 on Build GRCh38
rs751522954 Chr.18: 31399284-31399284 on Build GRCh38
rs901160093 Chr.18: 31399344-31399344 on Build GRCh38
rs763569095 Chr.18: 31399241-31399241 on Build GRCh38
rs530136665 Chr.18: 31399367-31399367 on Build GRCh38
rs781180015 Chr.18: 31399288-31399288 on Build GRCh38

View Full Product Details