Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 189695088-189695321 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKAR
Entrez Gene ID
150709
Gene Name
ankyrin and armadillo repeat containing
Gene Aliases
-
UniGene
Hs.60772
Gene Chromosome Location
Chr.2: 189674292-189763059 on Build GRCh38
Transcripts
NM_144708.3 XM_011510673.2 XM_011510674.1 XM_011510675.2 XM_011510676.2 XM_011510677.2 XM_011510679.2 XM_011510680.1 XM_011510681.1 XM_011510682.2 XM_011510685.2 XM_011510686.2 XM_011510687.2 XM_011510688.1 XM_011510689.2 XM_011510691.2 XM_011510692.2 XM_011510693.2 XM_011510694.2 XM_017003413.1 XM_017003414.1 XM_017003415.1 XM_017003416.1 XM_017003417.1 XM_017003418.1 XM_017003419.1 XM_017003420.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2712265 c.1423A>T p.T475S Chr.2: 189695096-189695096 on Build GRCh38
3838143 c.1426G>A p.D476N Chr.2: 189695099-189695099 on Build GRCh38
3838144 c.1213G>A p.D405N Chr.2: 189695099-189695099 on Build GRCh38
5914856 c.1265T>A p.M422K Chr.2: 189695151-189695151 on Build GRCh38
2712266 c.1210A>T p.T404S Chr.2: 189695096-189695096 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748469653 Chr.2: 189695178-189695179 on Build GRCh38
rs766157830 Chr.2: 189695146-189695146 on Build GRCh38
rs747259533 Chr.2: 189695203-189695203 on Build GRCh38
rs753805105 Chr.2: 189695153-189695153 on Build GRCh38
rs536908302 Chr.2: 189695263-189695263 on Build GRCh38
rs777000137 Chr.2: 189695131-189695131 on Build GRCh38
rs575210177 Chr.2: 189695189-189695189 on Build GRCh38
rs79029877 Chr.2: 189695209-189695209 on Build GRCh38
rs141464406 Chr.2: 189695120-189695120 on Build GRCh38
rs372066044 Chr.2: 189695171-189695171 on Build GRCh38
rs751279086 Chr.2: 189695162-189695162 on Build GRCh38
rs768257928 Chr.2: 189695113-189695113 on Build GRCh38
rs765270498 Chr.2: 189695163-189695163 on Build GRCh38
rs1009148079 Chr.2: 189695229-189695229 on Build GRCh38
rs995129929 Chr.2: 189695191-189695191 on Build GRCh38
rs867365300 Chr.2: 189695148-189695148 on Build GRCh38
rs150384782 Chr.2: 189695137-189695137 on Build GRCh38
rs1008545389 Chr.2: 189695223-189695223 on Build GRCh38
rs371045185 Chr.2: 189695170-189695170 on Build GRCh38
rs193293465 Chr.2: 189695233-189695233 on Build GRCh38
rs111666297 Chr.2: 189695154-189695154 on Build GRCh38
rs1048859660 Chr.2: 189695176-189695176 on Build GRCh38
rs781458099 Chr.2: 189695212-189695212 on Build GRCh38
rs910323439 Chr.2: 189695178-189695178 on Build GRCh38
rs375605787 Chr.2: 189695127-189695127 on Build GRCh38
rs767296937 Chr.2: 189695134-189695134 on Build GRCh38
rs901524512 Chr.2: 189695213-189695213 on Build GRCh38
rs772828251 Chr.2: 189695135-189695135 on Build GRCh38

View Full Product Details