Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 26770070-26770337 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADRA1A
Entrez Gene ID
148
Gene Name
adrenoceptor alpha 1A
Gene Aliases
ADRA1C, ADRA1L1, ALPHA1AAR
UniGene
Hs.52931
Gene Chromosome Location
Chr.8: 26738113-26870994 on Build GRCh38
Transcripts
NM_000680.3 NM_001322502.1 NM_001322503.1 NM_033302.3 NM_033303.4 NM_033304.3 NR_136343.1 XM_006716292.3 XM_006716293.3 XM_017013094.1 XM_017013095.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4523461 c.1216G>A p.G406R Chr.8: 26770334-26770334 on Build GRCh38
4896325 c.1222G>A p.A408T Chr.8: 26770328-26770328 on Build GRCh38
238716 c.1387G>A p.G463R Chr.8: 26770163-26770163 on Build GRCh38
1569103 c.1222G>T p.A408S Chr.8: 26770328-26770328 on Build GRCh38
1569101 c.1222G>T p.A408S Chr.8: 26770328-26770328 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766671226 Chr.8: 26770163-26770163 on Build GRCh38
rs779788145 Chr.8: 26770286-26770286 on Build GRCh38
rs146235804 Chr.8: 26770239-26770239 on Build GRCh38
rs762370840 Chr.8: 26770232-26770232 on Build GRCh38
rs866461368 Chr.8: 26770187-26770187 on Build GRCh38
rs761063889 Chr.8: 26770222-26770222 on Build GRCh38
rs777539457 Chr.8: 26770261-26770261 on Build GRCh38
rs776937254 Chr.8: 26770160-26770160 on Build GRCh38
rs769974972 Chr.8: 26770131-26770131 on Build GRCh38
rs774842013 Chr.8: 26770278-26770278 on Build GRCh38
rs777627486 Chr.8: 26770191-26770191 on Build GRCh38
rs376763061 Chr.8: 26770305-26770305 on Build GRCh38
rs908462360 Chr.8: 26770297-26770297 on Build GRCh38
rs528402543 Chr.8: 26770089-26770089 on Build GRCh38
rs751790994 Chr.8: 26770257-26770257 on Build GRCh38
rs759805643 Chr.8: 26770239-26770239 on Build GRCh38
rs958681910 Chr.8: 26770229-26770229 on Build GRCh38
rs2229126 Chr.8: 26770155-26770155 on Build GRCh38
rs759221908 Chr.8: 26770119-26770119 on Build GRCh38
rs3730247 Chr.8: 26770309-26770309 on Build GRCh38
rs771722367 Chr.8: 26770282-26770282 on Build GRCh38
rs775973909 Chr.8: 26770139-26770139 on Build GRCh38
rs367786537 Chr.8: 26770283-26770283 on Build GRCh38
rs373717543 Chr.8: 26770190-26770190 on Build GRCh38
rs372056090 Chr.8: 26770146-26770146 on Build GRCh38
rs200827445 Chr.8: 26770299-26770299 on Build GRCh38
rs763315626 Chr.8: 26770230-26770230 on Build GRCh38
rs777034772 Chr.8: 26770300-26770300 on Build GRCh38
rs576498291 Chr.8: 26770315-26770315 on Build GRCh38
rs867611987 Chr.8: 26770162-26770162 on Build GRCh38
rs201746874 Chr.8: 26770164-26770164 on Build GRCh38
rs747894155 Chr.8: 26770171-26770171 on Build GRCh38
rs957686968 Chr.8: 26770108-26770108 on Build GRCh38
rs767335342 Chr.8: 26770124-26770124 on Build GRCh38
rs150174675 Chr.8: 26770221-26770221 on Build GRCh38
rs769685470 Chr.8: 26770099-26770099 on Build GRCh38
rs372173736 Chr.8: 26770295-26770295 on Build GRCh38
rs770485145 Chr.8: 26770112-26770112 on Build GRCh38
rs1048102 Chr.8: 26770259-26770259 on Build GRCh38
rs142294326 Chr.8: 26770107-26770107 on Build GRCh38
rs773895990 Chr.8: 26770197-26770197 on Build GRCh38
rs761153886 Chr.8: 26770143-26770143 on Build GRCh38
rs903403718 Chr.8: 26770182-26770182 on Build GRCh38
rs770838608 Chr.8: 26770196-26770196 on Build GRCh38
rs757219537 Chr.8: 26770160-26770160 on Build GRCh38
rs757196717 Chr.8: 26770273-26770273 on Build GRCh38
rs758500506 Chr.8: 26770246-26770246 on Build GRCh38
rs61760513 Chr.8: 26770090-26770090 on Build GRCh38
rs745335442 Chr.8: 26770280-26770280 on Build GRCh38
rs749249774 Chr.8: 26770194-26770194 on Build GRCh38
rs138751075 Chr.8: 26770226-26770226 on Build GRCh38
rs137939451 Chr.8: 26770253-26770253 on Build GRCh38
rs769878756 Chr.8: 26770260-26770261 on Build GRCh38
rs753632480 Chr.8: 26770267-26770267 on Build GRCh38
rs747988651 Chr.8: 26770098-26770098 on Build GRCh38
rs765357565 Chr.8: 26770161-26770161 on Build GRCh38
rs773161924 Chr.8: 26770104-26770104 on Build GRCh38
rs765717995 Chr.8: 26770235-26770235 on Build GRCh38
rs778755383 Chr.8: 26770276-26770276 on Build GRCh38
rs776927741 Chr.8: 26770231-26770231 on Build GRCh38
rs773807450 Chr.8: 26770117-26770117 on Build GRCh38
rs771818468 Chr.8: 26770206-26770206 on Build GRCh38

View Full Product Details