Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 65082479-65082740 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP68
Entrez Gene ID
23177
Gene Name
centrosomal protein 68
Gene Aliases
KIAA0582
UniGene
Hs.709257
Gene Chromosome Location
Chr.2: 65056361-65087008 on Build GRCh38
Transcripts
NM_001319100.1 NM_001319101.1 NM_015147.2 NR_134966.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3055265 c.2241G>A p.M747I Chr.2: 65082672-65082672 on Build GRCh38
1022272 c.2130C>T p.I710I Chr.2: 65082561-65082561 on Build GRCh38
4583592 c.2216C>T p.T739I Chr.2: 65082647-65082647 on Build GRCh38
1690618 c.2138A>G p.Q713R Chr.2: 65082569-65082569 on Build GRCh38
3407933 c.2131G>A p.A711T Chr.2: 65082562-65082562 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs560970351 Chr.2: 65082603-65082603 on Build GRCh38
rs762322353 Chr.2: 65082617-65082617 on Build GRCh38
rs777921262 Chr.2: 65082591-65082591 on Build GRCh38
rs757294495 Chr.2: 65082654-65082654 on Build GRCh38
rs773300961 Chr.2: 65082621-65082621 on Build GRCh38
rs745422882 Chr.2: 65082661-65082661 on Build GRCh38
rs763990694 Chr.2: 65082647-65082647 on Build GRCh38
rs759350706 Chr.2: 65082528-65082530 on Build GRCh38
rs760928907 Chr.2: 65082625-65082625 on Build GRCh38
rs929448639 Chr.2: 65082672-65082672 on Build GRCh38
rs778719307 Chr.2: 65082570-65082570 on Build GRCh38
rs779641254 Chr.2: 65082677-65082677 on Build GRCh38
rs141590287 Chr.2: 65082598-65082598 on Build GRCh38
rs370298053 Chr.2: 65082561-65082561 on Build GRCh38
rs201278860 Chr.2: 65082602-65082602 on Build GRCh38
rs543825974 Chr.2: 65082507-65082507 on Build GRCh38
rs752356164 Chr.2: 65082708-65082709 on Build GRCh38
rs767483708 Chr.2: 65082529-65082529 on Build GRCh38
rs774727196 Chr.2: 65082707-65082707 on Build GRCh38
rs749652442 Chr.2: 65082685-65082685 on Build GRCh38
rs772400656 Chr.2: 65082669-65082669 on Build GRCh38
rs752528667 Chr.2: 65082632-65082632 on Build GRCh38
rs751608894 Chr.2: 65082524-65082524 on Build GRCh38
rs893586940 Chr.2: 65082579-65082579 on Build GRCh38
rs763472081 Chr.2: 65082519-65082519 on Build GRCh38
rs1008494196 Chr.2: 65082666-65082666 on Build GRCh38
rs764557987 Chr.2: 65082522-65082522 on Build GRCh38
rs775752708 Chr.2: 65082506-65082506 on Build GRCh38
rs758321362 Chr.2: 65082580-65082580 on Build GRCh38
rs767543442 Chr.2: 65082619-65082619 on Build GRCh38
rs780804376 Chr.2: 65082655-65082655 on Build GRCh38
rs373667566 Chr.2: 65082700-65082700 on Build GRCh38
rs750576074 Chr.2: 65082558-65082558 on Build GRCh38
rs767254642 Chr.2: 65082611-65082612 on Build GRCh38
rs1023389015 Chr.2: 65082594-65082594 on Build GRCh38
rs960126646 Chr.2: 65082575-65082575 on Build GRCh38
rs746501565 Chr.2: 65082592-65082592 on Build GRCh38
rs918838217 Chr.2: 65082670-65082670 on Build GRCh38
rs368398491 Chr.2: 65082690-65082690 on Build GRCh38
rs200403553 Chr.2: 65082631-65082631 on Build GRCh38
rs560205369 Chr.2: 65082676-65082676 on Build GRCh38
rs769202655 Chr.2: 65082611-65082611 on Build GRCh38
rs575479626 Chr.2: 65082595-65082595 on Build GRCh38
rs774897048 Chr.2: 65082612-65082612 on Build GRCh38
rs920881877 Chr.2: 65082658-65082658 on Build GRCh38
rs758181928 Chr.2: 65082646-65082646 on Build GRCh38
rs540340877 Chr.2: 65082674-65082674 on Build GRCh38
rs761949542 Chr.2: 65082528-65082528 on Build GRCh38
rs905408112 Chr.2: 65082549-65082549 on Build GRCh38
rs1046702957 Chr.2: 65082544-65082544 on Build GRCh38
rs751473103 Chr.2: 65082651-65082651 on Build GRCh38
rs767715250 Chr.2: 65082550-65082550 on Build GRCh38
rs775905529 Chr.2: 65082601-65082601 on Build GRCh38
rs1053951511 Chr.2: 65082577-65082577 on Build GRCh38
rs561057413 Chr.2: 65082691-65082691 on Build GRCh38
rs769356766 Chr.2: 65082599-65082599 on Build GRCh38

View Full Product Details