Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 223746944-223747217 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAPN2
Entrez Gene ID
824
Gene Name
calpain 2
Gene Aliases
CANP2, CANPL2, CANPml, mCANP
UniGene
Hs.350899
Gene Chromosome Location
Chr.1: 223701593-223776018 on Build GRCh38
Transcripts
NM_001146068.1 NM_001748.4 XM_011544286.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7359432 c.410A>G p.Y137C Chr.1: 223747080-223747080 on Build GRCh38
904428 c.729+1G>A p.? Chr.1: 223747166-223747166 on Build GRCh38
6061721 c.593G>T p.G198V Chr.1: 223747029-223747029 on Build GRCh38
7359430 c.401C>T p.A134V Chr.1: 223747071-223747071 on Build GRCh38
1960496 c.644A>G p.Y215C Chr.1: 223747080-223747080 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs139316104 Chr.1: 223747048-223747048 on Build GRCh38
rs762612610 Chr.1: 223746971-223746971 on Build GRCh38
rs756736356 Chr.1: 223747162-223747162 on Build GRCh38
rs751937740 Chr.1: 223747019-223747019 on Build GRCh38
rs760291910 Chr.1: 223747170-223747170 on Build GRCh38
rs760130328 Chr.1: 223747065-223747065 on Build GRCh38
rs749225005 Chr.1: 223747037-223747037 on Build GRCh38
rs747747881 Chr.1: 223747175-223747175 on Build GRCh38
rs776289415 Chr.1: 223747102-223747103 on Build GRCh38
rs763969373 Chr.1: 223746972-223746972 on Build GRCh38
rs753271009 Chr.1: 223747005-223747005 on Build GRCh38
rs747935113 Chr.1: 223747163-223747163 on Build GRCh38
rs768514311 Chr.1: 223747038-223747038 on Build GRCh38
rs367715962 Chr.1: 223746978-223746978 on Build GRCh38
rs759453896 Chr.1: 223747059-223747059 on Build GRCh38
rs575386583 Chr.1: 223747165-223747165 on Build GRCh38
rs140704789 Chr.1: 223747143-223747143 on Build GRCh38
rs767224404 Chr.1: 223746986-223746986 on Build GRCh38
rs778728946 Chr.1: 223747153-223747153 on Build GRCh38
rs201961877 Chr.1: 223747080-223747080 on Build GRCh38
rs764599934 Chr.1: 223747188-223747188 on Build GRCh38
rs1037976491 Chr.1: 223747112-223747112 on Build GRCh38
rs147824152 Chr.1: 223747092-223747092 on Build GRCh38
rs749920294 Chr.1: 223746997-223746997 on Build GRCh38
rs767967830 Chr.1: 223747194-223747194 on Build GRCh38
rs549405629 Chr.1: 223747166-223747166 on Build GRCh38
rs6663755 Chr.1: 223746977-223746977 on Build GRCh38
rs753331410 Chr.1: 223747084-223747084 on Build GRCh38
rs765853769 Chr.1: 223747070-223747070 on Build GRCh38
rs780088909 Chr.1: 223747031-223747031 on Build GRCh38
rs747013394 Chr.1: 223747024-223747025 on Build GRCh38
rs780149356 Chr.1: 223747141-223747141 on Build GRCh38
rs990085558 Chr.1: 223747002-223747002 on Build GRCh38
rs770639720 Chr.1: 223747179-223747179 on Build GRCh38
rs769768965 Chr.1: 223747140-223747140 on Build GRCh38
rs757705052 Chr.1: 223747116-223747116 on Build GRCh38
rs541384226 Chr.1: 223747004-223747004 on Build GRCh38
rs571892662 Chr.1: 223747180-223747180 on Build GRCh38
rs774746796 Chr.1: 223747191-223747191 on Build GRCh38
rs758879117 Chr.1: 223747014-223747014 on Build GRCh38
rs761570360 Chr.1: 223747049-223747049 on Build GRCh38
rs750693781 Chr.1: 223747132-223747132 on Build GRCh38
rs768050477 Chr.1: 223747024-223747024 on Build GRCh38
rs771653698 Chr.1: 223747060-223747060 on Build GRCh38
rs772882922 Chr.1: 223747061-223747061 on Build GRCh38
rs757492985 Chr.1: 223747020-223747020 on Build GRCh38
rs751897611 Chr.1: 223747111-223747111 on Build GRCh38
rs779819965 Chr.1: 223746985-223746985 on Build GRCh38
rs762216620 Chr.1: 223747193-223747193 on Build GRCh38
rs758986562 Chr.1: 223747184-223747184 on Build GRCh38
rs17600 Chr.1: 223747018-223747018 on Build GRCh38
rs761588429 Chr.1: 223747021-223747021 on Build GRCh38
rs781521726 Chr.1: 223747127-223747127 on Build GRCh38
rs770020884 Chr.1: 223747029-223747029 on Build GRCh38
rs764569987 Chr.1: 223747100-223747100 on Build GRCh38

View Full Product Details