Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 47658588-47658861 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C7orf65
Entrez Gene ID
401335
Gene Name
chromosome 7 open reading frame 65
Gene Aliases
-
UniGene
Hs.520816
Gene Chromosome Location
Chr.7: 47655244-47661648 on Build GRCh38
Transcripts
NM_001123065.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3881390 c.132G>T p.K44N Chr.7: 47658754-47658754 on Build GRCh38
3995616 c.105G>A p.L35L Chr.7: 47658727-47658727 on Build GRCh38
1090127 c.134G>T p.R45I Chr.7: 47658756-47658756 on Build GRCh38
4521403 c.110G>A p.R37K Chr.7: 47658732-47658732 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs752953856 Chr.7: 47658673-47658673 on Build GRCh38
rs748422938 Chr.7: 47658700-47658702 on Build GRCh38
rs751554140 Chr.7: 47658654-47658654 on Build GRCh38
rs759596272 Chr.7: 47658749-47658749 on Build GRCh38
rs766588703 Chr.7: 47658643-47658643 on Build GRCh38
rs778084309 Chr.7: 47658678-47658678 on Build GRCh38
rs111962078 Chr.7: 47658687-47658687 on Build GRCh38
rs577969927 Chr.7: 47658627-47658627 on Build GRCh38
rs777313070 Chr.7: 47658837-47658837 on Build GRCh38
rs779566434 Chr.7: 47658688-47658688 on Build GRCh38
rs372605213 Chr.7: 47658774-47658774 on Build GRCh38
rs769751001 Chr.7: 47658733-47658733 on Build GRCh38
rs967212957 Chr.7: 47658685-47658685 on Build GRCh38
rs749846255 Chr.7: 47658686-47658686 on Build GRCh38
rs779617559 Chr.7: 47658701-47658701 on Build GRCh38
rs370755765 Chr.7: 47658797-47658797 on Build GRCh38
rs747832234 Chr.7: 47658730-47658730 on Build GRCh38
rs373837942 Chr.7: 47658640-47658640 on Build GRCh38
rs780721704 Chr.7: 47658798-47658798 on Build GRCh38
rs768366879 Chr.7: 47658712-47658712 on Build GRCh38
rs529375828 Chr.7: 47658751-47658751 on Build GRCh38
rs757872314 Chr.7: 47658833-47658833 on Build GRCh38
rs767577967 Chr.7: 47658672-47658672 on Build GRCh38
rs776202943 Chr.7: 47658723-47658723 on Build GRCh38
rs369106786 Chr.7: 47658696-47658696 on Build GRCh38
rs992732636 Chr.7: 47658835-47658835 on Build GRCh38
rs766354417 Chr.7: 47658744-47658744 on Build GRCh38
rs560113141 Chr.7: 47658747-47658747 on Build GRCh38
rs762952478 Chr.7: 47658642-47658642 on Build GRCh38
rs763015648 Chr.7: 47658743-47658743 on Build GRCh38
rs756540179 Chr.7: 47658675-47658675 on Build GRCh38
rs759457006 Chr.7: 47658756-47658756 on Build GRCh38
rs537031631 Chr.7: 47658625-47658625 on Build GRCh38
rs752906597 Chr.7: 47658755-47658755 on Build GRCh38
rs773093190 Chr.7: 47658742-47658742 on Build GRCh38
rs897969129 Chr.7: 47658764-47658764 on Build GRCh38
rs758679456 Chr.7: 47658739-47658740 on Build GRCh38
rs757763962 Chr.7: 47658789-47658789 on Build GRCh38
rs957851480 Chr.7: 47658615-47658615 on Build GRCh38
rs1033976378 Chr.7: 47658816-47658816 on Build GRCh38
rs549050631 Chr.7: 47658783-47658783 on Build GRCh38
rs375966382 Chr.7: 47658791-47658791 on Build GRCh38
rs755235396 Chr.7: 47658667-47658667 on Build GRCh38
rs979763199 Chr.7: 47658800-47658800 on Build GRCh38
rs781147633 Chr.7: 47658741-47658742 on Build GRCh38
rs779227909 Chr.7: 47658790-47658790 on Build GRCh38

View Full Product Details