Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 17301494-17301719 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABHD8
Entrez Gene ID
79575
Gene Name
abhydrolase domain containing 8
Gene Aliases
-
UniGene
Hs.515664
Gene Chromosome Location
Chr.19: 17292131-17303473 on Build GRCh38
Transcripts
NM_024527.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
992757 c.57C>T p.N19N Chr.19: 17301560-17301560 on Build GRCh38
7233225 c.27C>T p.I9I Chr.19: 17301590-17301590 on Build GRCh38
7251111 c.47C>T p.T16M Chr.19: 17301570-17301570 on Build GRCh38
4406590 c.14T>G p.V5G Chr.19: 17301603-17301603 on Build GRCh38
7277462 c.36C>A p.C12* Chr.19: 17301581-17301581 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769538954 Chr.19: 17301606-17301606 on Build GRCh38
rs762989608 Chr.19: 17301664-17301664 on Build GRCh38
rs771916946 Chr.19: 17301569-17301569 on Build GRCh38
rs561881827 Chr.19: 17301596-17301596 on Build GRCh38
rs888456437 Chr.19: 17301676-17301676 on Build GRCh38
rs780297252 Chr.19: 17301567-17301567 on Build GRCh38
rs552233794 Chr.19: 17301595-17301595 on Build GRCh38
rs759068394 Chr.19: 17301598-17301598 on Build GRCh38
rs374747961 Chr.19: 17301644-17301644 on Build GRCh38
rs761619410 Chr.19: 17301562-17301563 on Build GRCh38
rs147808769 Chr.19: 17301556-17301556 on Build GRCh38
rs949602157 Chr.19: 17301594-17301594 on Build GRCh38
rs775329949 Chr.19: 17301608-17301608 on Build GRCh38
rs371431966 Chr.19: 17301642-17301642 on Build GRCh38
rs146901681 Chr.19: 17301568-17301568 on Build GRCh38
rs764220135 Chr.19: 17301680-17301680 on Build GRCh38
rs746966639 Chr.19: 17301585-17301585 on Build GRCh38
rs764619853 Chr.19: 17301640-17301640 on Build GRCh38
rs546146109 Chr.19: 17301560-17301560 on Build GRCh38
rs891822402 Chr.19: 17301686-17301686 on Build GRCh38
rs1010885468 Chr.19: 17301689-17301689 on Build GRCh38
rs142856821 Chr.19: 17301538-17301538 on Build GRCh38
rs765645691 Chr.19: 17301523-17301523 on Build GRCh38
rs756237719 Chr.19: 17301674-17301674 on Build GRCh38
rs11086067 Chr.19: 17301590-17301590 on Build GRCh38
rs145834574 Chr.19: 17301617-17301617 on Build GRCh38
rs760548258 Chr.19: 17301620-17301620 on Build GRCh38
rs11086066 Chr.19: 17301557-17301557 on Build GRCh38
rs968935995 Chr.19: 17301526-17301526 on Build GRCh38
rs919856528 Chr.19: 17301607-17301607 on Build GRCh38
rs766340046 Chr.19: 17301655-17301655 on Build GRCh38
rs764675043 Chr.19: 17301540-17301540 on Build GRCh38
rs767457257 Chr.19: 17301647-17301647 on Build GRCh38
rs777686823 Chr.19: 17301572-17301572 on Build GRCh38
rs772071597 Chr.19: 17301571-17301571 on Build GRCh38
rs753140950 Chr.19: 17301527-17301527 on Build GRCh38
rs750486923 Chr.19: 17301652-17301652 on Build GRCh38
rs367827324 Chr.19: 17301561-17301561 on Build GRCh38
rs773080943 Chr.19: 17301604-17301604 on Build GRCh38
rs151037617 Chr.19: 17301543-17301543 on Build GRCh38
rs912265508 Chr.19: 17301566-17301566 on Build GRCh38

View Full Product Details