Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 12081529-12081800 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHTKD1
Entrez Gene ID
55526
Gene Name
dehydrogenase E1 and transketolase domain containing 1
Gene Aliases
AMOXAD, CMT2Q
UniGene
Hs.104980
Gene Chromosome Location
Chr.10: 12068917-12123228 on Build GRCh38
Transcripts
NM_018706.6

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7080275 c.291delG p.G98fs*9 Chr.10: 12081608-12081608 on Build GRCh38
4677330 c.274C>A p.L92M Chr.10: 12081591-12081591 on Build GRCh38
427139 c.262G>A p.E88K Chr.10: 12081579-12081579 on Build GRCh38
1250048 c.249G>A p.L83L Chr.10: 12081566-12081566 on Build GRCh38
7428348 c.267C>T p.I89I Chr.10: 12081584-12081584 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs202067344 Chr.10: 12081658-12081658 on Build GRCh38
rs1722445 Chr.10: 12081668-12081668 on Build GRCh38
rs751295417 Chr.10: 12081649-12081649 on Build GRCh38
rs759279882 Chr.10: 12081621-12081621 on Build GRCh38
rs754265170 Chr.10: 12081657-12081657 on Build GRCh38
rs775431352 Chr.10: 12081623-12081623 on Build GRCh38
rs748501030 Chr.10: 12081748-12081748 on Build GRCh38
rs930925223 Chr.10: 12081569-12081569 on Build GRCh38
rs754213936 Chr.10: 12081566-12081566 on Build GRCh38
rs557500018 Chr.10: 12081554-12081554 on Build GRCh38
rs767227256 Chr.10: 12081651-12081651 on Build GRCh38
rs751243967 Chr.10: 12081559-12081559 on Build GRCh38
rs769632958 Chr.10: 12081622-12081622 on Build GRCh38
rs772184008 Chr.10: 12081595-12081595 on Build GRCh38
rs923349372 Chr.10: 12081736-12081736 on Build GRCh38
rs201207624 Chr.10: 12081576-12081576 on Build GRCh38
rs746398709 Chr.10: 12081665-12081666 on Build GRCh38
rs763659313 Chr.10: 12081558-12081558 on Build GRCh38
rs10795929 Chr.10: 12081772-12081772 on Build GRCh38
rs140768224 Chr.10: 12081604-12081604 on Build GRCh38
rs752172609 Chr.10: 12081552-12081552 on Build GRCh38
rs898202081 Chr.10: 12081564-12081564 on Build GRCh38
rs781305913 Chr.10: 12081608-12081608 on Build GRCh38
rs761587264 Chr.10: 12081650-12081650 on Build GRCh38
rs780534443 Chr.10: 12081561-12081561 on Build GRCh38
rs933488704 Chr.10: 12081744-12081744 on Build GRCh38
rs755459607 Chr.10: 12081572-12081572 on Build GRCh38
rs61847985 Chr.10: 12081749-12081749 on Build GRCh38
rs779415876 Chr.10: 12081575-12081575 on Build GRCh38
rs778225434 Chr.10: 12081767-12081767 on Build GRCh38
rs747213281 Chr.10: 12081607-12081607 on Build GRCh38
rs1021626029 Chr.10: 12081690-12081690 on Build GRCh38
rs879691087 Chr.10: 12081667-12081668 on Build GRCh38
rs1043449463 Chr.10: 12081768-12081768 on Build GRCh38
rs975226054 Chr.10: 12081684-12081684 on Build GRCh38
rs10906066 Chr.10: 12081667-12081667 on Build GRCh38
rs138526346 Chr.10: 12081551-12081551 on Build GRCh38
rs368269356 Chr.10: 12081620-12081620 on Build GRCh38
rs375541349 Chr.10: 12081560-12081560 on Build GRCh38
rs560311647 Chr.10: 12081704-12081704 on Build GRCh38
rs561180822 Chr.10: 12081720-12081720 on Build GRCh38
rs199959700 Chr.10: 12081611-12081611 on Build GRCh38
rs35442324 Chr.10: 12081624-12081624 on Build GRCh38

View Full Product Details