Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 24676192-24676421 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NPAP1
Entrez Gene ID
23742
Gene Name
nuclear pore associated protein 1
Gene Aliases
C15orf2
UniGene
Hs.649663
Gene Chromosome Location
Chr.15: 24675394-24683446 on Build GRCh38
Transcripts
NM_018958.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3500073 c.517G>A p.D173N Chr.15: 24676384-24676384 on Build GRCh38
6359576 c.528A>T p.K176N Chr.15: 24676395-24676395 on Build GRCh38
4053679 c.475A>G p.R159G Chr.15: 24676342-24676342 on Build GRCh38
138057 c.326G>A p.G109E Chr.15: 24676193-24676193 on Build GRCh38
294183 c.447C>T p.T149T Chr.15: 24676314-24676314 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755346108 Chr.15: 24676347-24676347 on Build GRCh38
rs781264951 Chr.15: 24676384-24676384 on Build GRCh38
rs139493881 Chr.15: 24676268-24676268 on Build GRCh38
rs776500183 Chr.15: 24676307-24676307 on Build GRCh38
rs768064836 Chr.15: 24676392-24676392 on Build GRCh38
rs762513843 Chr.15: 24676264-24676264 on Build GRCh38
rs764805101 Chr.15: 24676258-24676258 on Build GRCh38
rs35737140 Chr.15: 24676365-24676365 on Build GRCh38
rs762414613 Chr.15: 24676315-24676315 on Build GRCh38
rs775189386 Chr.15: 24676314-24676314 on Build GRCh38
rs746840625 Chr.15: 24676229-24676229 on Build GRCh38
rs776441508 Chr.15: 24676231-24676231 on Build GRCh38
rs757328409 Chr.15: 24676382-24676382 on Build GRCh38
rs746945090 Chr.15: 24676381-24676381 on Build GRCh38
rs200484163 Chr.15: 24676400-24676400 on Build GRCh38
rs753827965 Chr.15: 24676390-24676390 on Build GRCh38
rs759204891 Chr.15: 24676244-24676244 on Build GRCh38
rs750902724 Chr.15: 24676266-24676266 on Build GRCh38
rs773179649 Chr.15: 24676228-24676228 on Build GRCh38
rs773732497 Chr.15: 24676322-24676322 on Build GRCh38
rs947425966 Chr.15: 24676250-24676250 on Build GRCh38
rs766320087 Chr.15: 24676238-24676239 on Build GRCh38
rs368120585 Chr.15: 24676389-24676389 on Build GRCh38
rs761113460 Chr.15: 24676329-24676329 on Build GRCh38
rs748122844 Chr.15: 24676212-24676212 on Build GRCh38
rs34391654 Chr.15: 24676346-24676346 on Build GRCh38
rs770806597 Chr.15: 24676230-24676230 on Build GRCh38
rs371525434 Chr.15: 24676298-24676298 on Build GRCh38
rs745650476 Chr.15: 24676311-24676311 on Build GRCh38
rs78599594 Chr.15: 24676289-24676289 on Build GRCh38
rs755127854 Chr.15: 24676279-24676279 on Build GRCh38
rs902491766 Chr.15: 24676233-24676233 on Build GRCh38
rs981637768 Chr.15: 24676350-24676350 on Build GRCh38
rs771960423 Chr.15: 24676219-24676219 on Build GRCh38
rs878989315 Chr.15: 24676328-24676328 on Build GRCh38
rs144485323 Chr.15: 24676265-24676265 on Build GRCh38
rs569950473 Chr.15: 24676363-24676363 on Build GRCh38
rs143409729 Chr.15: 24676286-24676286 on Build GRCh38
rs748223943 Chr.15: 24676284-24676284 on Build GRCh38
rs779181944 Chr.15: 24676283-24676283 on Build GRCh38
rs754075519 Chr.15: 24676278-24676278 on Build GRCh38
rs752923324 Chr.15: 24676364-24676364 on Build GRCh38
rs769393442 Chr.15: 24676312-24676312 on Build GRCh38
rs764031857 Chr.15: 24676374-24676374 on Build GRCh38
rs774932721 Chr.15: 24676263-24676263 on Build GRCh38
rs952419790 Chr.15: 24676291-24676291 on Build GRCh38
rs35870568 Chr.15: 24676321-24676321 on Build GRCh38
rs1049852251 Chr.15: 24676259-24676259 on Build GRCh38
rs766736241 Chr.15: 24676334-24676334 on Build GRCh38

View Full Product Details