Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 113388195-113388468 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALAD
Entrez Gene ID
210
Gene Name
aminolevulinate dehydratase
Gene Aliases
ALADH, PBGS
UniGene
Hs.1227
Gene Chromosome Location
Chr.9: 113386312-113401338 on Build GRCh38
Transcripts
NM_000031.5 NM_001003945.2 NM_001317745.1 XM_011518364.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7281079 c.1019-6A>C p.? Chr.9: 113388367-113388367 on Build GRCh38
7281078 c.932-6A>C p.? Chr.9: 113388367-113388367 on Build GRCh38
1104224 c.1022C>A p.A341D Chr.9: 113388358-113388358 on Build GRCh38
1597922 c.935C>A p.A312D Chr.9: 113388358-113388358 on Build GRCh38
1179242 c.1080A>C p.*360C Chr.9: 113388300-113388300 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758531402 Chr.9: 113388261-113388261 on Build GRCh38
rs139825145 Chr.9: 113388331-113388331 on Build GRCh38
rs755903342 Chr.9: 113388398-113388398 on Build GRCh38
rs906141027 Chr.9: 113388419-113388419 on Build GRCh38
rs781273728 Chr.9: 113388296-113388296 on Build GRCh38
rs747969789 Chr.9: 113388298-113388298 on Build GRCh38
rs777081320 Chr.9: 113388265-113388269 on Build GRCh38
rs767016856 Chr.9: 113388369-113388369 on Build GRCh38
rs940850440 Chr.9: 113388422-113388422 on Build GRCh38
rs983914261 Chr.9: 113388404-113388404 on Build GRCh38
rs367719995 Chr.9: 113388241-113388241 on Build GRCh38
rs771411981 Chr.9: 113388250-113388251 on Build GRCh38
rs938894418 Chr.9: 113388426-113388426 on Build GRCh38
rs769534528 Chr.9: 113388299-113388299 on Build GRCh38
rs1020631364 Chr.9: 113388306-113388306 on Build GRCh38
rs780322260 Chr.9: 113388262-113388262 on Build GRCh38
rs1019830721 Chr.9: 113388219-113388219 on Build GRCh38
rs750930674 Chr.9: 113388348-113388348 on Build GRCh38
rs755168259 Chr.9: 113388291-113388291 on Build GRCh38
rs188845125 Chr.9: 113388303-113388303 on Build GRCh38
rs759952918 Chr.9: 113388390-113388391 on Build GRCh38
rs192226264 Chr.9: 113388443-113388443 on Build GRCh38
rs1044636201 Chr.9: 113388222-113388222 on Build GRCh38
rs555208841 Chr.9: 113388413-113388413 on Build GRCh38
rs762227805 Chr.9: 113388332-113388332 on Build GRCh38
rs899416104 Chr.9: 113388442-113388442 on Build GRCh38
rs377113920 Chr.9: 113388254-113388254 on Build GRCh38
rs749099723 Chr.9: 113388400-113388400 on Build GRCh38
rs749260844 Chr.9: 113388301-113388301 on Build GRCh38
rs758984721 Chr.9: 113388326-113388326 on Build GRCh38
rs1036516767 Chr.9: 113388434-113388434 on Build GRCh38
rs371063254 Chr.9: 113388412-113388412 on Build GRCh38
rs557510313 Chr.9: 113388353-113388353 on Build GRCh38
rs537911204 Chr.9: 113388439-113388439 on Build GRCh38
rs781461345 Chr.9: 113388388-113388388 on Build GRCh38
rs771691451 Chr.9: 113388330-113388330 on Build GRCh38
rs777500783 Chr.9: 113388399-113388399 on Build GRCh38
rs891929836 Chr.9: 113388440-113388440 on Build GRCh38
rs1044408379 Chr.9: 113388415-113388416 on Build GRCh38
rs1042375 Chr.9: 113388236-113388236 on Build GRCh38
rs143589885 Chr.9: 113388390-113388390 on Build GRCh38
rs372621957 Chr.9: 113388253-113388253 on Build GRCh38
rs112609758 Chr.9: 113388249-113388249 on Build GRCh38
rs965252079 Chr.9: 113388238-113388238 on Build GRCh38
rs35581515 Chr.9: 113388330-113388331 on Build GRCh38
rs763947225 Chr.9: 113388416-113388416 on Build GRCh38
rs1042374 Chr.9: 113388265-113388265 on Build GRCh38
rs373487651 Chr.9: 113388333-113388333 on Build GRCh38
rs751029544 Chr.9: 113388260-113388260 on Build GRCh38
rs1057318877 Chr.9: 113388427-113388427 on Build GRCh38
rs78691919 Chr.9: 113388305-113388305 on Build GRCh38
rs751722843 Chr.9: 113388279-113388279 on Build GRCh38
rs568594714 Chr.9: 113388444-113388444 on Build GRCh38
rs765774746 Chr.9: 113388255-113388255 on Build GRCh38
rs377627387 Chr.9: 113388371-113388371 on Build GRCh38
rs111390615 Chr.9: 113388370-113388370 on Build GRCh38

View Full Product Details