Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 34090250-34090521 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EMC7
Entrez Gene ID
56851
Gene Name
ER membrane protein complex subunit 7
Gene Aliases
C11orf3, C15orf24, HT022, ORF1-FL1
UniGene
Hs.160565
Gene Chromosome Location
Chr.15: 34084022-34101852 on Build GRCh38
Transcripts
NM_020154.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5817609 c.492A>G p.P164P Chr.15: 34090320-34090320 on Build GRCh38
4494479 c.433C>T p.P145S Chr.15: 34090379-34090379 on Build GRCh38
6871151 c.391G>A p.V131I Chr.15: 34090421-34090421 on Build GRCh38
2267835 c.462G>A p.S154S Chr.15: 34090350-34090350 on Build GRCh38
2267837 c.404C>T p.P135L Chr.15: 34090408-34090408 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs535294114 Chr.15: 34090414-34090414 on Build GRCh38
rs767261000 Chr.15: 34090290-34090290 on Build GRCh38
rs11557327 Chr.15: 34090378-34090378 on Build GRCh38
rs770602135 Chr.15: 34090380-34090380 on Build GRCh38
rs140273811 Chr.15: 34090375-34090375 on Build GRCh38
rs960108847 Chr.15: 34090388-34090388 on Build GRCh38
rs376147512 Chr.15: 34090383-34090383 on Build GRCh38
rs750692480 Chr.15: 34090346-34090346 on Build GRCh38
rs764094450 Chr.15: 34090319-34090319 on Build GRCh38
rs770555284 Chr.15: 34090272-34090272 on Build GRCh38
rs776617754 Chr.15: 34090447-34090447 on Build GRCh38
rs750533108 Chr.15: 34090466-34090466 on Build GRCh38
rs752555548 Chr.15: 34090294-34090294 on Build GRCh38
rs866005480 Chr.15: 34090429-34090429 on Build GRCh38
rs150305850 Chr.15: 34090341-34090341 on Build GRCh38
rs751716052 Chr.15: 34090482-34090482 on Build GRCh38
rs3751642 Chr.15: 34090276-34090276 on Build GRCh38
rs190604325 Chr.15: 34090351-34090351 on Build GRCh38
rs112390189 Chr.15: 34090314-34090314 on Build GRCh38
rs768843263 Chr.15: 34090356-34090356 on Build GRCh38
rs763784272 Chr.15: 34090437-34090437 on Build GRCh38
rs761708257 Chr.15: 34090449-34090449 on Build GRCh38
rs370272260 Chr.15: 34090415-34090415 on Build GRCh38
rs758341598 Chr.15: 34090470-34090470 on Build GRCh38
rs758668112 Chr.15: 34090350-34090350 on Build GRCh38
rs760344541 Chr.15: 34090304-34090304 on Build GRCh38
rs766635045 Chr.15: 34090471-34090471 on Build GRCh38
rs373701824 Chr.15: 34090420-34090420 on Build GRCh38
rs781778397 Chr.15: 34090368-34090368 on Build GRCh38
rs774940926 Chr.15: 34090419-34090419 on Build GRCh38
rs760533840 Chr.15: 34090428-34090428 on Build GRCh38
rs778959248 Chr.15: 34090345-34090345 on Build GRCh38
rs765110916 Chr.15: 34090452-34090452 on Build GRCh38
rs972824055 Chr.15: 34090438-34090438 on Build GRCh38
rs753715035 Chr.15: 34090335-34090335 on Build GRCh38
rs747402157 Chr.15: 34090353-34090353 on Build GRCh38
rs759237658 Chr.15: 34090277-34090277 on Build GRCh38

View Full Product Details