Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 53494127-53494376 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AKTIP
Entrez Gene ID
64400
Gene Name
AKT interacting protein
Gene Aliases
FT1, FTS
UniGene
Hs.380897
Gene Chromosome Location
Chr.16: 53491028-53504411 on Build GRCh38
Transcripts
NM_001012398.2 NM_001308325.1 NM_022476.3 XM_005256095.4 XM_005256096.4 XM_005256097.4 XM_005256098.4 XM_017023564.1 XM_017023565.1 XM_017023566.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3402363 c.619C>T p.Q207* Chr.16: 53494229-53494229 on Build GRCh38
4151352 c.653A>G p.K218R Chr.16: 53494195-53494195 on Build GRCh38
5754870 c.599T>C p.V200A Chr.16: 53494342-53494342 on Build GRCh38
6213373 c.594T>A p.A198A Chr.16: 53494347-53494347 on Build GRCh38
4660948 c.627_628insT p.K210fs*1 Chr.16: 53494220-53494221 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774284749 Chr.16: 53494181-53494181 on Build GRCh38
rs768429868 Chr.16: 53494330-53494330 on Build GRCh38
rs1012088845 Chr.16: 53494354-53494354 on Build GRCh38
rs749069548 Chr.16: 53494177-53494177 on Build GRCh38
rs749049970 Chr.16: 53494324-53494324 on Build GRCh38
rs761884339 Chr.16: 53494202-53494202 on Build GRCh38
rs772281688 Chr.16: 53494280-53494281 on Build GRCh38
rs746263084 Chr.16: 53494278-53494281 on Build GRCh38
rs779706980 Chr.16: 53494322-53494322 on Build GRCh38
rs753191952 Chr.16: 53494276-53494276 on Build GRCh38
rs766802657 Chr.16: 53494224-53494224 on Build GRCh38
rs911361305 Chr.16: 53494193-53494193 on Build GRCh38
rs151134332 Chr.16: 53494187-53494187 on Build GRCh38
rs769769588 Chr.16: 53494162-53494162 on Build GRCh38
rs756896209 Chr.16: 53494287-53494287 on Build GRCh38
rs748061956 Chr.16: 53494349-53494349 on Build GRCh38
rs780837326 Chr.16: 53494316-53494316 on Build GRCh38
rs767578006 Chr.16: 53494207-53494207 on Build GRCh38
rs773479084 Chr.16: 53494208-53494208 on Build GRCh38
rs528168629 Chr.16: 53494180-53494180 on Build GRCh38
rs755325401 Chr.16: 53494258-53494258 on Build GRCh38
rs1053737418 Chr.16: 53494265-53494265 on Build GRCh38
rs778887599 Chr.16: 53494342-53494342 on Build GRCh38
rs1008322968 Chr.16: 53494206-53494206 on Build GRCh38
rs1017995839 Chr.16: 53494310-53494310 on Build GRCh38
rs372351551 Chr.16: 53494325-53494325 on Build GRCh38
rs765679177 Chr.16: 53494262-53494262 on Build GRCh38
rs745513215 Chr.16: 53494152-53494152 on Build GRCh38
rs760860184 Chr.16: 53494219-53494219 on Build GRCh38
rs559351790 Chr.16: 53494169-53494169 on Build GRCh38
rs9927310 Chr.16: 53494167-53494167 on Build GRCh38
rs935313689 Chr.16: 53494247-53494248 on Build GRCh38
rs745564595 Chr.16: 53494317-53494317 on Build GRCh38
rs779853418 Chr.16: 53494306-53494306 on Build GRCh38
rs772184838 Chr.16: 53494355-53494355 on Build GRCh38
rs754189096 Chr.16: 53494242-53494242 on Build GRCh38
rs746599433 Chr.16: 53494324-53494324 on Build GRCh38
rs893823904 Chr.16: 53494280-53494280 on Build GRCh38

View Full Product Details