Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 53915284-53915555 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CACNG7
Entrez Gene ID
59284
Gene Name
calcium voltage-gated channel auxiliary subunit gamma 7
Gene Aliases
-
UniGene
Hs.631597
Gene Chromosome Location
Chr.19: 53909425-53943944 on Build GRCh38
Transcripts
NM_031896.4 XM_005259124.3 XM_017027093.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5528065 c.414C>T p.F138F Chr.19: 53915495-53915495 on Build GRCh38
359359 c.370C>G p.R124G Chr.19: 53915451-53915451 on Build GRCh38
259120 c.361G>T p.G121C Chr.19: 53915442-53915442 on Build GRCh38
6618736 c.360C>T p.I120I Chr.19: 53915441-53915441 on Build GRCh38
1712764 c.310C>T p.P104S Chr.19: 53915391-53915391 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769597460 Chr.19: 53915354-53915354 on Build GRCh38
rs748659461 Chr.19: 53915399-53915399 on Build GRCh38
rs779427253 Chr.19: 53915330-53915330 on Build GRCh38
rs780604612 Chr.19: 53915405-53915405 on Build GRCh38
rs756341554 Chr.19: 53915387-53915387 on Build GRCh38
rs754071753 Chr.19: 53915464-53915464 on Build GRCh38
rs759175000 Chr.19: 53915344-53915344 on Build GRCh38
rs147913695 Chr.19: 53915426-53915426 on Build GRCh38
rs764059997 Chr.19: 53915368-53915368 on Build GRCh38
rs761942078 Chr.19: 53915376-53915376 on Build GRCh38
rs267605662 Chr.19: 53915451-53915451 on Build GRCh38
rs762782484 Chr.19: 53915420-53915420 on Build GRCh38
rs201185188 Chr.19: 53915534-53915534 on Build GRCh38
rs373486264 Chr.19: 53915328-53915328 on Build GRCh38
rs376155498 Chr.19: 53915490-53915490 on Build GRCh38
rs758875080 Chr.19: 53915480-53915480 on Build GRCh38
rs768543537 Chr.19: 53915504-53915504 on Build GRCh38
rs752951314 Chr.19: 53915535-53915535 on Build GRCh38
rs899033509 Chr.19: 53915488-53915488 on Build GRCh38
rs369713864 Chr.19: 53915326-53915326 on Build GRCh38
rs755702985 Chr.19: 53915496-53915496 on Build GRCh38
rs267605661 Chr.19: 53915450-53915450 on Build GRCh38
rs750629734 Chr.19: 53915384-53915384 on Build GRCh38
rs755340207 Chr.19: 53915394-53915394 on Build GRCh38
rs771864611 Chr.19: 53915514-53915514 on Build GRCh38
rs371290517 Chr.19: 53915501-53915501 on Build GRCh38
rs774064816 Chr.19: 53915508-53915508 on Build GRCh38
rs746449508 Chr.19: 53915334-53915334 on Build GRCh38
rs769278610 Chr.19: 53915411-53915411 on Build GRCh38
rs933001525 Chr.19: 53915412-53915412 on Build GRCh38
rs776332659 Chr.19: 53915343-53915343 on Build GRCh38
rs572176547 Chr.19: 53915356-53915356 on Build GRCh38
rs751560841 Chr.19: 53915374-53915374 on Build GRCh38
rs748901775 Chr.19: 53915528-53915529 on Build GRCh38
rs543056281 Chr.19: 53915520-53915520 on Build GRCh38
rs745326651 Chr.19: 53915489-53915489 on Build GRCh38
rs779334025 Chr.19: 53915396-53915396 on Build GRCh38
rs866063392 Chr.19: 53915461-53915461 on Build GRCh38
rs1011186103 Chr.19: 53915351-53915351 on Build GRCh38
rs1026402611 Chr.19: 53915515-53915515 on Build GRCh38
rs967080427 Chr.19: 53915378-53915378 on Build GRCh38
rs1033004119 Chr.19: 53915311-53915311 on Build GRCh38
rs766519700 Chr.19: 53915456-53915456 on Build GRCh38
rs770360020 Chr.19: 53915340-53915340 on Build GRCh38
rs775471058 Chr.19: 53915355-53915355 on Build GRCh38
rs564240369 Chr.19: 53915409-53915409 on Build GRCh38
rs773206528 Chr.19: 53915517-53915517 on Build GRCh38
rs958774872 Chr.19: 53915445-53915445 on Build GRCh38
rs770469040 Chr.19: 53915401-53915401 on Build GRCh38
rs553878683 Chr.19: 53915427-53915427 on Build GRCh38
rs778361539 Chr.19: 53915483-53915483 on Build GRCh38
rs750421026 Chr.19: 53915439-53915439 on Build GRCh38
rs765655432 Chr.19: 53915472-53915472 on Build GRCh38
rs202017942 Chr.19: 53915530-53915530 on Build GRCh38
rs768292427 Chr.19: 53915529-53915529 on Build GRCh38
rs767563429 Chr.19: 53915377-53915377 on Build GRCh38
rs545594390 Chr.19: 53915390-53915390 on Build GRCh38
rs775309749 Chr.19: 53915414-53915414 on Build GRCh38
rs139985451 Chr.19: 53915465-53915465 on Build GRCh38
rs989719272 Chr.19: 53915338-53915338 on Build GRCh38
rs754163354 Chr.19: 53915393-53915393 on Build GRCh38
rs768633291 Chr.19: 53915422-53915422 on Build GRCh38
rs766503259 Chr.19: 53915529-53915529 on Build GRCh38
rs779563899 Chr.19: 53915340-53915341 on Build GRCh38
rs34480556 Chr.19: 53915441-53915442 on Build GRCh38
rs748000116 Chr.19: 53915510-53915510 on Build GRCh38
rs915437673 Chr.19: 53915350-53915350 on Build GRCh38

View Full Product Details