Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 116820961-116821208 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APOA4
Entrez Gene ID
337
Gene Name
apolipoprotein A4
Gene Aliases
-
UniGene
Hs.1247
Gene Chromosome Location
Chr.11: 116820702-116823295 on Build GRCh38
Transcripts
NM_000482.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3415556 c.1080G>T p.E360D Chr.11: 116820978-116820978 on Build GRCh38
405124 c.882delG p.H295fs*46 Chr.11: 116821176-116821176 on Build GRCh38
923369 c.858G>T p.Q286H Chr.11: 116821200-116821200 on Build GRCh38
4905447 c.910C>T p.R304* Chr.11: 116821148-116821148 on Build GRCh38
1240851 c.895C>A p.Q299K Chr.11: 116821163-116821163 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775248272 Chr.11: 116821110-116821110 on Build GRCh38
rs201032623 Chr.11: 116821041-116821041 on Build GRCh38
rs753125110 Chr.11: 116821149-116821149 on Build GRCh38
rs771207711 Chr.11: 116821109-116821109 on Build GRCh38
rs565211148 Chr.11: 116821066-116821066 on Build GRCh38
rs370490075 Chr.11: 116821186-116821186 on Build GRCh38
rs773341406 Chr.11: 116821028-116821028 on Build GRCh38
rs376352095 Chr.11: 116820987-116820987 on Build GRCh38
rs145761354 Chr.11: 116821052-116821052 on Build GRCh38
rs781747963 Chr.11: 116821148-116821148 on Build GRCh38
rs5108 Chr.11: 116821139-116821139 on Build GRCh38
rs1042833464 Chr.11: 116820992-116820992 on Build GRCh38
rs549843769 Chr.11: 116821141-116821141 on Build GRCh38
rs769788860 Chr.11: 116821089-116821089 on Build GRCh38
rs759328385 Chr.11: 116821031-116821031 on Build GRCh38
rs773393056 Chr.11: 116821102-116821102 on Build GRCh38
rs780657386 Chr.11: 116820991-116820991 on Build GRCh38
rs147577451 Chr.11: 116821004-116821004 on Build GRCh38
rs148203811 Chr.11: 116821055-116821055 on Build GRCh38
rs150264487 Chr.11: 116821145-116821145 on Build GRCh38
rs760472018 Chr.11: 116821120-116821120 on Build GRCh38
rs781766665 Chr.11: 116821079-116821079 on Build GRCh38
rs973577436 Chr.11: 116821166-116821166 on Build GRCh38
rs150624574 Chr.11: 116821147-116821147 on Build GRCh38
rs755486311 Chr.11: 116821073-116821073 on Build GRCh38
rs764903189 Chr.11: 116821059-116821059 on Build GRCh38
rs769994902 Chr.11: 116821013-116821013 on Build GRCh38
rs756688595 Chr.11: 116821163-116821163 on Build GRCh38
rs767280079 Chr.11: 116821036-116821036 on Build GRCh38
rs749437133 Chr.11: 116821106-116821106 on Build GRCh38
rs534105044 Chr.11: 116820984-116820984 on Build GRCh38
rs747488895 Chr.11: 116821000-116821000 on Build GRCh38
rs545034382 Chr.11: 116821053-116821053 on Build GRCh38
rs368128806 Chr.11: 116821038-116821038 on Build GRCh38
rs373920026 Chr.11: 116821128-116821128 on Build GRCh38
rs5107 Chr.11: 116821170-116821170 on Build GRCh38
rs371977184 Chr.11: 116821080-116821080 on Build GRCh38
rs977366818 Chr.11: 116821087-116821087 on Build GRCh38
rs377752186 Chr.11: 116821127-116821127 on Build GRCh38
rs146353487 Chr.11: 116821001-116821001 on Build GRCh38
rs748191355 Chr.11: 116821007-116821007 on Build GRCh38
rs201021435 Chr.11: 116821142-116821142 on Build GRCh38
rs762156180 Chr.11: 116821137-116821138 on Build GRCh38
rs5109 Chr.11: 116821050-116821050 on Build GRCh38
rs1038330230 Chr.11: 116820999-116820999 on Build GRCh38
rs113263292 Chr.11: 116821060-116821060 on Build GRCh38
rs955492329 Chr.11: 116821119-116821119 on Build GRCh38
rs771079346 Chr.11: 116821180-116821180 on Build GRCh38
rs760598613 Chr.11: 116821042-116821042 on Build GRCh38

View Full Product Details