Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 69153162-69153415 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCA10
Entrez Gene ID
10349
Gene Name
ATP binding cassette subfamily A member 10
Gene Aliases
EST698739
UniGene
Hs.25377
Gene Chromosome Location
Chr.17: 69148007-69244815 on Build GRCh38
Transcripts
NM_080282.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3521433 c.4117G>A p.E1373K Chr.17: 69153324-69153324 on Build GRCh38
318413 c.4106G>T p.G1369V Chr.17: 69153335-69153335 on Build GRCh38
3737471 c.4097C>G p.P1366R Chr.17: 69153344-69153344 on Build GRCh38
4439215 c.4076C>T p.S1359L Chr.17: 69153365-69153365 on Build GRCh38
5343007 c.4072C>T p.P1358S Chr.17: 69153369-69153369 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775612811 Chr.17: 69153254-69153254 on Build GRCh38
rs1026792991 Chr.17: 69153342-69153342 on Build GRCh38
rs904043184 Chr.17: 69153249-69153249 on Build GRCh38
rs370369427 Chr.17: 69153265-69153265 on Build GRCh38
rs773220832 Chr.17: 69153258-69153258 on Build GRCh38
rs774282619 Chr.17: 69153368-69153368 on Build GRCh38
rs530952082 Chr.17: 69153226-69153226 on Build GRCh38
rs201375015 Chr.17: 69153324-69153324 on Build GRCh38
rs751286341 Chr.17: 69153305-69153305 on Build GRCh38
rs201554524 Chr.17: 69153391-69153391 on Build GRCh38
rs201197046 Chr.17: 69153372-69153372 on Build GRCh38
rs1031674104 Chr.17: 69153273-69153273 on Build GRCh38
rs768808451 Chr.17: 69153350-69153350 on Build GRCh38
rs779392624 Chr.17: 69153336-69153336 on Build GRCh38
rs891411287 Chr.17: 69153289-69153289 on Build GRCh38
rs368430285 Chr.17: 69153358-69153358 on Build GRCh38
rs777109521 Chr.17: 69153325-69153325 on Build GRCh38
rs755799243 Chr.17: 69153387-69153387 on Build GRCh38
rs938654051 Chr.17: 69153385-69153385 on Build GRCh38
rs747184545 Chr.17: 69153344-69153344 on Build GRCh38
rs776538863 Chr.17: 69153352-69153352 on Build GRCh38
rs749842617 Chr.17: 69153328-69153328 on Build GRCh38
rs149740416 Chr.17: 69153240-69153240 on Build GRCh38
rs928536174 Chr.17: 69153361-69153361 on Build GRCh38
rs766209028 Chr.17: 69153363-69153363 on Build GRCh38
rs78875590 Chr.17: 69153343-69153343 on Build GRCh38
rs766232477 Chr.17: 69153296-69153296 on Build GRCh38
rs1043862814 Chr.17: 69153213-69153213 on Build GRCh38
rs752431471 Chr.17: 69153373-69153373 on Build GRCh38
rs764101694 Chr.17: 69153257-69153257 on Build GRCh38
rs200562513 Chr.17: 69153284-69153284 on Build GRCh38
rs906931014 Chr.17: 69153378-69153378 on Build GRCh38
rs754749374 Chr.17: 69153310-69153310 on Build GRCh38
rs767286512 Chr.17: 69153312-69153312 on Build GRCh38
rs199596306 Chr.17: 69153333-69153333 on Build GRCh38
rs999709609 Chr.17: 69153266-69153266 on Build GRCh38
rs374412556 Chr.17: 69153337-69153337 on Build GRCh38
rs752343610 Chr.17: 69153321-69153321 on Build GRCh38
rs763730655 Chr.17: 69153360-69153360 on Build GRCh38
rs759421451 Chr.17: 69153369-69153369 on Build GRCh38
rs760747573 Chr.17: 69153256-69153256 on Build GRCh38

View Full Product Details