Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 103575246-103575511 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMY2B
Entrez Gene ID
280
Gene Name
amylase, alpha 2B (pancreatic)
Gene Aliases
AMY2, AMY3, HXA
UniGene
Hs.484588
Gene Chromosome Location
Chr.1: 103554644-103579534 on Build GRCh38
Transcripts
NM_020978.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5039807 c.970G>A p.G324R Chr.1: 103575314-103575314 on Build GRCh38
6120225 c.994G>T p.D332Y Chr.1: 103575338-103575338 on Build GRCh38
1244913 c.1047T>A p.G349G Chr.1: 103575486-103575486 on Build GRCh38
414211 c.915C>A p.D305E Chr.1: 103575259-103575259 on Build GRCh38
6264015 c.1008T>C p.Y336Y Chr.1: 103575447-103575447 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs542691992 Chr.1: 103575366-103575367 on Build GRCh38
rs374177865 Chr.1: 103575363-103575363 on Build GRCh38
rs752701317 Chr.1: 103575272-103575272 on Build GRCh38
rs776177641 Chr.1: 103575337-103575337 on Build GRCh38
rs761519391 Chr.1: 103575381-103575381 on Build GRCh38
rs747507777 Chr.1: 103575437-103575437 on Build GRCh38
rs781653913 Chr.1: 103575458-103575458 on Build GRCh38
rs775272824 Chr.1: 103575472-103575472 on Build GRCh38
rs750640477 Chr.1: 103575350-103575350 on Build GRCh38
rs771258177 Chr.1: 103575335-103575335 on Build GRCh38
rs12075462 Chr.1: 103575345-103575345 on Build GRCh38
rs984694934 Chr.1: 103575286-103575286 on Build GRCh38
rs767334674 Chr.1: 103575483-103575483 on Build GRCh38
rs201867231 Chr.1: 103575349-103575349 on Build GRCh38
rs377079253 Chr.1: 103575300-103575300 on Build GRCh38
rs746200954 Chr.1: 103575460-103575460 on Build GRCh38
rs768148171 Chr.1: 103575475-103575475 on Build GRCh38
rs140978983 Chr.1: 103575299-103575299 on Build GRCh38
rs757065720 Chr.1: 103575296-103575296 on Build GRCh38
rs752831735 Chr.1: 103575410-103575410 on Build GRCh38
rs761762278 Chr.1: 103575481-103575481 on Build GRCh38
rs977482756 Chr.1: 103575358-103575358 on Build GRCh38
rs779102366 Chr.1: 103575401-103575401 on Build GRCh38
rs371219365 Chr.1: 103575269-103575269 on Build GRCh38
rs766832915 Chr.1: 103575365-103575366 on Build GRCh38
rs778916272 Chr.1: 103575302-103575302 on Build GRCh38
rs769545055 Chr.1: 103575342-103575342 on Build GRCh38
rs773031300 Chr.1: 103575485-103575485 on Build GRCh38
rs548551542 Chr.1: 103575285-103575285 on Build GRCh38
rs145913863 Chr.1: 103575455-103575455 on Build GRCh38
rs778146000 Chr.1: 103575434-103575434 on Build GRCh38
rs767305223 Chr.1: 103575382-103575382 on Build GRCh38
rs529999526 Chr.1: 103575275-103575275 on Build GRCh38
rs751457981 Chr.1: 103575379-103575379 on Build GRCh38
rs748205188 Chr.1: 103575304-103575304 on Build GRCh38
rs199878320 Chr.1: 103575389-103575389 on Build GRCh38
rs778328913 Chr.1: 103575318-103575318 on Build GRCh38
rs374864399 Chr.1: 103575297-103575297 on Build GRCh38
rs763999096 Chr.1: 103575367-103575367 on Build GRCh38
rs201657725 Chr.1: 103575366-103575366 on Build GRCh38
rs562580345 Chr.1: 103575390-103575390 on Build GRCh38
rs140209167 Chr.1: 103575270-103575270 on Build GRCh38
rs144622525 Chr.1: 103575471-103575471 on Build GRCh38
rs759212934 Chr.1: 103575338-103575338 on Build GRCh38
rs764797763 Chr.1: 103575271-103575271 on Build GRCh38
rs751380783 Chr.1: 103575291-103575291 on Build GRCh38
rs116293837 Chr.1: 103575474-103575474 on Build GRCh38
rs200884633 Chr.1: 103575315-103575315 on Build GRCh38
rs774187619 Chr.1: 103575480-103575480 on Build GRCh38
rs758444697 Chr.1: 103575421-103575421 on Build GRCh38
rs755242322 Chr.1: 103575394-103575394 on Build GRCh38
rs200799723 Chr.1: 103575317-103575317 on Build GRCh38

View Full Product Details