Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 28237608-28237861 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATPIF1
Entrez Gene ID
93974
Gene Name
ATPase inhibitory factor 1
Gene Aliases
ATPI, ATPIP, IP
UniGene
Hs.744914
Gene Chromosome Location
Chr.1: 28236091-28238105 on Build GRCh38
Transcripts
NM_016311.4 NM_178190.2 NM_178191.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5955052 c.193G>A p.E65K Chr.1: 28237850-28237850 on Build GRCh38
4832653 c.199G>C p.E67Q Chr.1: 28237780-28237780 on Build GRCh38
7231821 c.215G>A p.*72* Chr.1: 28237796-28237796 on Build GRCh38
7228235 c.187A>G p.R63G Chr.1: 28237768-28237768 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs904070877 Chr.1: 28237722-28237722 on Build GRCh38
rs778930273 Chr.1: 28237739-28237739 on Build GRCh38
rs112006347 Chr.1: 28237835-28237835 on Build GRCh38
rs766752378 Chr.1: 28237728-28237728 on Build GRCh38
rs758222441 Chr.1: 28237765-28237765 on Build GRCh38
rs757140062 Chr.1: 28237738-28237738 on Build GRCh38
rs752878076 Chr.1: 28237736-28237736 on Build GRCh38
rs948073072 Chr.1: 28237655-28237655 on Build GRCh38
rs113261974 Chr.1: 28237834-28237834 on Build GRCh38
rs369695947 Chr.1: 28237715-28237715 on Build GRCh38
rs376753470 Chr.1: 28237702-28237702 on Build GRCh38
rs900567898 Chr.1: 28237683-28237683 on Build GRCh38
rs777602793 Chr.1: 28237698-28237700 on Build GRCh38
rs141948364 Chr.1: 28237769-28237769 on Build GRCh38
rs997588141 Chr.1: 28237706-28237706 on Build GRCh38
rs774827581 Chr.1: 28237812-28237812 on Build GRCh38
rs751735945 Chr.1: 28237731-28237731 on Build GRCh38
rs763478808 Chr.1: 28237800-28237800 on Build GRCh38
rs935581566 Chr.1: 28237737-28237737 on Build GRCh38
rs8559 Chr.1: 28237741-28237741 on Build GRCh38
rs763390750 Chr.1: 28237833-28237844 on Build GRCh38
rs773657021 Chr.1: 28237795-28237795 on Build GRCh38
rs769304298 Chr.1: 28237792-28237792 on Build GRCh38
rs780027111 Chr.1: 28237767-28237767 on Build GRCh38
rs759662061 Chr.1: 28237824-28237824 on Build GRCh38
rs759838729 Chr.1: 28237732-28237732 on Build GRCh38
rs368919521 Chr.1: 28237825-28237825 on Build GRCh38
rs533689697 Chr.1: 28237801-28237801 on Build GRCh38
rs1034029245 Chr.1: 28237817-28237817 on Build GRCh38
rs9508 Chr.1: 28237768-28237768 on Build GRCh38
rs1008839767 Chr.1: 28237789-28237789 on Build GRCh38
rs1039011263 Chr.1: 28237680-28237680 on Build GRCh38
rs568776345 Chr.1: 28237648-28237648 on Build GRCh38
rs760640267 Chr.1: 28237720-28237720 on Build GRCh38
rs747624396 Chr.1: 28237774-28237774 on Build GRCh38
rs941990529 Chr.1: 28237641-28237641 on Build GRCh38
rs767469518 Chr.1: 28237733-28237733 on Build GRCh38

View Full Product Details