Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 58252684-58252911 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC113
Entrez Gene ID
29070
Gene Name
coiled-coil domain containing 113
Gene Aliases
HSPC065
UniGene
Hs.11614
Gene Chromosome Location
Chr.16: 58196863-58283836 on Build GRCh38
Transcripts
NM_001142302.1 NM_014157.3 XM_011523038.2 XM_011523039.2 XM_011523040.2 XM_011523041.2 XM_011523042.2 XM_017023176.1 XM_017023177.1 XM_017023178.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5851125 c.211G>A p.A71T Chr.16: 58252836-58252836 on Build GRCh38
5620424 c.206T>C p.I69T Chr.16: 58252831-58252831 on Build GRCh38
6195197 c.218A>G p.D73G Chr.16: 58252843-58252843 on Build GRCh38
5692578 c.135G>C p.E45D Chr.16: 58252760-58252760 on Build GRCh38
7179156 c.142G>C p.E48Q Chr.16: 58252767-58252767 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs185015668 Chr.16: 58252812-58252812 on Build GRCh38
rs750033417 Chr.16: 58252798-58252798 on Build GRCh38
rs765664558 Chr.16: 58252781-58252781 on Build GRCh38
rs747174505 Chr.16: 58252833-58252833 on Build GRCh38
rs550520231 Chr.16: 58252775-58252775 on Build GRCh38
rs773604432 Chr.16: 58252815-58252815 on Build GRCh38
rs111436398 Chr.16: 58252858-58252858 on Build GRCh38
rs748437962 Chr.16: 58252730-58252730 on Build GRCh38
rs568951247 Chr.16: 58252779-58252779 on Build GRCh38
rs747880097 Chr.16: 58252814-58252814 on Build GRCh38
rs551786429 Chr.16: 58252788-58252788 on Build GRCh38
rs948059786 Chr.16: 58252778-58252778 on Build GRCh38
rs375584362 Chr.16: 58252803-58252803 on Build GRCh38
rs753848299 Chr.16: 58252806-58252806 on Build GRCh38
rs772256237 Chr.16: 58252737-58252737 on Build GRCh38
rs200002781 Chr.16: 58252809-58252809 on Build GRCh38
rs529347659 Chr.16: 58252754-58252754 on Build GRCh38
rs755701219 Chr.16: 58252801-58252801 on Build GRCh38
rs752982246 Chr.16: 58252782-58252782 on Build GRCh38
rs760139041 Chr.16: 58252866-58252866 on Build GRCh38
rs767266077 Chr.16: 58252725-58252725 on Build GRCh38
rs866977136 Chr.16: 58252800-58252800 on Build GRCh38
rs771020066 Chr.16: 58252853-58252853 on Build GRCh38
rs761616001 Chr.16: 58252787-58252787 on Build GRCh38
rs777973771 Chr.16: 58252825-58252825 on Build GRCh38
rs773336052 Chr.16: 58252738-58252738 on Build GRCh38
rs760735689 Chr.16: 58252740-58252740 on Build GRCh38
rs368475564 Chr.16: 58252861-58252861 on Build GRCh38
rs113343928 Chr.16: 58252755-58252755 on Build GRCh38

View Full Product Details