Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 972031-972304 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP2A2
Entrez Gene ID
161
Gene Name
adaptor related protein complex 2 alpha 2 subunit
Gene Aliases
ADTAB, CLAPA2, HIP-9, HIP9, HYPJ
UniGene
Hs.19121
Gene Chromosome Location
Chr.11: 925809-1012245 on Build GRCh38
Transcripts
NM_001242837.1 NM_012305.3 XM_011519929.1 XM_011519930.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2062059 c.370A>G p.N124D Chr.11: 972152-972152 on Build GRCh38
4037807 c.330C>T p.R110R Chr.11: 972112-972112 on Build GRCh38
2062061 c.406G>T p.A136S Chr.11: 972188-972188 on Build GRCh38
1358043 c.343G>A p.A115T Chr.11: 972125-972125 on Build GRCh38
7292936 c.365G>A p.S122N Chr.11: 972147-972147 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs7933889 Chr.11: 972270-972270 on Build GRCh38
rs765372174 Chr.11: 972247-972247 on Build GRCh38
rs763177214 Chr.11: 972250-972250 on Build GRCh38
rs763389818 Chr.11: 972188-972188 on Build GRCh38
rs757060254 Chr.11: 972193-972193 on Build GRCh38
rs768724916 Chr.11: 972057-972057 on Build GRCh38
rs778559415 Chr.11: 972278-972278 on Build GRCh38
rs759978873 Chr.11: 972239-972239 on Build GRCh38
rs762607300 Chr.11: 972106-972106 on Build GRCh38
rs752922457 Chr.11: 972150-972150 on Build GRCh38
rs745798520 Chr.11: 972203-972203 on Build GRCh38
rs528656716 Chr.11: 972273-972273 on Build GRCh38
rs757486268 Chr.11: 972190-972198 on Build GRCh38
rs556658081 Chr.11: 972059-972059 on Build GRCh38
rs375422112 Chr.11: 972253-972253 on Build GRCh38
rs773931850 Chr.11: 972219-972219 on Build GRCh38
rs766748347 Chr.11: 972248-972248 on Build GRCh38
rs755014932 Chr.11: 972143-972143 on Build GRCh38
rs769361795 Chr.11: 972058-972058 on Build GRCh38
rs998528469 Chr.11: 972259-972260 on Build GRCh38
rs200256869 Chr.11: 972190-972190 on Build GRCh38
rs776886746 Chr.11: 972282-972282 on Build GRCh38
rs545499861 Chr.11: 972275-972275 on Build GRCh38
rs780014620 Chr.11: 972271-972271 on Build GRCh38
rs571569960 Chr.11: 972109-972109 on Build GRCh38
rs765834089 Chr.11: 972129-972129 on Build GRCh38
rs766894467 Chr.11: 972135-972135 on Build GRCh38
rs1046856409 Chr.11: 972112-972112 on Build GRCh38
rs771202088 Chr.11: 972224-972224 on Build GRCh38
rs771274317 Chr.11: 972280-972280 on Build GRCh38
rs780864083 Chr.11: 972268-972268 on Build GRCh38
rs117612040 Chr.11: 972118-972118 on Build GRCh38
rs919983704 Chr.11: 972227-972227 on Build GRCh38
rs368541164 Chr.11: 972149-972149 on Build GRCh38
rs751877003 Chr.11: 972262-972262 on Build GRCh38
rs775011494 Chr.11: 972214-972214 on Build GRCh38
rs111597325 Chr.11: 972257-972257 on Build GRCh38
rs769236848 Chr.11: 972204-972204 on Build GRCh38
rs747765935 Chr.11: 972279-972279 on Build GRCh38
rs1016694991 Chr.11: 972132-972132 on Build GRCh38
rs371262427 Chr.11: 972124-972124 on Build GRCh38
rs775058159 Chr.11: 972103-972103 on Build GRCh38
rs375531485 Chr.11: 972140-972140 on Build GRCh38
rs942404129 Chr.11: 972258-972258 on Build GRCh38
rs777134957 Chr.11: 972226-972226 on Build GRCh38
rs1040686640 Chr.11: 972260-972260 on Build GRCh38
rs781186303 Chr.11: 972194-972194 on Build GRCh38
rs372522058 Chr.11: 972187-972187 on Build GRCh38
rs905541763 Chr.11: 972276-972276 on Build GRCh38
rs539232320 Chr.11: 972264-972264 on Build GRCh38
rs761093838 Chr.11: 972223-972223 on Build GRCh38
rs758742768 Chr.11: 972254-972254 on Build GRCh38
rs748836668 Chr.11: 972215-972215 on Build GRCh38
rs768273850 Chr.11: 972217-972217 on Build GRCh38

View Full Product Details