Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 17507552-17507821 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAP2
Entrez Gene ID
10486
Gene Name
CAP, adenylate cyclase-associated protein, 2 (yeast)
Gene Aliases
-
UniGene
Hs.132902
Gene Chromosome Location
Chr.6: 17393216-17557792 on Build GRCh38
Transcripts
NM_006366.2 XM_011514233.2 NR_110855.1

Gene Symbol
LOC101928491
Entrez Gene ID
101928491
Gene Name
uncharacterized LOC101928491
Gene Aliases
-
UniGene
Hs.622876
Gene Chromosome Location
Chr.6: 17501360-17511265 on Build GRCh38
Transcripts
NM_006366.2 XM_011514233.2 NR_110855.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5869806 c.454C>T p.P152S Chr.6: 17507650-17507650 on Build GRCh38
5967937 c.506G>T p.R169M Chr.6: 17507702-17507702 on Build GRCh38
7359406 c.474G>C p.E158D Chr.6: 17507670-17507670 on Build GRCh38
3623044 c.503A>G p.N168S Chr.6: 17507699-17507699 on Build GRCh38
5869808 c.477G>A p.M159I Chr.6: 17507673-17507673 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757590490 Chr.6: 17507739-17507739 on Build GRCh38
rs767425282 Chr.6: 17507629-17507629 on Build GRCh38
rs778806530 Chr.6: 17507680-17507680 on Build GRCh38
rs372057498 Chr.6: 17507624-17507624 on Build GRCh38
rs202134882 Chr.6: 17507670-17507670 on Build GRCh38
rs774665618 Chr.6: 17507728-17507729 on Build GRCh38
rs760532114 Chr.6: 17507652-17507652 on Build GRCh38
rs771387880 Chr.6: 17507765-17507765 on Build GRCh38
rs772446004 Chr.6: 17507592-17507592 on Build GRCh38
rs778067921 Chr.6: 17507597-17507597 on Build GRCh38
rs373415673 Chr.6: 17507759-17507759 on Build GRCh38
rs758545277 Chr.6: 17507729-17507729 on Build GRCh38
rs146122387 Chr.6: 17507758-17507758 on Build GRCh38
rs900404840 Chr.6: 17507763-17507763 on Build GRCh38
rs199864799 Chr.6: 17507776-17507776 on Build GRCh38
rs777003309 Chr.6: 17507607-17507607 on Build GRCh38
rs762566898 Chr.6: 17507612-17507612 on Build GRCh38
rs763107046 Chr.6: 17507597-17507597 on Build GRCh38
rs1051146814 Chr.6: 17507662-17507662 on Build GRCh38
rs778055994 Chr.6: 17507653-17507653 on Build GRCh38
rs1011275312 Chr.6: 17507764-17507764 on Build GRCh38
rs768032048 Chr.6: 17507620-17507620 on Build GRCh38
rs369973316 Chr.6: 17507748-17507748 on Build GRCh38
rs762352770 Chr.6: 17507734-17507737 on Build GRCh38
rs144444597 Chr.6: 17507731-17507731 on Build GRCh38
rs201738681 Chr.6: 17507698-17507698 on Build GRCh38
rs930128979 Chr.6: 17507585-17507585 on Build GRCh38
rs766490475 Chr.6: 17507666-17507666 on Build GRCh38
rs997697040 Chr.6: 17507736-17507736 on Build GRCh38
rs139737796 Chr.6: 17507730-17507730 on Build GRCh38
rs530636738 Chr.6: 17507786-17507786 on Build GRCh38
rs201244167 Chr.6: 17507770-17507770 on Build GRCh38
rs747516198 Chr.6: 17507603-17507603 on Build GRCh38
rs771489826 Chr.6: 17507606-17507606 on Build GRCh38
rs766721451 Chr.6: 17507589-17507589 on Build GRCh38
rs750186666 Chr.6: 17507638-17507638 on Build GRCh38
rs769073379 Chr.6: 17507596-17507597 on Build GRCh38
rs142527859 Chr.6: 17507704-17507704 on Build GRCh38
rs755133331 Chr.6: 17507679-17507679 on Build GRCh38
rs761519773 Chr.6: 17507628-17507628 on Build GRCh38

View Full Product Details