Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 43184837-43185106 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CUL9
Entrez Gene ID
23113
Gene Name
cullin 9
Gene Aliases
H7AP1, PARC
UniGene
Hs.485434
Gene Chromosome Location
Chr.6: 43182170-43224587 on Build GRCh38
Transcripts
NM_015089.3 XM_011514422.1 XM_011514423.2 XM_011514424.1 XM_011514425.1 XM_011514426.1 XM_011514428.1 XM_011514430.1 XM_017010589.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5626302 c.565A>G p.M189V Chr.6: 43184875-43184875 on Build GRCh38
6638239 c.569T>C p.L190P Chr.6: 43184879-43184879 on Build GRCh38
7239687 c.592G>T p.A198S Chr.6: 43184902-43184902 on Build GRCh38
1729478 c.536A>T p.E179V Chr.6: 43184846-43184846 on Build GRCh38
40441 c.548G>A p.R183H Chr.6: 43184858-43184858 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs558820727 Chr.6: 43184970-43184970 on Build GRCh38
rs895022243 Chr.6: 43185037-43185037 on Build GRCh38
rs35604391 Chr.6: 43185055-43185056 on Build GRCh38
rs1049579691 Chr.6: 43185073-43185073 on Build GRCh38
rs764962364 Chr.6: 43184904-43184904 on Build GRCh38
rs780432976 Chr.6: 43184968-43184968 on Build GRCh38
rs773291608 Chr.6: 43184861-43184861 on Build GRCh38
rs1042676623 Chr.6: 43185032-43185033 on Build GRCh38
rs759068521 Chr.6: 43184898-43184898 on Build GRCh38
rs72867558 Chr.6: 43185055-43185055 on Build GRCh38
rs1054876232 Chr.6: 43185015-43185015 on Build GRCh38
rs557353517 Chr.6: 43184971-43184971 on Build GRCh38
rs768006179 Chr.6: 43184922-43184922 on Build GRCh38
rs947075755 Chr.6: 43185019-43185019 on Build GRCh38
rs868130797 Chr.6: 43184917-43184917 on Build GRCh38
rs868094459 Chr.6: 43184956-43184956 on Build GRCh38
rs748263558 Chr.6: 43184858-43184858 on Build GRCh38
rs768458784 Chr.6: 43184911-43184912 on Build GRCh38
rs41274930 Chr.6: 43184985-43184985 on Build GRCh38
rs750987964 Chr.6: 43184941-43184941 on Build GRCh38
rs543843460 Chr.6: 43184964-43184964 on Build GRCh38
rs770676984 Chr.6: 43184889-43184889 on Build GRCh38
rs762454887 Chr.6: 43184913-43184913 on Build GRCh38
rs772080051 Chr.6: 43184860-43184860 on Build GRCh38
rs945001591 Chr.6: 43184899-43184899 on Build GRCh38
rs905810105 Chr.6: 43185049-43185049 on Build GRCh38
rs760546219 Chr.6: 43184867-43184867 on Build GRCh38
rs774921382 Chr.6: 43184906-43184906 on Build GRCh38
rs539556444 Chr.6: 43184953-43184953 on Build GRCh38
rs538972823 Chr.6: 43184957-43184957 on Build GRCh38
rs776312174 Chr.6: 43184895-43184895 on Build GRCh38
rs975340330 Chr.6: 43184857-43184857 on Build GRCh38
rs755419257 Chr.6: 43185054-43185054 on Build GRCh38

View Full Product Details