Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 92355019-92355246 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAT3
Entrez Gene ID
120114
Gene Name
FAT atypical cadherin 3
Gene Aliases
CDHF15, CDHR10, hFat3
UniGene
Hs.98523
Gene Chromosome Location
Chr.11: 92224636-92896473 on Build GRCh38
Transcripts
NM_001008781.2 XM_017017178.1 XM_017017179.1 XM_017017180.1 XM_017017181.1 XM_017017182.1 XM_017017183.1 XM_017017184.1 XM_017017186.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6070896 c.2928C>G p.V976V Chr.11: 92355040-92355040 on Build GRCh38
4037493 c.3112C>T p.L1038F Chr.11: 92355224-92355224 on Build GRCh38
5790519 c.3029T>C p.V1010A Chr.11: 92355141-92355141 on Build GRCh38
7184974 c.2931T>C p.N977N Chr.11: 92355043-92355043 on Build GRCh38
6070891 c.3096G>T p.V1032V Chr.11: 92355208-92355208 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs187314509 Chr.11: 92355199-92355199 on Build GRCh38
rs369427789 Chr.11: 92355147-92355147 on Build GRCh38
rs766450180 Chr.11: 92355110-92355110 on Build GRCh38
rs779693833 Chr.11: 92355173-92355173 on Build GRCh38
rs777521459 Chr.11: 92355042-92355042 on Build GRCh38
rs375939567 Chr.11: 92355085-92355085 on Build GRCh38
rs539020470 Chr.11: 92355090-92355090 on Build GRCh38
rs557128937 Chr.11: 92355216-92355216 on Build GRCh38
rs765323524 Chr.11: 92355217-92355217 on Build GRCh38
rs746696427 Chr.11: 92355049-92355049 on Build GRCh38
rs181670458 Chr.11: 92355144-92355144 on Build GRCh38
rs751247669 Chr.11: 92355160-92355160 on Build GRCh38
rs780758673 Chr.11: 92355081-92355081 on Build GRCh38
rs759676852 Chr.11: 92355139-92355139 on Build GRCh38
rs775508425 Chr.11: 92355222-92355222 on Build GRCh38
rs866833607 Chr.11: 92355146-92355146 on Build GRCh38
rs745533159 Chr.11: 92355163-92355163 on Build GRCh38
rs772618201 Chr.11: 92355100-92355100 on Build GRCh38
rs376758242 Chr.11: 92355162-92355162 on Build GRCh38
rs747388173 Chr.11: 92355203-92355203 on Build GRCh38
rs373884887 Chr.11: 92355159-92355159 on Build GRCh38
rs762382379 Chr.11: 92355097-92355097 on Build GRCh38
rs771319509 Chr.11: 92355207-92355207 on Build GRCh38
rs961960612 Chr.11: 92355217-92355217 on Build GRCh38
rs776757183 Chr.11: 92355215-92355215 on Build GRCh38
rs535283582 Chr.11: 92355125-92355125 on Build GRCh38
rs748692596 Chr.11: 92355187-92355187 on Build GRCh38
rs1027630627 Chr.11: 92355107-92355107 on Build GRCh38
rs199580154 Chr.11: 92355143-92355143 on Build GRCh38
rs201896979 Chr.11: 92355089-92355089 on Build GRCh38
rs755729530 Chr.11: 92355169-92355169 on Build GRCh38
rs772636695 Chr.11: 92355191-92355191 on Build GRCh38
rs566236434 Chr.11: 92355104-92355104 on Build GRCh38
rs756949096 Chr.11: 92355161-92355161 on Build GRCh38
rs776896055 Chr.11: 92355129-92355129 on Build GRCh38
rs140704029 Chr.11: 92355103-92355103 on Build GRCh38
rs770450284 Chr.11: 92355074-92355074 on Build GRCh38

View Full Product Details