Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 44335637-44335894 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C21orf2
Entrez Gene ID
755
Gene Name
chromosome 21 open reading frame 2
Gene Aliases
LRRC76, YF5/A2
UniGene
Hs.517331
Gene Chromosome Location
Chr.21: 44328944-44339417 on Build GRCh38
Transcripts
NM_001271440.1 NM_001271441.1 NM_001271442.1 NM_004928.2 XM_006724051.3 XM_006724053.3 XM_017028470.1 XM_017028471.1 XM_017028472.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6196013 c.137C>T p.T46M Chr.21: 44335764-44335764 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs750472057 Chr.21: 44335714-44335714 on Build GRCh38
rs889274820 Chr.21: 44335698-44335698 on Build GRCh38
rs982267515 Chr.21: 44335756-44335756 on Build GRCh38
rs897021988 Chr.21: 44335787-44335787 on Build GRCh38
rs750561784 Chr.21: 44335782-44335782 on Build GRCh38
rs767420984 Chr.21: 44335778-44335778 on Build GRCh38
rs376087157 Chr.21: 44335875-44335893 on Build GRCh38
rs200038076 Chr.21: 44335741-44335741 on Build GRCh38
rs748929175 Chr.21: 44335856-44335856 on Build GRCh38
rs766422381 Chr.21: 44335825-44335825 on Build GRCh38
rs749459116 Chr.21: 44335725-44335725 on Build GRCh38
rs751412265 Chr.21: 44335850-44335851 on Build GRCh38
rs755186566 Chr.21: 44335834-44335834 on Build GRCh38
rs779288110 Chr.21: 44335749-44335749 on Build GRCh38
rs761779834 Chr.21: 44335775-44335775 on Build GRCh38
rs939925757 Chr.21: 44335808-44335808 on Build GRCh38
rs146104157 Chr.21: 44335763-44335763 on Build GRCh38
rs189695721 Chr.21: 44335852-44335852 on Build GRCh38
rs775012349 Chr.21: 44335762-44335762 on Build GRCh38
rs202196237 Chr.21: 44335724-44335724 on Build GRCh38
rs186382635 Chr.21: 44335672-44335672 on Build GRCh38
rs752657657 Chr.21: 44335840-44335842 on Build GRCh38
rs370014560 Chr.21: 44335821-44335821 on Build GRCh38
rs774096260 Chr.21: 44335772-44335772 on Build GRCh38
rs762837824 Chr.21: 44335844-44335845 on Build GRCh38
rs763871034 Chr.21: 44335848-44335849 on Build GRCh38
rs889292055 Chr.21: 44335858-44335858 on Build GRCh38
rs754027698 Chr.21: 44335832-44335832 on Build GRCh38
rs112926736 Chr.21: 44335870-44335870 on Build GRCh38
rs200830387 Chr.21: 44335726-44335726 on Build GRCh38
rs779571401 Chr.21: 44335854-44335854 on Build GRCh38
rs970527998 Chr.21: 44335755-44335755 on Build GRCh38
rs763821454 Chr.21: 44335764-44335764 on Build GRCh38
rs201329990 Chr.21: 44335748-44335748 on Build GRCh38
rs751468650 Chr.21: 44335710-44335710 on Build GRCh38
rs924132138 Chr.21: 44335699-44335699 on Build GRCh38
rs997989180 Chr.21: 44335687-44335687 on Build GRCh38
rs1027355126 Chr.21: 44335836-44335836 on Build GRCh38
rs779185167 Chr.21: 44335845-44335845 on Build GRCh38
rs757142399 Chr.21: 44335711-44335711 on Build GRCh38
rs909781389 Chr.21: 44335794-44335794 on Build GRCh38
rs372854948 Chr.21: 44335750-44335750 on Build GRCh38
rs138100132 Chr.21: 44335760-44335760 on Build GRCh38
rs769425801 Chr.21: 44335828-44335828 on Build GRCh38
rs749414890 Chr.21: 44335668-44335668 on Build GRCh38
rs562586179 Chr.21: 44335860-44335860 on Build GRCh38
rs147855042 Chr.21: 44335853-44335853 on Build GRCh38
rs201127621 Chr.21: 44335851-44335851 on Build GRCh38
rs375698855 Chr.21: 44335751-44335751 on Build GRCh38
rs765036174 Chr.21: 44335825-44335828 on Build GRCh38
rs1007821232 Chr.21: 44335701-44335701 on Build GRCh38
rs113602050 Chr.21: 44335742-44335742 on Build GRCh38
rs1030536546 Chr.21: 44335688-44335688 on Build GRCh38
rs142252957 Chr.21: 44335713-44335713 on Build GRCh38
rs139328963 Chr.21: 44335869-44335887 on Build GRCh38
rs868517153 Chr.21: 44335783-44335783 on Build GRCh38
rs756184388 Chr.21: 44335789-44335789 on Build GRCh38
rs912172384 Chr.21: 44335681-44335681 on Build GRCh38
rs182333890 Chr.21: 44335857-44335857 on Build GRCh38

View Full Product Details