Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 16159371-16159642 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCC6
Entrez Gene ID
368
Gene Name
ATP binding cassette subfamily C member 6
Gene Aliases
ABC34, ARA, EST349056, GACI2, MLP1, MOAT-E, MOATE, MRP6, PXE, PXE1, URG7
UniGene
Hs.442182
Gene Chromosome Location
Chr.16: 16149565-16223616 on Build GRCh38
Transcripts
NM_001171.5 XM_011522479.2 XM_011522480.1 XM_011522481.2 XM_017023212.1 XM_017023213.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1376141 c.3726G>A p.T1242T Chr.16: 16159491-16159491 on Build GRCh38
967549 c.3704G>A p.R1235Q Chr.16: 16159513-16159513 on Build GRCh38
1283181 c.3729C>G p.P1243P Chr.16: 16159488-16159488 on Build GRCh38
6042947 c.3703C>T p.R1235W Chr.16: 16159514-16159514 on Build GRCh38
4545220 c.3733G>A p.E1245K Chr.16: 16159484-16159484 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs59049137 Chr.16: 16159435-16159435 on Build GRCh38
rs906374625 Chr.16: 16159618-16159618 on Build GRCh38
rs773961518 Chr.16: 16159485-16159485 on Build GRCh38
rs368931767 Chr.16: 16159527-16159527 on Build GRCh38
rs972225942 Chr.16: 16159546-16159546 on Build GRCh38
rs945926144 Chr.16: 16159400-16159400 on Build GRCh38
rs371191765 Chr.16: 16159557-16159557 on Build GRCh38
rs770483331 Chr.16: 16159540-16159540 on Build GRCh38
rs281865557 Chr.16: 16159482-16159482 on Build GRCh38
rs768576172 Chr.16: 16159437-16159437 on Build GRCh38
rs63750402 Chr.16: 16159514-16159514 on Build GRCh38
rs371493792 Chr.16: 16159568-16159568 on Build GRCh38
rs755947755 Chr.16: 16159598-16159598 on Build GRCh38
rs745938225 Chr.16: 16159509-16159509 on Build GRCh38
rs141449320 Chr.16: 16159492-16159492 on Build GRCh38
rs114017587 Chr.16: 16159565-16159565 on Build GRCh38
rs749436704 Chr.16: 16159433-16159433 on Build GRCh38
rs940876455 Chr.16: 16159531-16159531 on Build GRCh38
rs752499858 Chr.16: 16159455-16159456 on Build GRCh38
rs376412306 Chr.16: 16159427-16159427 on Build GRCh38
rs1041519747 Chr.16: 16159404-16159404 on Build GRCh38
rs762552663 Chr.16: 16159458-16159458 on Build GRCh38
rs63751215 Chr.16: 16159556-16159556 on Build GRCh38
rs141728905 Chr.16: 16159526-16159526 on Build GRCh38
rs914524231 Chr.16: 16159621-16159621 on Build GRCh38
rs150145577 Chr.16: 16159499-16159499 on Build GRCh38
rs72657701 Chr.16: 16159494-16159494 on Build GRCh38
rs747993729 Chr.16: 16159518-16159518 on Build GRCh38
rs1024789925 Chr.16: 16159406-16159406 on Build GRCh38
rs757431953 Chr.16: 16159456-16159456 on Build GRCh38
rs72653747 Chr.16: 16159500-16159500 on Build GRCh38
rs775947674 Chr.16: 16159563-16159563 on Build GRCh38
rs777360866 Chr.16: 16159606-16159606 on Build GRCh38
rs1046269881 Chr.16: 16159615-16159615 on Build GRCh38
rs920811008 Chr.16: 16159547-16159547 on Build GRCh38
rs769432314 Chr.16: 16159519-16159519 on Build GRCh38
rs138700741 Chr.16: 16159513-16159513 on Build GRCh38
rs386789396 Chr.16: 16159434-16159435 on Build GRCh38
rs764318423 Chr.16: 16159573-16159573 on Build GRCh38
rs749050638 Chr.16: 16159426-16159427 on Build GRCh38
rs772707425 Chr.16: 16159453-16159453 on Build GRCh38
rs63750125 Chr.16: 16159541-16159541 on Build GRCh38
rs776633873 Chr.16: 16159440-16159440 on Build GRCh38
rs63749796 Chr.16: 16159505-16159505 on Build GRCh38
rs769533776 Chr.16: 16159449-16159449 on Build GRCh38
rs398078529 Chr.16: 16159434-16159434 on Build GRCh38
rs72664212 Chr.16: 16159586-16159586 on Build GRCh38
rs754104618 Chr.16: 16159578-16159578 on Build GRCh38
rs372710375 Chr.16: 16159472-16159472 on Build GRCh38
rs914408711 Chr.16: 16159396-16159396 on Build GRCh38
rs777846973 Chr.16: 16159431-16159431 on Build GRCh38
rs762007802 Chr.16: 16159581-16159581 on Build GRCh38
rs562368920 Chr.16: 16159464-16159464 on Build GRCh38
rs199786977 Chr.16: 16159473-16159473 on Build GRCh38
rs778069919 Chr.16: 16159426-16159426 on Build GRCh38
rs765483994 Chr.16: 16159583-16159583 on Build GRCh38
rs563265838 Chr.16: 16159474-16159474 on Build GRCh38
rs749109161 Chr.16: 16159620-16159620 on Build GRCh38
rs374927491 Chr.16: 16159593-16159593 on Build GRCh38
rs72653745 Chr.16: 16159549-16159549 on Build GRCh38
rs398028845 Chr.16: 16159435-16159435 on Build GRCh38
rs944125276 Chr.16: 16159428-16159428 on Build GRCh38
rs56822940 Chr.16: 16159427-16159427 on Build GRCh38
rs63751001 Chr.16: 16159555-16159555 on Build GRCh38
rs72653746 Chr.16: 16159508-16159508 on Build GRCh38
rs747942423 Chr.16: 16159446-16159446 on Build GRCh38
rs72653748 Chr.16: 16159495-16159495 on Build GRCh38
rs63749992 Chr.16: 16159502-16159502 on Build GRCh38
rs779018991 Chr.16: 16159524-16159525 on Build GRCh38
rs778791988 Chr.16: 16159506-16159506 on Build GRCh38
rs546587473 Chr.16: 16159587-16159587 on Build GRCh38
rs201806988 Chr.16: 16159479-16159479 on Build GRCh38
rs58538400 Chr.16: 16159481-16159481 on Build GRCh38

View Full Product Details