Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 180911436-180911673 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTNL8
Entrez Gene ID
79908
Gene Name
butyrophilin like 8
Gene Aliases
BTN9.2
UniGene
Hs.189109
Gene Chromosome Location
Chr.5: 180899077-180952166 on Build GRCh38
Transcripts
NM_001040462.2 NM_001159707.1 NM_001159708.1 NM_001159709.1 NM_001159710.1 NM_024850.2 XM_011534649.1 XM_011534650.2 XM_011534651.2 XM_011534652.2 XM_011534653.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6103616 c.588C>A p.T196T Chr.5: 180911529-180911529 on Build GRCh38
1642711 c.556C>T p.H186Y Chr.5: 180911497-180911497 on Build GRCh38
3614991 c.670G>A p.G224R Chr.5: 180911611-180911611 on Build GRCh38
1642712 c.556C>T p.H186Y Chr.5: 180911497-180911497 on Build GRCh38
6103617 c.588C>A p.T196T Chr.5: 180911529-180911529 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749175716 Chr.5: 180911564-180911564 on Build GRCh38
rs759058379 Chr.5: 180911511-180911511 on Build GRCh38
rs878937428 Chr.5: 180911651-180911651 on Build GRCh38
rs201314326 Chr.5: 180911477-180911477 on Build GRCh38
rs148293230 Chr.5: 180911649-180911649 on Build GRCh38
rs928392180 Chr.5: 180911603-180911603 on Build GRCh38
rs762340755 Chr.5: 180911596-180911596 on Build GRCh38
rs377631162 Chr.5: 180911595-180911595 on Build GRCh38
rs201891387 Chr.5: 180911602-180911602 on Build GRCh38
rs754119428 Chr.5: 180911530-180911530 on Build GRCh38
rs776245269 Chr.5: 180911629-180911629 on Build GRCh38
rs762431858 Chr.5: 180911533-180911533 on Build GRCh38
rs958670515 Chr.5: 180911619-180911619 on Build GRCh38
rs374252379 Chr.5: 180911585-180911585 on Build GRCh38
rs766478838 Chr.5: 180911626-180911626 on Build GRCh38
rs574786683 Chr.5: 180911535-180911535 on Build GRCh38
rs369025007 Chr.5: 180911496-180911496 on Build GRCh38
rs540479681 Chr.5: 180911545-180911545 on Build GRCh38
rs763403199 Chr.5: 180911617-180911617 on Build GRCh38
rs141844193 Chr.5: 180911506-180911506 on Build GRCh38
rs751803412 Chr.5: 180911542-180911542 on Build GRCh38
rs745572100 Chr.5: 180911568-180911568 on Build GRCh38
rs377605273 Chr.5: 180911541-180911541 on Build GRCh38
rs762079358 Chr.5: 180911587-180911587 on Build GRCh38
rs746524916 Chr.5: 180911567-180911567 on Build GRCh38
rs376649884 Chr.5: 180911495-180911495 on Build GRCh38
rs372363727 Chr.5: 180911461-180911461 on Build GRCh38
rs143894862 Chr.5: 180911566-180911566 on Build GRCh38
rs762544045 Chr.5: 180911500-180911500 on Build GRCh38
rs374590204 Chr.5: 180911628-180911628 on Build GRCh38
rs749267396 Chr.5: 180911645-180911645 on Build GRCh38
rs750568279 Chr.5: 180911536-180911536 on Build GRCh38
rs753576169 Chr.5: 180911628-180911630 on Build GRCh38
rs375445903 Chr.5: 180911632-180911634 on Build GRCh38
rs200912428 Chr.5: 180911494-180911494 on Build GRCh38
rs138408550 Chr.5: 180911468-180911468 on Build GRCh38
rs751610961 Chr.5: 180911627-180911627 on Build GRCh38
rs755872032 Chr.5: 180911637-180911637 on Build GRCh38
rs369175545 Chr.5: 180911547-180911547 on Build GRCh38
rs769831495 Chr.5: 180911551-180911551 on Build GRCh38
rs757228791 Chr.5: 180911647-180911647 on Build GRCh38
rs373005299 Chr.5: 180911499-180911499 on Build GRCh38
rs146434143 Chr.5: 180911584-180911584 on Build GRCh38
rs554947046 Chr.5: 180911513-180911513 on Build GRCh38
rs755118033 Chr.5: 180911544-180911544 on Build GRCh38
rs980662952 Chr.5: 180911572-180911572 on Build GRCh38
rs374616316 Chr.5: 180911648-180911648 on Build GRCh38
rs770494151 Chr.5: 180911503-180911503 on Build GRCh38
rs560299365 Chr.5: 180911561-180911561 on Build GRCh38

View Full Product Details