Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 38054652-38054877 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DSCR4
Entrez Gene ID
10281
Gene Name
Down syndrome critical region 4
Gene Aliases
DCRB, DSCRB
UniGene
Hs.23251
Gene Chromosome Location
Chr.21: 38054011-38121360 on Build GRCh38
Transcripts
NM_005867.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
395898 c.263G>A p.C88Y Chr.21: 38054741-38054741 on Build GRCh38
3912089 c.300C>T p.L100L Chr.21: 38054704-38054704 on Build GRCh38
2845054 c.340G>A p.A114T Chr.21: 38054664-38054664 on Build GRCh38
74441 c.322A>G p.R108G Chr.21: 38054682-38054682 on Build GRCh38
4134927 c.285T>C p.L95L Chr.21: 38054719-38054719 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773450833 Chr.21: 38054816-38054816 on Build GRCh38
rs916680116 Chr.21: 38054819-38054819 on Build GRCh38
rs748238072 Chr.21: 38054740-38054741 on Build GRCh38
rs377625455 Chr.21: 38054818-38054818 on Build GRCh38
rs370355127 Chr.21: 38054787-38054787 on Build GRCh38
rs754097235 Chr.21: 38054686-38054686 on Build GRCh38
rs916154461 Chr.21: 38054852-38054853 on Build GRCh38
rs770598123 Chr.21: 38054760-38054760 on Build GRCh38
rs769105103 Chr.21: 38054795-38054795 on Build GRCh38
rs751772004 Chr.21: 38054745-38054745 on Build GRCh38
rs577166865 Chr.21: 38054700-38054700 on Build GRCh38
rs772381845 Chr.21: 38054742-38054743 on Build GRCh38
rs776544423 Chr.21: 38054788-38054792 on Build GRCh38
rs748022221 Chr.21: 38054726-38054726 on Build GRCh38
rs781175876 Chr.21: 38054722-38054722 on Build GRCh38
rs748903050 Chr.21: 38054753-38054753 on Build GRCh38
rs769682330 Chr.21: 38054740-38054740 on Build GRCh38
rs762209853 Chr.21: 38054806-38054806 on Build GRCh38
rs991761975 Chr.21: 38054758-38054758 on Build GRCh38
rs773670558 Chr.21: 38054764-38054764 on Build GRCh38
rs192313727 Chr.21: 38054691-38054691 on Build GRCh38
rs761263119 Chr.21: 38054774-38054774 on Build GRCh38
rs537333690 Chr.21: 38054677-38054677 on Build GRCh38
rs878978507 Chr.21: 38054715-38054715 on Build GRCh38
rs775222121 Chr.21: 38054817-38054817 on Build GRCh38
rs963386981 Chr.21: 38054701-38054701 on Build GRCh38
rs765479707 Chr.21: 38054690-38054690 on Build GRCh38
rs777617061 Chr.21: 38054749-38054749 on Build GRCh38
rs779954662 Chr.21: 38054699-38054699 on Build GRCh38
rs754812344 Chr.21: 38054703-38054703 on Build GRCh38

View Full Product Details