Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 196910190-196910417 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CFHR4
Entrez Gene ID
10877
Gene Name
complement factor H related 4
Gene Aliases
CFHL4, FHR-4, FHR4
UniGene
Hs.710100
Gene Chromosome Location
Chr.1: 196888014-196918911 on Build GRCh38
Transcripts
NM_001201550.2 NM_001201551.1 NM_006684.4 XM_006711129.3 XM_017000110.1 XM_017000111.1 XM_017000112.1 XM_017000113.1 XM_017000114.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3480882 c.932G>A p.G311E Chr.1: 196910416-196910416 on Build GRCh38
3976771 c.867C>A p.Y289* Chr.1: 196910351-196910351 on Build GRCh38
4903450 c.817C>T p.P273S Chr.1: 196910301-196910301 on Build GRCh38
3687836 c.857G>A p.R286H Chr.1: 196910341-196910341 on Build GRCh38
5467964 c.880A>G p.T294A Chr.1: 196910364-196910364 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs867140793 Chr.1: 196910302-196910302 on Build GRCh38
rs905279177 Chr.1: 196910250-196910250 on Build GRCh38
rs756020612 Chr.1: 196910319-196910319 on Build GRCh38
rs770991524 Chr.1: 196910337-196910344 on Build GRCh38
rs188298928 Chr.1: 196910341-196910341 on Build GRCh38
rs753643477 Chr.1: 196910333-196910333 on Build GRCh38
rs569248104 Chr.1: 196910338-196910338 on Build GRCh38
rs777174191 Chr.1: 196910293-196910293 on Build GRCh38
rs199583121 Chr.1: 196910307-196910307 on Build GRCh38
rs764444563 Chr.1: 196910316-196910316 on Build GRCh38
rs773940527 Chr.1: 196910266-196910266 on Build GRCh38
rs910856197 Chr.1: 196910324-196910324 on Build GRCh38
rs1003594535 Chr.1: 196910258-196910258 on Build GRCh38
rs1022530373 Chr.1: 196910297-196910297 on Build GRCh38
rs777012805 Chr.1: 196910385-196910385 on Build GRCh38
rs777524466 Chr.1: 196910371-196910371 on Build GRCh38
rs540816872 Chr.1: 196910263-196910263 on Build GRCh38
rs772730258 Chr.1: 196910277-196910277 on Build GRCh38
rs534869827 Chr.1: 196910267-196910267 on Build GRCh38
rs770523337 Chr.1: 196910383-196910383 on Build GRCh38
rs780179186 Chr.1: 196910247-196910247 on Build GRCh38
rs780015139 Chr.1: 196910331-196910331 on Build GRCh38
rs921604716 Chr.1: 196910368-196910368 on Build GRCh38
rs771482209 Chr.1: 196910350-196910350 on Build GRCh38
rs184131375 Chr.1: 196910223-196910223 on Build GRCh38
rs752827321 Chr.1: 196910308-196910308 on Build GRCh38
rs747541418 Chr.1: 196910268-196910268 on Build GRCh38
rs762884559 Chr.1: 196910314-196910314 on Build GRCh38
rs778653315 Chr.1: 196910339-196910339 on Build GRCh38
rs959411831 Chr.1: 196910238-196910238 on Build GRCh38
rs956889654 Chr.1: 196910276-196910276 on Build GRCh38
rs528613959 Chr.1: 196910261-196910261 on Build GRCh38
rs552795605 Chr.1: 196910354-196910354 on Build GRCh38
rs766701460 Chr.1: 196910287-196910287 on Build GRCh38
rs751699894 Chr.1: 196910318-196910318 on Build GRCh38
rs202044205 Chr.1: 196910279-196910279 on Build GRCh38
rs929814541 Chr.1: 196910340-196910340 on Build GRCh38
rs562079661 Chr.1: 196910272-196910272 on Build GRCh38
rs41265219 Chr.1: 196910303-196910303 on Build GRCh38
rs749214375 Chr.1: 196910255-196910255 on Build GRCh38

View Full Product Details