Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 241633742-241634015 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHML
Entrez Gene ID
1122
Gene Name
CHM like, Rab escort protein 2
Gene Aliases
REP2
UniGene
Hs.654545
Gene Chromosome Location
Chr.1: 241628864-241635930 on Build GRCh38
Transcripts
NM_001821.3 NM_014322.2

Gene Symbol
OPN3
Entrez Gene ID
23596
Gene Name
opsin 3
Gene Aliases
ECPN, PPP1R116
UniGene
Hs.409081
Gene Chromosome Location
Chr.1: 241593150-241640399 on Build GRCh38
Transcripts
NM_001821.3 NM_014322.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5031633 c.1818C>A p.F606L Chr.1: 241633949-241633949 on Build GRCh38
6628457 c.1914A>G p.L638L Chr.1: 241633853-241633853 on Build GRCh38
2050005 c.1824T>A p.P608P Chr.1: 241633943-241633943 on Build GRCh38
1185802 c.1793A>C p.Q598P Chr.1: 241633974-241633974 on Build GRCh38
77466 c.1882C>T p.P628S Chr.1: 241633885-241633885 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772627535 Chr.1: 241633782-241633782 on Build GRCh38
rs750692689 Chr.1: 241633780-241633789 on Build GRCh38
rs747095226 Chr.1: 241633804-241633804 on Build GRCh38
rs201624623 Chr.1: 241633856-241633856 on Build GRCh38
rs745548979 Chr.1: 241633854-241633854 on Build GRCh38
rs1054959882 Chr.1: 241633989-241633989 on Build GRCh38
rs751665271 Chr.1: 241633928-241633928 on Build GRCh38
rs767353388 Chr.1: 241633951-241633952 on Build GRCh38
rs776566714 Chr.1: 241633867-241633867 on Build GRCh38
rs768091835 Chr.1: 241633863-241633863 on Build GRCh38
rs752604774 Chr.1: 241633977-241633978 on Build GRCh38
rs144022344 Chr.1: 241633873-241633873 on Build GRCh38
rs765054000 Chr.1: 241633881-241633881 on Build GRCh38
rs374188147 Chr.1: 241633859-241633859 on Build GRCh38
rs750635085 Chr.1: 241633784-241633784 on Build GRCh38
rs368558008 Chr.1: 241633766-241633766 on Build GRCh38
rs763138761 Chr.1: 241633888-241633888 on Build GRCh38
rs748606837 Chr.1: 241633843-241633843 on Build GRCh38
rs538044891 Chr.1: 241633853-241633853 on Build GRCh38
rs755122437 Chr.1: 241633933-241633933 on Build GRCh38
rs778050094 Chr.1: 241633968-241633968 on Build GRCh38
rs749652365 Chr.1: 241633982-241633982 on Build GRCh38
rs76973247 Chr.1: 241633781-241633781 on Build GRCh38
rs760232961 Chr.1: 241633860-241633860 on Build GRCh38
rs754721864 Chr.1: 241633872-241633872 on Build GRCh38
rs912333621 Chr.1: 241633953-241633953 on Build GRCh38
rs201439073 Chr.1: 241633789-241633789 on Build GRCh38
rs753704460 Chr.1: 241633770-241633770 on Build GRCh38
rs770303697 Chr.1: 241633850-241633850 on Build GRCh38
rs534065519 Chr.1: 241633954-241633954 on Build GRCh38
rs766614640 Chr.1: 241633920-241633920 on Build GRCh38
rs771688883 Chr.1: 241633992-241633992 on Build GRCh38
rs781370771 Chr.1: 241633940-241633940 on Build GRCh38
rs780044404 Chr.1: 241633790-241633790 on Build GRCh38
rs781765555 Chr.1: 241633830-241633830 on Build GRCh38
rs372520346 Chr.1: 241633776-241633776 on Build GRCh38
rs774960334 Chr.1: 241633923-241633923 on Build GRCh38
rs750207699 Chr.1: 241633885-241633885 on Build GRCh38
rs755604697 Chr.1: 241633807-241633807 on Build GRCh38
rs553907341 Chr.1: 241633994-241633994 on Build GRCh38
rs932351146 Chr.1: 241633913-241633913 on Build GRCh38
rs920004999 Chr.1: 241633941-241633941 on Build GRCh38
rs753286099 Chr.1: 241633947-241633947 on Build GRCh38

View Full Product Details