Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 33577070-33577297 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAMTS12
Entrez Gene ID
81792
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 12
Gene Aliases
PRO4389
UniGene
Hs.12680
Gene Chromosome Location
Chr.5: 33523535-33892180 on Build GRCh38
Transcripts
NM_001324512.1 NM_030955.3 XM_017009905.1 XM_017009906.1 XM_017009907.1 XM_017009908.1 XM_017009909.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5908466 c.2954C>T p.P985L Chr.5: 33577072-33577072 on Build GRCh38
5630787 c.2895G>C p.R965R Chr.5: 33577131-33577131 on Build GRCh38
5952562 c.2938G>A p.D980N Chr.5: 33577088-33577088 on Build GRCh38
256246 c.2899C>T p.R967C Chr.5: 33577127-33577127 on Build GRCh38
273882 c.2894G>A p.R965Q Chr.5: 33577132-33577132 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745531836 Chr.5: 33577175-33577175 on Build GRCh38
rs767143473 Chr.5: 33577100-33577100 on Build GRCh38
rs201935843 Chr.5: 33577189-33577189 on Build GRCh38
rs750865031 Chr.5: 33577116-33577116 on Build GRCh38
rs367882059 Chr.5: 33577133-33577133 on Build GRCh38
rs899912281 Chr.5: 33577139-33577139 on Build GRCh38
rs752828513 Chr.5: 33577206-33577206 on Build GRCh38
rs756263283 Chr.5: 33577207-33577207 on Build GRCh38
rs749212498 Chr.5: 33577152-33577152 on Build GRCh38
rs762253270 Chr.5: 33577103-33577103 on Build GRCh38
rs758813862 Chr.5: 33577123-33577123 on Build GRCh38
rs1022702885 Chr.5: 33577226-33577226 on Build GRCh38
rs776343027 Chr.5: 33577198-33577198 on Build GRCh38
rs544451042 Chr.5: 33577255-33577255 on Build GRCh38
rs748210600 Chr.5: 33577138-33577138 on Build GRCh38
rs372910215 Chr.5: 33577191-33577191 on Build GRCh38
rs760273828 Chr.5: 33577184-33577184 on Build GRCh38
rs774224405 Chr.5: 33577171-33577171 on Build GRCh38
rs377084884 Chr.5: 33577094-33577094 on Build GRCh38
rs183388124 Chr.5: 33577222-33577222 on Build GRCh38
rs553050048 Chr.5: 33577163-33577163 on Build GRCh38
rs765736039 Chr.5: 33577109-33577109 on Build GRCh38
rs766719558 Chr.5: 33577126-33577126 on Build GRCh38
rs140294636 Chr.5: 33577093-33577093 on Build GRCh38
rs971117167 Chr.5: 33577242-33577242 on Build GRCh38
rs541891837 Chr.5: 33577219-33577219 on Build GRCh38
rs192176463 Chr.5: 33577190-33577190 on Build GRCh38
rs777802460 Chr.5: 33577150-33577150 on Build GRCh38
rs563669246 Chr.5: 33577132-33577132 on Build GRCh38
rs530992437 Chr.5: 33577248-33577248 on Build GRCh38
rs1017195989 Chr.5: 33577213-33577213 on Build GRCh38
rs375366015 Chr.5: 33577127-33577127 on Build GRCh38
rs370210174 Chr.5: 33577098-33577098 on Build GRCh38
rs774893888 Chr.5: 33577176-33577176 on Build GRCh38
rs756032392 Chr.5: 33577147-33577147 on Build GRCh38

View Full Product Details