Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 292573-292838 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PGGHG
Entrez Gene ID
80162
Gene Name
protein-glucosylgalactosylhydroxylysine glucosidase
Gene Aliases
ATHL1
UniGene
Hs.353181
Gene Chromosome Location
Chr.11: 287760-295688 on Build GRCh38
Transcripts
NM_025092.4 XM_011520383.2 XM_011520384.2 XM_011520385.2 XM_011520386.2 XM_011520387.2 XM_017018355.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2113647 c.523G>A p.D175N Chr.11: 292573-292573 on Build GRCh38
6207614 c.1073G>T p.C358F Chr.11: 292592-292592 on Build GRCh38
5522163 c.1119C>T p.A373A Chr.11: 292638-292638 on Build GRCh38
2113649 c.535G>C p.E179Q Chr.11: 292585-292585 on Build GRCh38
1585680 c.1126T>G p.L376V Chr.11: 292645-292645 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs149924011 Chr.11: 292603-292603 on Build GRCh38
rs774815107 Chr.11: 292633-292633 on Build GRCh38
rs532427019 Chr.11: 292702-292702 on Build GRCh38
rs755784553 Chr.11: 292604-292604 on Build GRCh38
rs1008106150 Chr.11: 292635-292635 on Build GRCh38
rs145002734 Chr.11: 292639-292639 on Build GRCh38
rs751539733 Chr.11: 292726-292726 on Build GRCh38
rs148680130 Chr.11: 292671-292671 on Build GRCh38
rs753557852 Chr.11: 292648-292648 on Build GRCh38
rs752094043 Chr.11: 292593-292593 on Build GRCh38
rs193253835 Chr.11: 292638-292638 on Build GRCh38
rs201699493 Chr.11: 292605-292605 on Build GRCh38
rs759331565 Chr.11: 292711-292711 on Build GRCh38
rs777778425 Chr.11: 292621-292621 on Build GRCh38
rs140683809 Chr.11: 292744-292744 on Build GRCh38
rs767915781 Chr.11: 292601-292601 on Build GRCh38
rs978701733 Chr.11: 292697-292697 on Build GRCh38
rs374726787 Chr.11: 292627-292627 on Build GRCh38
rs370134855 Chr.11: 292689-292689 on Build GRCh38
rs776118411 Chr.11: 292710-292710 on Build GRCh38
rs764086715 Chr.11: 292721-292721 on Build GRCh38
rs754426046 Chr.11: 292649-292649 on Build GRCh38
rs11246056 Chr.11: 292805-292805 on Build GRCh38
rs371810242 Chr.11: 292644-292644 on Build GRCh38
rs749571408 Chr.11: 292683-292683 on Build GRCh38
rs746935151 Chr.11: 292609-292609 on Build GRCh38
rs989113188 Chr.11: 292778-292778 on Build GRCh38
rs1043428459 Chr.11: 292802-292802 on Build GRCh38
rs568096396 Chr.11: 292809-292809 on Build GRCh38
rs758468353 Chr.11: 292654-292654 on Build GRCh38
rs764826682 Chr.11: 292713-292713 on Build GRCh38
rs774477538 Chr.11: 292688-292688 on Build GRCh38
rs1026815173 Chr.11: 292637-292637 on Build GRCh38
rs560201194 Chr.11: 292690-292690 on Build GRCh38
rs775069650 Chr.11: 292668-292668 on Build GRCh38
rs183604938 Chr.11: 292756-292756 on Build GRCh38
rs543435538 Chr.11: 292789-292789 on Build GRCh38
rs1006777736 Chr.11: 292645-292645 on Build GRCh38
rs371739590 Chr.11: 292608-292608 on Build GRCh38
rs771963570 Chr.11: 292616-292616 on Build GRCh38
rs759761373 Chr.11: 292626-292626 on Build GRCh38
rs765879602 Chr.11: 292647-292647 on Build GRCh38
rs369079985 Chr.11: 292632-292632 on Build GRCh38
rs373066201 Chr.11: 292706-292706 on Build GRCh38
rs913463388 Chr.11: 292779-292779 on Build GRCh38
rs757989538 Chr.11: 292720-292720 on Build GRCh38
rs139998628 Chr.11: 292595-292595 on Build GRCh38
rs375741439 Chr.11: 292653-292653 on Build GRCh38
rs776080639 Chr.11: 292738-292738 on Build GRCh38
rs780092877 Chr.11: 292681-292681 on Build GRCh38
rs762905133 Chr.11: 292631-292631 on Build GRCh38
rs11602423 Chr.11: 292798-292798 on Build GRCh38
rs760355642 Chr.11: 292673-292673 on Build GRCh38
rs574718267 Chr.11: 292658-292658 on Build GRCh38
rs768689080 Chr.11: 292685-292685 on Build GRCh38
rs752196874 Chr.11: 292716-292716 on Build GRCh38
rs777870311 Chr.11: 292664-292664 on Build GRCh38
rs115225588 Chr.11: 292667-292667 on Build GRCh38
rs762113759 Chr.11: 292598-292598 on Build GRCh38
rs754519348 Chr.11: 292606-292606 on Build GRCh38

View Full Product Details