Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 96883735-96883994 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ERAP1
Entrez Gene ID
51752
Gene Name
endoplasmic reticulum aminopeptidase 1
Gene Aliases
A-LAP, ALAP, APPILS, ARTS-1, ARTS1, ERAAP, ERAAP1, PILS-AP, PILSAP
UniGene
Hs.666524
Gene Chromosome Location
Chr.5: 96759245-96935983 on Build GRCh38
Transcripts
XM_011543480.2 XM_011543481.2 XM_011543484.2 XM_011543485.2 XM_011543486.2 XM_017009580.1 XM_017009581.1 XM_011543544.1

Gene Symbol
ERAP2
Entrez Gene ID
64167
Gene Name
endoplasmic reticulum aminopeptidase 2
Gene Aliases
L-RAP, LRAP
UniGene
Hs.482910
Gene Chromosome Location
Chr.5: 96875940-96919703 on Build GRCh38
Transcripts
XM_011543480.2 XM_011543481.2 XM_011543484.2 XM_011543485.2 XM_011543486.2 XM_017009580.1 XM_017009581.1 XM_011543544.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4424795 c.685A>T p.R229W Chr.5: 96883901-96883901 on Build GRCh38
4915086 c.623T>C p.F208S Chr.5: 96883839-96883839 on Build GRCh38
7216831 c.641C>T p.P214L Chr.5: 96883857-96883857 on Build GRCh38
3776939 c.598G>C p.E200Q Chr.5: 96883814-96883814 on Build GRCh38
6214234 c.605C>A p.T202N Chr.5: 96883821-96883821 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs79422968 Chr.5: 96883853-96883853 on Build GRCh38
rs777647974 Chr.5: 96883891-96883892 on Build GRCh38
rs915945750 Chr.5: 96883780-96883780 on Build GRCh38
rs188408214 Chr.5: 96883961-96883961 on Build GRCh38
rs3733905 Chr.5: 96883857-96883857 on Build GRCh38
rs919861748 Chr.5: 96883799-96883799 on Build GRCh38
rs73150323 Chr.5: 96883830-96883830 on Build GRCh38
rs148145360 Chr.5: 96883829-96883829 on Build GRCh38
rs761999795 Chr.5: 96883794-96883794 on Build GRCh38
rs748231518 Chr.5: 96883888-96883888 on Build GRCh38
rs749134867 Chr.5: 96883951-96883951 on Build GRCh38
rs769707979 Chr.5: 96883885-96883885 on Build GRCh38
rs199510383 Chr.5: 96883890-96883890 on Build GRCh38
rs781024551 Chr.5: 96883939-96883939 on Build GRCh38
rs777181457 Chr.5: 96883968-96883968 on Build GRCh38
rs138972366 Chr.5: 96883889-96883889 on Build GRCh38
rs372040296 Chr.5: 96883905-96883905 on Build GRCh38
rs773509300 Chr.5: 96883893-96883893 on Build GRCh38
rs773494377 Chr.5: 96883801-96883801 on Build GRCh38
rs776579246 Chr.5: 96883769-96883769 on Build GRCh38
rs201654009 Chr.5: 96883781-96883781 on Build GRCh38
rs376759795 Chr.5: 96883931-96883931 on Build GRCh38
rs183470843 Chr.5: 96883940-96883940 on Build GRCh38
rs1008012664 Chr.5: 96883886-96883886 on Build GRCh38
rs758234017 Chr.5: 96883877-96883880 on Build GRCh38
rs368480890 Chr.5: 96883895-96883895 on Build GRCh38
rs780076212 Chr.5: 96883945-96883945 on Build GRCh38
rs760174507 Chr.5: 96883911-96883911 on Build GRCh38
rs200920921 Chr.5: 96883936-96883936 on Build GRCh38
rs908009571 Chr.5: 96883868-96883868 on Build GRCh38
rs771424061 Chr.5: 96883897-96883897 on Build GRCh38
rs745873777 Chr.5: 96883864-96883864 on Build GRCh38
rs777650524 Chr.5: 96883954-96883954 on Build GRCh38
rs757064973 Chr.5: 96883861-96883861 on Build GRCh38
rs764904138 Chr.5: 96883834-96883834 on Build GRCh38
rs768729926 Chr.5: 96883776-96883776 on Build GRCh38
rs373773533 Chr.5: 96883856-96883856 on Build GRCh38
rs778910462 Chr.5: 96883887-96883887 on Build GRCh38
rs377453003 Chr.5: 96883858-96883858 on Build GRCh38
rs765813173 Chr.5: 96883917-96883917 on Build GRCh38
rs145672042 Chr.5: 96883800-96883800 on Build GRCh38
rs747920343 Chr.5: 96883772-96883772 on Build GRCh38
rs956158334 Chr.5: 96883793-96883793 on Build GRCh38
rs745754987 Chr.5: 96883774-96883774 on Build GRCh38
rs772210333 Chr.5: 96883953-96883953 on Build GRCh38
rs760051554 Chr.5: 96883831-96883831 on Build GRCh38
rs375419588 Chr.5: 96883825-96883825 on Build GRCh38
rs752188887 Chr.5: 96883920-96883920 on Build GRCh38
rs770813759 Chr.5: 96883766-96883766 on Build GRCh38
rs1038274260 Chr.5: 96883790-96883790 on Build GRCh38

View Full Product Details