Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 14469303-14469528 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COPB1
Entrez Gene ID
1315
Gene Name
coatomer protein complex subunit beta 1
Gene Aliases
COPB
UniGene
Hs.339278
Gene Chromosome Location
Chr.11: 14457503-14499895 on Build GRCh38
Transcripts
NM_001144061.1 NM_001144062.1 NM_016451.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
687131 c.1792T>C p.S598P Chr.11: 14469509-14469509 on Build GRCh38
687132 c.1885G>A p.D629N Chr.11: 14469416-14469416 on Build GRCh38
6847328 c.1963A>T p.K655* Chr.11: 14469338-14469338 on Build GRCh38
1661250 c.1938A>C p.K646N Chr.11: 14469363-14469363 on Build GRCh38
4877149 c.1835G>A p.R612Q Chr.11: 14469466-14469466 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775287362 Chr.11: 14469352-14469354 on Build GRCh38
rs567395905 Chr.11: 14469418-14469418 on Build GRCh38
rs776554381 Chr.11: 14469406-14469406 on Build GRCh38
rs759828340 Chr.11: 14469456-14469456 on Build GRCh38
rs768074101 Chr.11: 14469465-14469465 on Build GRCh38
rs768589584 Chr.11: 14469386-14469386 on Build GRCh38
rs974462035 Chr.11: 14469488-14469488 on Build GRCh38
rs771743899 Chr.11: 14469369-14469369 on Build GRCh38
rs1006041516 Chr.11: 14469466-14469466 on Build GRCh38
rs762703980 Chr.11: 14469432-14469432 on Build GRCh38
rs781534973 Chr.11: 14469332-14469332 on Build GRCh38
rs868250133 Chr.11: 14469387-14469387 on Build GRCh38
rs564234227 Chr.11: 14469378-14469378 on Build GRCh38
rs769349616 Chr.11: 14469419-14469419 on Build GRCh38
rs1016047567 Chr.11: 14469469-14469469 on Build GRCh38
rs147940792 Chr.11: 14469480-14469480 on Build GRCh38
rs144780995 Chr.11: 14469428-14469428 on Build GRCh38
rs778139308 Chr.11: 14469353-14469353 on Build GRCh38
rs779940106 Chr.11: 14469371-14469371 on Build GRCh38
rs751899671 Chr.11: 14469452-14469452 on Build GRCh38
rs757263393 Chr.11: 14469464-14469464 on Build GRCh38
rs765975347 Chr.11: 14469447-14469447 on Build GRCh38
rs749584129 Chr.11: 14469359-14469359 on Build GRCh38

View Full Product Details