Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 44857609-44857882 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF2B3
Entrez Gene ID
8891
Gene Name
eukaryotic translation initiation factor 2B subunit gamma
Gene Aliases
EIF-2B, EIF2Bgamma
UniGene
Hs.533549
Gene Chromosome Location
Chr.1: 44850522-44986722 on Build GRCh38
Transcripts
NM_001261418.1 NM_020365.4 XM_017002747.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6389016 c.1228T>C p.C410R Chr.1: 44857782-44857782 on Build GRCh38
1343020 c.1306G>T p.A436S Chr.1: 44857704-44857704 on Build GRCh38
3419270 c.1245C>T p.I415I Chr.1: 44857765-44857765 on Build GRCh38
6126630 c.1237G>T p.A413S Chr.1: 44857773-44857773 on Build GRCh38
910092 c.1295T>G p.I432S Chr.1: 44857715-44857715 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762959486 Chr.1: 44857733-44857733 on Build GRCh38
rs765453086 Chr.1: 44857721-44857721 on Build GRCh38
rs770622953 Chr.1: 44857802-44857802 on Build GRCh38
rs886046358 Chr.1: 44857723-44857723 on Build GRCh38
rs960555866 Chr.1: 44857633-44857633 on Build GRCh38
rs772495393 Chr.1: 44857731-44857731 on Build GRCh38
rs563359395 Chr.1: 44857816-44857816 on Build GRCh38
rs755755644 Chr.1: 44857775-44857775 on Build GRCh38
rs748821477 Chr.1: 44857797-44857797 on Build GRCh38
rs751561316 Chr.1: 44857749-44857749 on Build GRCh38
rs139902464 Chr.1: 44857788-44857788 on Build GRCh38
rs529847913 Chr.1: 44857764-44857764 on Build GRCh38
rs919028678 Chr.1: 44857822-44857822 on Build GRCh38
rs762168275 Chr.1: 44857847-44857847 on Build GRCh38
rs776084152 Chr.1: 44857676-44857676 on Build GRCh38
rs754879236 Chr.1: 44857763-44857763 on Build GRCh38
rs376165243 Chr.1: 44857842-44857842 on Build GRCh38
rs761993572 Chr.1: 44857698-44857698 on Build GRCh38
rs547795033 Chr.1: 44857765-44857765 on Build GRCh38
rs750471493 Chr.1: 44857727-44857727 on Build GRCh38
rs769214135 Chr.1: 44857692-44857692 on Build GRCh38
rs369355291 Chr.1: 44857846-44857846 on Build GRCh38
rs772511334 Chr.1: 44857845-44857845 on Build GRCh38
rs532129358 Chr.1: 44857855-44857855 on Build GRCh38
rs201617348 Chr.1: 44857781-44857781 on Build GRCh38
rs773739308 Chr.1: 44857826-44857826 on Build GRCh38
rs1051418199 Chr.1: 44857835-44857835 on Build GRCh38
rs777419400 Chr.1: 44857793-44857793 on Build GRCh38
rs902143849 Chr.1: 44857772-44857772 on Build GRCh38
rs761251836 Chr.1: 44857680-44857680 on Build GRCh38
rs777026699 Chr.1: 44857696-44857696 on Build GRCh38
rs766429223 Chr.1: 44857738-44857738 on Build GRCh38
rs553737269 Chr.1: 44857758-44857758 on Build GRCh38
rs943279992 Chr.1: 44857852-44857852 on Build GRCh38

View Full Product Details