Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 2109920-2110177 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP3D1
Entrez Gene ID
8943
Gene Name
adaptor related protein complex 3 delta 1 subunit
Gene Aliases
ADTD, hBLVR
UniGene
Hs.512815
Gene Chromosome Location
Chr.19: 2100988-2151620 on Build GRCh38
Transcripts
NM_001261826.1 NM_003938.6 XM_006722932.2 XM_017027422.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3531288 c.3265-1G>A p.? Chr.19: 2109959-2109959 on Build GRCh38
4280409 c.2931G>T p.K977N Chr.19: 2110160-2110160 on Build GRCh38
4280407 c.2969C>T p.A990V Chr.19: 2109945-2109945 on Build GRCh38
4784299 c.3242G>A p.G1081E Chr.19: 2110158-2110158 on Build GRCh38
3531286 c.3079-1G>A p.? Chr.19: 2109959-2109959 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767578595 Chr.19: 2110051-2110051 on Build GRCh38
rs763764314 Chr.19: 2109952-2109952 on Build GRCh38
rs751610165 Chr.19: 2110102-2110102 on Build GRCh38
rs566400421 Chr.19: 2109964-2109964 on Build GRCh38
rs776324869 Chr.19: 2110124-2110124 on Build GRCh38
rs374725507 Chr.19: 2109953-2109953 on Build GRCh38
rs773919127 Chr.19: 2109982-2109982 on Build GRCh38
rs367951579 Chr.19: 2110076-2110076 on Build GRCh38
rs368539081 Chr.19: 2110106-2110106 on Build GRCh38
rs938929196 Chr.19: 2110149-2110149 on Build GRCh38
rs547984913 Chr.19: 2109943-2109943 on Build GRCh38
rs774897782 Chr.19: 2109998-2109998 on Build GRCh38
rs766273969 Chr.19: 2110153-2110153 on Build GRCh38
rs760186162 Chr.19: 2110000-2110000 on Build GRCh38
rs367889389 Chr.19: 2109983-2109983 on Build GRCh38
rs555224621 Chr.19: 2110121-2110121 on Build GRCh38
rs760252452 Chr.19: 2109947-2109947 on Build GRCh38
rs748569899 Chr.19: 2109976-2109976 on Build GRCh38
rs771406981 Chr.19: 2109997-2109997 on Build GRCh38
rs751818228 Chr.19: 2109983-2109984 on Build GRCh38
rs773020774 Chr.19: 2110140-2110140 on Build GRCh38
rs965662532 Chr.19: 2110050-2110050 on Build GRCh38
rs570328730 Chr.19: 2110070-2110070 on Build GRCh38
rs763704577 Chr.19: 2110005-2110005 on Build GRCh38
rs757494706 Chr.19: 2109990-2109990 on Build GRCh38
rs368379367 Chr.19: 2110114-2110114 on Build GRCh38
rs768790210 Chr.19: 2109969-2109969 on Build GRCh38
rs778290390 Chr.19: 2110110-2110110 on Build GRCh38
rs752474782 Chr.19: 2110086-2110086 on Build GRCh38
rs577459436 Chr.19: 2110103-2110103 on Build GRCh38
rs370612433 Chr.19: 2109945-2109945 on Build GRCh38
rs749627361 Chr.19: 2110112-2110112 on Build GRCh38
rs953823307 Chr.19: 2110077-2110077 on Build GRCh38
rs781643139 Chr.19: 2110004-2110010 on Build GRCh38
rs749831158 Chr.19: 2109995-2109995 on Build GRCh38
rs200617697 Chr.19: 2110157-2110157 on Build GRCh38
rs750356074 Chr.19: 2109962-2109962 on Build GRCh38
rs750159778 Chr.19: 2110088-2110088 on Build GRCh38
rs888418364 Chr.19: 2110006-2110006 on Build GRCh38
rs758271852 Chr.19: 2110094-2110094 on Build GRCh38
rs781424280 Chr.19: 2109970-2109970 on Build GRCh38
rs752197630 Chr.19: 2109944-2109944 on Build GRCh38
rs756654646 Chr.19: 2110107-2110108 on Build GRCh38
rs537890782 Chr.19: 2110071-2110071 on Build GRCh38
rs199856286 Chr.19: 2110115-2110115 on Build GRCh38
rs998065277 Chr.19: 2110056-2110056 on Build GRCh38
rs113684349 Chr.19: 2110109-2110109 on Build GRCh38
rs776237023 Chr.19: 2110008-2110008 on Build GRCh38
rs888836735 Chr.19: 2110125-2110125 on Build GRCh38
rs2074958 Chr.19: 2110020-2110020 on Build GRCh38
rs762759003 Chr.19: 2110144-2110144 on Build GRCh38
rs1015545266 Chr.19: 2110027-2110027 on Build GRCh38
rs778863085 Chr.19: 2109957-2109957 on Build GRCh38
rs77265501 Chr.19: 2109989-2109989 on Build GRCh38
rs199895376 Chr.19: 2110137-2110137 on Build GRCh38
rs746310920 Chr.19: 2110092-2110093 on Build GRCh38
rs770248777 Chr.19: 2109980-2109980 on Build GRCh38
rs781601590 Chr.19: 2110104-2110104 on Build GRCh38
rs71337082 Chr.19: 2110010-2110010 on Build GRCh38
rs534639518 Chr.19: 2109963-2109963 on Build GRCh38
rs761466920 Chr.19: 2110130-2110130 on Build GRCh38
rs374556619 Chr.19: 2110087-2110087 on Build GRCh38
rs766297772 Chr.19: 2110096-2110096 on Build GRCh38
rs894598626 Chr.19: 2109978-2109978 on Build GRCh38
rs375224762 Chr.19: 2110113-2110113 on Build GRCh38

View Full Product Details