Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 13754258-13754483 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC130
Entrez Gene ID
81576
Gene Name
coiled-coil domain containing 130
Gene Aliases
-
UniGene
Hs.24998
Gene Chromosome Location
Chr.19: 13731760-13763298 on Build GRCh38
Transcripts
NM_001320561.1 NM_001320564.1 NM_001320565.1 NM_001320566.1 NM_001320567.1 NM_001320568.1 NM_001320569.1 NM_030818.3 XM_005260086.4 XM_011528326.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4492016 c.38C>T p.P13L Chr.19: 13754323-13754323 on Build GRCh38
3932899 c.52G>C p.E18Q Chr.19: 13754337-13754337 on Build GRCh38
6414977 c.53A>T p.E18V Chr.19: 13754338-13754338 on Build GRCh38
474221 c.51T>C p.P17P Chr.19: 13754336-13754336 on Build GRCh38
2817995 c.52G>A p.E18K Chr.19: 13754337-13754337 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377472319 Chr.19: 13754398-13754398 on Build GRCh38
rs756718507 Chr.19: 13754389-13754389 on Build GRCh38
rs573663025 Chr.19: 13754358-13754358 on Build GRCh38
rs376116418 Chr.19: 13754357-13754357 on Build GRCh38
rs759494804 Chr.19: 13754315-13754315 on Build GRCh38
rs531767273 Chr.19: 13754422-13754422 on Build GRCh38
rs993099376 Chr.19: 13754344-13754345 on Build GRCh38
rs435207 Chr.19: 13754461-13754461 on Build GRCh38
rs886544892 Chr.19: 13754432-13754432 on Build GRCh38
rs922734308 Chr.19: 13754416-13754416 on Build GRCh38
rs200133003 Chr.19: 13754348-13754348 on Build GRCh38
rs200521047 Chr.19: 13754343-13754343 on Build GRCh38
rs772785462 Chr.19: 13754281-13754281 on Build GRCh38
rs776496891 Chr.19: 13754312-13754312 on Build GRCh38
rs771668885 Chr.19: 13754317-13754317 on Build GRCh38
rs964069846 Chr.19: 13754397-13754397 on Build GRCh38
rs760445366 Chr.19: 13754285-13754285 on Build GRCh38
rs766170095 Chr.19: 13754304-13754304 on Build GRCh38
rs1009051762 Chr.19: 13754439-13754439 on Build GRCh38
rs751796069 Chr.19: 13754355-13754355 on Build GRCh38
rs370590169 Chr.19: 13754364-13754364 on Build GRCh38
rs447668 Chr.19: 13754462-13754462 on Build GRCh38
rs35632776 Chr.19: 13754461-13754462 on Build GRCh38
rs756876738 Chr.19: 13754342-13754342 on Build GRCh38
rs1019103657 Chr.19: 13754460-13754460 on Build GRCh38
rs544334278 Chr.19: 13754401-13754401 on Build GRCh38
rs879763378 Chr.19: 13754461-13754462 on Build GRCh38
rs746327603 Chr.19: 13754388-13754388 on Build GRCh38
rs34136555 Chr.19: 13754354-13754354 on Build GRCh38
rs113791690 Chr.19: 13754461-13754461 on Build GRCh38
rs73922726 Chr.19: 13754421-13754421 on Build GRCh38
rs765262615 Chr.19: 13754342-13754342 on Build GRCh38

View Full Product Details