Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 99329456-99329681 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNNM1
Entrez Gene ID
26507
Gene Name
cyclin and CBS domain divalent metal cation transport mediator 1
Gene Aliases
ACDP1, CLP-1
UniGene
Hs.274579
Gene Chromosome Location
Chr.10: 99329099-99394330 on Build GRCh38
Transcripts
NM_020348.2 XM_005269692.3 XM_011539629.1 XM_011539630.1 XM_011539631.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6128266 c.196G>A p.E66K Chr.10: 99329583-99329583 on Build GRCh38
6631290 c.222C>T p.V74V Chr.10: 99329609-99329609 on Build GRCh38
6406718 c.254C>T p.A85V Chr.10: 99329641-99329641 on Build GRCh38
5084099 c.190G>A p.A64T Chr.10: 99329577-99329577 on Build GRCh38
1248563 c.233C>T p.P78L Chr.10: 99329620-99329620 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749661449 Chr.10: 99329621-99329621 on Build GRCh38
rs752414699 Chr.10: 99329660-99329660 on Build GRCh38
rs759052277 Chr.10: 99329644-99329644 on Build GRCh38
rs776207895 Chr.10: 99329643-99329643 on Build GRCh38
rs998906677 Chr.10: 99329653-99329653 on Build GRCh38
rs758246458 Chr.10: 99329570-99329570 on Build GRCh38
rs777684690 Chr.10: 99329577-99329577 on Build GRCh38
rs778611554 Chr.10: 99329554-99329554 on Build GRCh38
rs746041012 Chr.10: 99329597-99329597 on Build GRCh38
rs754689003 Chr.10: 99329549-99329549 on Build GRCh38
rs942126643 Chr.10: 99329522-99329522 on Build GRCh38
rs752502645 Chr.10: 99329562-99329562 on Build GRCh38
rs997732778 Chr.10: 99329627-99329627 on Build GRCh38
rs764854856 Chr.10: 99329657-99329657 on Build GRCh38
rs765765196 Chr.10: 99329641-99329641 on Build GRCh38
rs765961265 Chr.10: 99329495-99329495 on Build GRCh38
rs527685732 Chr.10: 99329490-99329490 on Build GRCh38
rs746396934 Chr.10: 99329664-99329664 on Build GRCh38
rs569360605 Chr.10: 99329580-99329580 on Build GRCh38
rs966791673 Chr.10: 99329498-99329498 on Build GRCh38
rs746988712 Chr.10: 99329579-99329579 on Build GRCh38
rs774974042 Chr.10: 99329629-99329629 on Build GRCh38
rs775943402 Chr.10: 99329605-99329605 on Build GRCh38
rs900673029 Chr.10: 99329603-99329603 on Build GRCh38
rs551214160 Chr.10: 99329542-99329542 on Build GRCh38
rs374753323 Chr.10: 99329601-99329601 on Build GRCh38
rs762401365 Chr.10: 99329638-99329638 on Build GRCh38
rs769179313 Chr.10: 99329626-99329626 on Build GRCh38

View Full Product Details