Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 49530949-49531206 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DAG1
Entrez Gene ID
1605
Gene Name
dystroglycan 1
Gene Aliases
156DAG, A3a, AGRNR, DAG, MDDGA9, MDDGC7, MDDGC9
UniGene
Hs.76111
Gene Chromosome Location
Chr.3: 49468703-49535618 on Build GRCh38
Transcripts
NM_001165928.3 NM_001177634.2 NM_001177635.2 NM_001177636.2 NM_001177637.2 NM_001177638.2 NM_001177639.2 NM_001177640.2 NM_001177641.2 NM_001177642.2 NM_001177643.2 NM_001177644.2 NM_004393.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4118503 c.497C>T p.S166L Chr.3: 49531008-49531008 on Build GRCh38
5978639 c.568G>C p.V190L Chr.3: 49531079-49531079 on Build GRCh38
4118504 c.643C>T p.R215W Chr.3: 49531154-49531154 on Build GRCh38
77555 c.682G>T p.V228L Chr.3: 49531193-49531193 on Build GRCh38
1309271 c.655G>A p.E219K Chr.3: 49531166-49531166 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749461367 Chr.3: 49530974-49530974 on Build GRCh38
rs529287786 Chr.3: 49531180-49531180 on Build GRCh38
rs746894568 Chr.3: 49531087-49531087 on Build GRCh38
rs199654893 Chr.3: 49530973-49530973 on Build GRCh38
rs757690278 Chr.3: 49531065-49531065 on Build GRCh38
rs750166378 Chr.3: 49531031-49531031 on Build GRCh38
rs545200027 Chr.3: 49531079-49531079 on Build GRCh38
rs775575894 Chr.3: 49531082-49531082 on Build GRCh38
rs368359780 Chr.3: 49530984-49530984 on Build GRCh38
rs765923041 Chr.3: 49531147-49531147 on Build GRCh38
rs772976299 Chr.3: 49531134-49531134 on Build GRCh38
rs193922955 Chr.3: 49531086-49531086 on Build GRCh38
rs763906245 Chr.3: 49531014-49531014 on Build GRCh38
rs201118488 Chr.3: 49531120-49531120 on Build GRCh38
rs41290704 Chr.3: 49531110-49531110 on Build GRCh38
rs757948629 Chr.3: 49531035-49531035 on Build GRCh38
rs745947173 Chr.3: 49530991-49530991 on Build GRCh38
rs144753260 Chr.3: 49531155-49531155 on Build GRCh38
rs894480547 Chr.3: 49531141-49531141 on Build GRCh38
rs766034304 Chr.3: 49531042-49531042 on Build GRCh38
rs200094365 Chr.3: 49531118-49531118 on Build GRCh38
rs201869343 Chr.3: 49531046-49531046 on Build GRCh38
rs146574353 Chr.3: 49531145-49531145 on Build GRCh38
rs794727500 Chr.3: 49531009-49531009 on Build GRCh38
rs764969676 Chr.3: 49531025-49531025 on Build GRCh38
rs145540662 Chr.3: 49531070-49531070 on Build GRCh38
rs890598451 Chr.3: 49531122-49531122 on Build GRCh38
rs907269872 Chr.3: 49531162-49531162 on Build GRCh38
rs556413209 Chr.3: 49531008-49531008 on Build GRCh38
rs749602970 Chr.3: 49531063-49531063 on Build GRCh38
rs562046526 Chr.3: 49531154-49531154 on Build GRCh38
rs768487898 Chr.3: 49531102-49531102 on Build GRCh38
rs147153370 Chr.3: 49531021-49531021 on Build GRCh38
rs754413131 Chr.3: 49531183-49531183 on Build GRCh38
rs148393291 Chr.3: 49530982-49530982 on Build GRCh38
rs778142627 Chr.3: 49531061-49531061 on Build GRCh38
rs760549124 Chr.3: 49531012-49531012 on Build GRCh38
rs145871499 Chr.3: 49531124-49531124 on Build GRCh38
rs756584618 Chr.3: 49531050-49531050 on Build GRCh38
rs772182851 Chr.3: 49530999-49530999 on Build GRCh38
rs998718075 Chr.3: 49531170-49531170 on Build GRCh38
rs1045779842 Chr.3: 49531159-49531159 on Build GRCh38

View Full Product Details