Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 159325760-159326019 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BAZ2B
Entrez Gene ID
29994
Gene Name
bromodomain adjacent to zinc finger domain 2B
Gene Aliases
WALp4
UniGene
Hs.470369
Gene Chromosome Location
Chr.2: 159315312-159712439 on Build GRCh38
Transcripts
NM_001289975.1 NM_013450.3 XM_005246488.2 XM_005246489.4 XM_005246492.4 XM_006712461.3 XM_011511038.2 XM_011511039.1 XM_011511042.1 XM_011511043.1 XM_011511044.1 XM_011511045.1 XM_011511047.1 XM_011511048.2 XM_011511050.1 XM_011511051.1 XM_011511052.1 XM_011511056.1 XM_017003918.1 XM_017003919.1 XM_017003920.1 XM_017003921.1 XM_017003922.1 XM_017003923.1 XM_017003924.1 XM_017003925.1 XM_017003926.1 XM_017003927.1 XM_017003928.1 XM_017003929.1 XM_017003930.1 XM_017003931.1 XM_017003932.1 XM_017003933.1 XM_017003934.1 XM_017003935.1 XM_017003936.1 XM_017003937.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6563207 c.6056C>T p.S2019F Chr.2: 159325806-159325806 on Build GRCh38
6431038 c.6013G>A p.G2005S Chr.2: 159325849-159325849 on Build GRCh38
4554081 c.6102G>A p.K2034K Chr.2: 159325760-159325760 on Build GRCh38
3743928 c.6067A>G p.S2023G Chr.2: 159325795-159325795 on Build GRCh38
3736217 c.5999A>G p.N2000S Chr.2: 159325863-159325863 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779496770 Chr.2: 159325950-159325950 on Build GRCh38
rs765295946 Chr.2: 159325927-159325928 on Build GRCh38
rs999475360 Chr.2: 159325981-159325981 on Build GRCh38
rs750092971 Chr.2: 159325880-159325880 on Build GRCh38
rs779049002 Chr.2: 159325953-159325954 on Build GRCh38
rs748956349 Chr.2: 159325827-159325827 on Build GRCh38
rs778547353 Chr.2: 159325837-159325837 on Build GRCh38
rs772671668 Chr.2: 159325848-159325848 on Build GRCh38
rs926935507 Chr.2: 159325908-159325908 on Build GRCh38
rs751468658 Chr.2: 159325937-159325937 on Build GRCh38
rs964709554 Chr.2: 159325805-159325805 on Build GRCh38
rs373982448 Chr.2: 159325856-159325856 on Build GRCh38
rs760992884 Chr.2: 159325788-159325788 on Build GRCh38
rs532331487 Chr.2: 159325804-159325804 on Build GRCh38
rs369418579 Chr.2: 159325814-159325814 on Build GRCh38
rs747518402 Chr.2: 159325951-159325951 on Build GRCh38
rs749995117 Chr.2: 159325942-159325942 on Build GRCh38
rs778395615 Chr.2: 159325930-159325930 on Build GRCh38
rs777672179 Chr.2: 159325822-159325822 on Build GRCh38
rs895517019 Chr.2: 159325987-159325987 on Build GRCh38
rs761866580 Chr.2: 159325928-159325928 on Build GRCh38
rs745369584 Chr.2: 159325841-159325841 on Build GRCh38
rs752713978 Chr.2: 159325888-159325888 on Build GRCh38
rs767454873 Chr.2: 159325946-159325946 on Build GRCh38
rs415793 Chr.2: 159325791-159325791 on Build GRCh38
rs547598843 Chr.2: 159325819-159325819 on Build GRCh38
rs966656926 Chr.2: 159325958-159325958 on Build GRCh38
rs750412559 Chr.2: 159325798-159325798 on Build GRCh38
rs554369554 Chr.2: 159325928-159325928 on Build GRCh38
rs776822333 Chr.2: 159325802-159325803 on Build GRCh38
rs756804716 Chr.2: 159325834-159325834 on Build GRCh38
rs765363163 Chr.2: 159325878-159325878 on Build GRCh38
rs200299773 Chr.2: 159325866-159325866 on Build GRCh38
rs549709911 Chr.2: 159325797-159325797 on Build GRCh38
rs1056818697 Chr.2: 159325932-159325932 on Build GRCh38
rs148382881 Chr.2: 159325863-159325863 on Build GRCh38
rs763996775 Chr.2: 159325892-159325892 on Build GRCh38
rs376860541 Chr.2: 159325862-159325862 on Build GRCh38
rs769091896 Chr.2: 159325966-159325966 on Build GRCh38
rs764345917 Chr.2: 159325790-159325790 on Build GRCh38
rs767326540 Chr.2: 159325807-159325807 on Build GRCh38
rs747463243 Chr.2: 159325859-159325859 on Build GRCh38
rs913097416 Chr.2: 159325877-159325877 on Build GRCh38
rs756052909 Chr.2: 159325890-159325890 on Build GRCh38
rs761982232 Chr.2: 159325793-159325793 on Build GRCh38
rs753677056 Chr.2: 159325927-159325927 on Build GRCh38

View Full Product Details