Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 31422742-31423015 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DROSHA
Entrez Gene ID
29102
Gene Name
drosha ribonuclease III
Gene Aliases
ETOHI2, HSA242976, RANSE3L, RN3, RNASE3L, RNASEN
UniGene
Hs.97997
Gene Chromosome Location
Chr.5: 31400494-31532175 on Build GRCh38
Transcripts
NM_001100412.1 NM_013235.4 XM_005248291.3 XM_005248292.3 XM_005248293.3 XM_005248294.3 XM_011514033.2 XM_017009399.1 XM_017009400.1 XM_017009401.1 XM_017009402.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6193025 c.3302C>A p.S1101Y Chr.5: 31422904-31422904 on Build GRCh38
3429371 c.3284G>A p.R1095Q Chr.5: 31422922-31422922 on Build GRCh38
3014375 c.3295G>T p.E1099* Chr.5: 31422911-31422911 on Build GRCh38
5466430 c.3363T>C p.V1121V Chr.5: 31422843-31422843 on Build GRCh38
3615357 c.3361G>T p.V1121F Chr.5: 31422845-31422845 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747258421 Chr.5: 31422829-31422829 on Build GRCh38
rs374888259 Chr.5: 31422936-31422936 on Build GRCh38
rs2287584 Chr.5: 31422900-31422900 on Build GRCh38
rs748009983 Chr.5: 31422902-31422902 on Build GRCh38
rs371709358 Chr.5: 31422930-31422930 on Build GRCh38
rs1006321001 Chr.5: 31422894-31422894 on Build GRCh38
rs769711908 Chr.5: 31422869-31422869 on Build GRCh38
rs375079038 Chr.5: 31422849-31422849 on Build GRCh38
rs767129327 Chr.5: 31422957-31422957 on Build GRCh38
rs755701109 Chr.5: 31422984-31422984 on Build GRCh38
rs776736153 Chr.5: 31422865-31422865 on Build GRCh38
rs748062041 Chr.5: 31422866-31422866 on Build GRCh38
rs758695166 Chr.5: 31422794-31422794 on Build GRCh38
rs780286427 Chr.5: 31422819-31422819 on Build GRCh38
rs1012494996 Chr.5: 31422956-31422956 on Build GRCh38
rs577273045 Chr.5: 31422964-31422964 on Build GRCh38
rs768751793 Chr.5: 31422770-31422770 on Build GRCh38
rs762765224 Chr.5: 31422901-31422901 on Build GRCh38
rs759300980 Chr.5: 31422949-31422949 on Build GRCh38
rs368160941 Chr.5: 31422778-31422778 on Build GRCh38

View Full Product Details